Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119885663-119886190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN4
Entrez Gene ID
5775
Gene Name
protein tyrosine phosphatase, non-receptor type 4
Gene Aliases
MEG, PTPMEG, PTPMEG1
UniGene
Hs.469809
Gene Chromosome Location
Chr.2: 119759631-119984899 on Build GRCh38
Transcripts
NM_002830.3 XM_011511560.2 XM_011511562.2 XM_017004596.1 XM_017004597.1 XM_017004598.1 XM_017004599.1 XM_017004600.1 XM_017004601.1 XM_017004602.1 XM_017004603.1 XM_017004604.1 XM_017004605.1 XM_017004606.1 XM_017004607.1 XM_017004608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4154495 c.630A>T p.T210T Chr.2: 119885837-119885837 on Build GRCh38
1291289 c.612A>G p.E204E Chr.2: 119885819-119885819 on Build GRCh38
5859994 c.670G>A p.A224T Chr.2: 119885877-119885877 on Build GRCh38
4084519 c.635G>A p.R212H Chr.2: 119885842-119885842 on Build GRCh38
716042 c.653G>C p.G218A Chr.2: 119885860-119885860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533758568 Chr.2: 119886051-119886051 on Build GRCh38
rs770717048 Chr.2: 119885762-119885762 on Build GRCh38
rs1014398809 Chr.2: 119886033-119886033 on Build GRCh38
rs944758468 Chr.2: 119885742-119885742 on Build GRCh38
rs932023046 Chr.2: 119885702-119885702 on Build GRCh38
rs780299990 Chr.2: 119886054-119886054 on Build GRCh38
rs548914986 Chr.2: 119885739-119885739 on Build GRCh38
rs749842506 Chr.2: 119885967-119885967 on Build GRCh38
rs899783430 Chr.2: 119885752-119885752 on Build GRCh38
rs187466061 Chr.2: 119886141-119886141 on Build GRCh38
rs1004160166 Chr.2: 119886074-119886074 on Build GRCh38
rs565256787 Chr.2: 119886118-119886118 on Build GRCh38
rs779166189 Chr.2: 119885842-119885842 on Build GRCh38
rs1015767607 Chr.2: 119885995-119885995 on Build GRCh38
rs868358880 Chr.2: 119886132-119886132 on Build GRCh38
rs112079258 Chr.2: 119886131-119886131 on Build GRCh38
rs74866537 Chr.2: 119885704-119885705 on Build GRCh38
rs11892365 Chr.2: 119885850-119885850 on Build GRCh38
rs759307375 Chr.2: 119885733-119885733 on Build GRCh38
rs373958913 Chr.2: 119885908-119885908 on Build GRCh38
rs767733787 Chr.2: 119885819-119885819 on Build GRCh38
rs558314253 Chr.2: 119886123-119886123 on Build GRCh38
rs1036549163 Chr.2: 119885796-119885796 on Build GRCh38
rs376525667 Chr.2: 119885857-119885857 on Build GRCh38
rs767189237 Chr.2: 119885785-119885785 on Build GRCh38
rs772624067 Chr.2: 119885751-119885751 on Build GRCh38
rs986107710 Chr.2: 119886104-119886104 on Build GRCh38
rs944703992 Chr.2: 119886124-119886124 on Build GRCh38
rs758809995 Chr.2: 119885875-119885875 on Build GRCh38
rs757271269 Chr.2: 119885840-119885840 on Build GRCh38
rs911944498 Chr.2: 119885723-119885723 on Build GRCh38
rs752733102 Chr.2: 119885791-119885791 on Build GRCh38
rs769043780 Chr.2: 119885920-119885920 on Build GRCh38
rs567204388 Chr.2: 119885925-119885925 on Build GRCh38
rs555300562 Chr.2: 119886062-119886062 on Build GRCh38
rs959646609 Chr.2: 119886099-119886099 on Build GRCh38
rs796195760 Chr.2: 119885747-119885748 on Build GRCh38
rs888130228 Chr.2: 119885924-119885924 on Build GRCh38
rs911984063 Chr.2: 119885735-119885735 on Build GRCh38
rs536900151 Chr.2: 119885726-119885726 on Build GRCh38
rs762482436 Chr.2: 119885760-119885760 on Build GRCh38
rs555891212 Chr.2: 119886165-119886165 on Build GRCh38
rs750608861 Chr.2: 119885759-119885760 on Build GRCh38
rs370609847 Chr.2: 119885901-119885901 on Build GRCh38
rs764053345 Chr.2: 119885829-119885829 on Build GRCh38
rs770563499 Chr.2: 119885926-119885926 on Build GRCh38
rs758664911 Chr.2: 119885899-119885900 on Build GRCh38
rs759472967 Chr.2: 119885782-119885782 on Build GRCh38
rs959792798 Chr.2: 119885968-119885968 on Build GRCh38
rs774131346 Chr.2: 119885780-119885780 on Build GRCh38
rs925874515 Chr.2: 119886063-119886063 on Build GRCh38
rs371599364 Chr.2: 119885932-119885932 on Build GRCh38
rs760556953 Chr.2: 119885822-119885822 on Build GRCh38
rs570515802 Chr.2: 119886128-119886128 on Build GRCh38
rs754376689 Chr.2: 119885747-119885748 on Build GRCh38
rs753868954 Chr.2: 119885833-119885833 on Build GRCh38
rs953261257 Chr.2: 119886094-119886094 on Build GRCh38
rs1020014487 Chr.2: 119886040-119886040 on Build GRCh38
rs757509202 Chr.2: 119885748-119885751 on Build GRCh38

View Full Product Details