Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17511722-17512213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR2
Entrez Gene ID
27443
Gene Name
CECR2, histone acetyl-lysine reader
Gene Aliases
-
UniGene
Hs.231895
Gene Chromosome Location
Chr.22: 17359949-17558155 on Build GRCh38
Transcripts
NM_001290046.1 NM_001290047.1 XM_011546128.2 XM_011546129.2 XM_011546132.2 XM_017028785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1291205 c.754C>T p.R252C Chr.22: 17511819-17511819 on Build GRCh38
1318792 c.877G>A p.V293I Chr.22: 17511885-17511885 on Build GRCh38
1592841 c.757C>A p.P253T Chr.22: 17511822-17511822 on Build GRCh38
1647465 c.800A>G p.D267G Chr.22: 17511865-17511865 on Build GRCh38
1031848 c.814C>A p.P272T Chr.22: 17511822-17511822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372166419 Chr.22: 17511828-17511828 on Build GRCh38
rs183665956 Chr.22: 17511904-17511904 on Build GRCh38
rs918340903 Chr.22: 17511745-17511745 on Build GRCh38
rs375479676 Chr.22: 17511946-17511946 on Build GRCh38
rs961774183 Chr.22: 17511992-17511992 on Build GRCh38
rs761062236 Chr.22: 17511911-17511911 on Build GRCh38
rs369053722 Chr.22: 17511884-17511884 on Build GRCh38
rs774377462 Chr.22: 17511812-17511812 on Build GRCh38
rs1012368886 Chr.22: 17512114-17512114 on Build GRCh38
rs750763487 Chr.22: 17511846-17511846 on Build GRCh38
rs754382702 Chr.22: 17511854-17511854 on Build GRCh38
rs748289376 Chr.22: 17511883-17511883 on Build GRCh38
rs1025278743 Chr.22: 17512188-17512188 on Build GRCh38
rs776452695 Chr.22: 17511821-17511821 on Build GRCh38
rs983958716 Chr.22: 17511763-17511763 on Build GRCh38
rs755549503 Chr.22: 17511923-17511923 on Build GRCh38
rs935435780 Chr.22: 17512101-17512101 on Build GRCh38
rs779115433 Chr.22: 17511882-17511882 on Build GRCh38
rs768785705 Chr.22: 17511810-17511810 on Build GRCh38
rs149739505 Chr.22: 17511829-17511829 on Build GRCh38
rs763676410 Chr.22: 17511833-17511833 on Build GRCh38
rs775023684 Chr.22: 17511913-17511913 on Build GRCh38
rs5747211 Chr.22: 17511820-17511820 on Build GRCh38
rs369496060 Chr.22: 17511839-17511839 on Build GRCh38
rs763008744 Chr.22: 17511901-17511901 on Build GRCh38
rs1041068666 Chr.22: 17512083-17512083 on Build GRCh38
rs145660179 Chr.22: 17512026-17512026 on Build GRCh38
rs370117659 Chr.22: 17511918-17511918 on Build GRCh38
rs906782637 Chr.22: 17512140-17512140 on Build GRCh38
rs972874385 Chr.22: 17511992-17511992 on Build GRCh38
rs372506978 Chr.22: 17511888-17511888 on Build GRCh38
rs1014595675 Chr.22: 17511991-17511991 on Build GRCh38
rs762920073 Chr.22: 17511790-17511791 on Build GRCh38
rs770566419 Chr.22: 17511806-17511806 on Build GRCh38
rs571610441 Chr.22: 17511980-17511980 on Build GRCh38
rs757509083 Chr.22: 17511766-17511766 on Build GRCh38
rs940161101 Chr.22: 17511831-17511831 on Build GRCh38
rs374911807 Chr.22: 17511850-17511850 on Build GRCh38
rs895280027 Chr.22: 17512111-17512111 on Build GRCh38
rs1039061518 Chr.22: 17511792-17511792 on Build GRCh38
rs999372501 Chr.22: 17511761-17511761 on Build GRCh38
rs567714852 Chr.22: 17512115-17512115 on Build GRCh38
rs759447732 Chr.22: 17511823-17511823 on Build GRCh38
rs966558480 Chr.22: 17511758-17511758 on Build GRCh38
rs981758638 Chr.22: 17511991-17511992 on Build GRCh38
rs61745636 Chr.22: 17511843-17511843 on Build GRCh38
rs944019383 Chr.22: 17512079-17512079 on Build GRCh38
rs560633584 Chr.22: 17511770-17511770 on Build GRCh38
rs141454025 Chr.22: 17512073-17512073 on Build GRCh38
rs994263273 Chr.22: 17512146-17512146 on Build GRCh38
rs893706161 Chr.22: 17511956-17511956 on Build GRCh38
rs1054372399 Chr.22: 17512106-17512106 on Build GRCh38
rs766906272 Chr.22: 17511917-17511917 on Build GRCh38
rs893662496 Chr.22: 17512117-17512117 on Build GRCh38
rs117314088 Chr.22: 17511935-17511935 on Build GRCh38
rs990890185 Chr.22: 17511799-17511799 on Build GRCh38
rs922505169 Chr.22: 17512002-17512002 on Build GRCh38
rs746344040 Chr.22: 17511774-17511774 on Build GRCh38
rs745626121 Chr.22: 17511886-17511886 on Build GRCh38
rs1042372198 Chr.22: 17512015-17512015 on Build GRCh38
rs528991283 Chr.22: 17511905-17511905 on Build GRCh38
rs972881063 Chr.22: 17511834-17511834 on Build GRCh38
rs200014459 Chr.22: 17511872-17511872 on Build GRCh38
rs767235260 Chr.22: 17511906-17511906 on Build GRCh38
rs780724878 Chr.22: 17511801-17511801 on Build GRCh38
rs933344713 Chr.22: 17512012-17512012 on Build GRCh38
rs780640836 Chr.22: 17511851-17511851 on Build GRCh38
rs373328675 Chr.22: 17511945-17511945 on Build GRCh38
rs902597189 Chr.22: 17512092-17512092 on Build GRCh38
rs1012509947 Chr.22: 17512134-17512134 on Build GRCh38
rs376311056 Chr.22: 17511803-17511803 on Build GRCh38
rs985755332 Chr.22: 17512056-17512059 on Build GRCh38
rs528325498 Chr.22: 17511942-17511942 on Build GRCh38
rs1023701761 Chr.22: 17511965-17511965 on Build GRCh38
rs777993980 Chr.22: 17511885-17511885 on Build GRCh38
rs775235194 Chr.22: 17511889-17511889 on Build GRCh38
rs750103297 Chr.22: 17511909-17511909 on Build GRCh38
rs1012421246 Chr.22: 17511858-17511858 on Build GRCh38
rs748458393 Chr.22: 17511818-17511818 on Build GRCh38
rs55670111 Chr.22: 17511747-17511747 on Build GRCh38
rs942593882 Chr.22: 17511958-17511958 on Build GRCh38
rs1003125038 Chr.22: 17512169-17512169 on Build GRCh38
rs749175883 Chr.22: 17511787-17511787 on Build GRCh38
rs763385948 Chr.22: 17512121-17512121 on Build GRCh38
rs763956084 Chr.22: 17511838-17511839 on Build GRCh38

View Full Product Details