Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20117417-20117920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP1
Entrez Gene ID
5902
Gene Name
RAN binding protein 1
Gene Aliases
HTF9A
UniGene
Hs.24763
Gene Chromosome Location
Chr.22: 20115938-20127357 on Build GRCh38
Transcripts
NM_001278639.1 NM_001278640.1 NM_002882.3 XM_011530289.2 XM_011530290.2 XM_011530291.2 XM_017028890.1 XM_017028892.1

SNP Detail

SNP ID Chromosome Location
rs1040350697 Chr.22: 20117629-20117629 on Build GRCh38
rs989283430 Chr.22: 20117616-20117616 on Build GRCh38
rs1024997125 Chr.22: 20117575-20117575 on Build GRCh38
rs901058964 Chr.22: 20117499-20117499 on Build GRCh38
rs2904566 Chr.22: 20117502-20117502 on Build GRCh38
rs1005336523 Chr.22: 20117681-20117681 on Build GRCh38
rs913259257 Chr.22: 20117621-20117621 on Build GRCh38
rs921927380 Chr.22: 20117876-20117876 on Build GRCh38
rs1033163650 Chr.22: 20117815-20117815 on Build GRCh38
rs980789624 Chr.22: 20117471-20117471 on Build GRCh38
rs745768671 Chr.22: 20117619-20117619 on Build GRCh38
rs756201278 Chr.22: 20117849-20117849 on Build GRCh38
rs767381766 Chr.22: 20117627-20117629 on Build GRCh38
rs576822287 Chr.22: 20117550-20117550 on Build GRCh38
rs927971553 Chr.22: 20117689-20117689 on Build GRCh38
rs541553781 Chr.22: 20117851-20117851 on Build GRCh38
rs542523991 Chr.22: 20117633-20117633 on Build GRCh38
rs555759275 Chr.22: 20117705-20117705 on Build GRCh38
rs750187557 Chr.22: 20117627-20117632 on Build GRCh38
rs939348107 Chr.22: 20117693-20117693 on Build GRCh38
rs564431389 Chr.22: 20117866-20117866 on Build GRCh38
rs556764880 Chr.22: 20117496-20117496 on Build GRCh38
rs370043493 Chr.22: 20117706-20117706 on Build GRCh38
rs926115025 Chr.22: 20117589-20117589 on Build GRCh38
rs888265731 Chr.22: 20117654-20117654 on Build GRCh38
rs1049629968 Chr.22: 20117638-20117642 on Build GRCh38
rs1028118632 Chr.22: 20117524-20117524 on Build GRCh38
rs970316658 Chr.22: 20117767-20117767 on Build GRCh38
rs936126905 Chr.22: 20117605-20117605 on Build GRCh38
rs976495079 Chr.22: 20117873-20117873 on Build GRCh38
rs533437495 Chr.22: 20117878-20117878 on Build GRCh38
rs901091152 Chr.22: 20117630-20117630 on Build GRCh38
rs865834622 Chr.22: 20117543-20117543 on Build GRCh38
rs1005305459 Chr.22: 20117545-20117545 on Build GRCh38
rs914139491 Chr.22: 20117822-20117822 on Build GRCh38
rs980318603 Chr.22: 20117769-20117769 on Build GRCh38
rs975556332 Chr.22: 20117454-20117454 on Build GRCh38
rs1014341618 Chr.22: 20117707-20117707 on Build GRCh38
rs921663870 Chr.22: 20117460-20117460 on Build GRCh38
rs988827483 Chr.22: 20117697-20117697 on Build GRCh38
rs545609133 Chr.22: 20117887-20117887 on Build GRCh38
rs992478341 Chr.22: 20117565-20117565 on Build GRCh38
rs868811636 Chr.22: 20117842-20117842 on Build GRCh38
rs1023940979 Chr.22: 20117729-20117729 on Build GRCh38
rs996699226 Chr.22: 20117514-20117514 on Build GRCh38
rs945335829 Chr.22: 20117482-20117482 on Build GRCh38
rs980246928 Chr.22: 20117585-20117585 on Build GRCh38
rs970244757 Chr.22: 20117584-20117584 on Build GRCh38
rs961321439 Chr.22: 20117564-20117564 on Build GRCh38
rs1015773940 Chr.22: 20117688-20117688 on Build GRCh38
rs540044660 Chr.22: 20117440-20117440 on Build GRCh38
rs778045025 Chr.22: 20117601-20117601 on Build GRCh38
rs867809433 Chr.22: 20117648-20117648 on Build GRCh38
rs913322937 Chr.22: 20117870-20117870 on Build GRCh38
rs767423266 Chr.22: 20117879-20117879 on Build GRCh38
rs888233221 Chr.22: 20117533-20117533 on Build GRCh38
rs779980115 Chr.22: 20117668-20117668 on Build GRCh38
rs896880067 Chr.22: 20117699-20117699 on Build GRCh38
rs933099845 Chr.22: 20117481-20117481 on Build GRCh38
rs757536134 Chr.22: 20117883-20117883 on Build GRCh38
rs374944780 Chr.22: 20117706-20117707 on Build GRCh38
rs1041344837 Chr.22: 20117494-20117494 on Build GRCh38

View Full Product Details