Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170109155-170109655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXI1
Entrez Gene ID
2299
Gene Name
forkhead box I1
Gene Aliases
FKH10, FKHL10, FREAC-6, FREAC6, HFH-3, HFH3
UniGene
Hs.87236
Gene Chromosome Location
Chr.5: 170105897-170109725 on Build GRCh38
Transcripts
NM_012188.4 NM_144769.2

SNP Detail

SNP ID Chromosome Location
rs774087435 Chr.5: 170109410-170109410 on Build GRCh38
rs386405655 Chr.5: 170109543-170109544 on Build GRCh38
rs928192477 Chr.5: 170109374-170109374 on Build GRCh38
rs978343342 Chr.5: 170109370-170109370 on Build GRCh38
rs932064307 Chr.5: 170109314-170109314 on Build GRCh38
rs915099050 Chr.5: 170109372-170109372 on Build GRCh38
rs869204485 Chr.5: 170109550-170109551 on Build GRCh38
rs966422889 Chr.5: 170109290-170109290 on Build GRCh38
rs1005032697 Chr.5: 170109526-170109526 on Build GRCh38
rs35964510 Chr.5: 170109549-170109550 on Build GRCh38
rs1028358335 Chr.5: 170109466-170109466 on Build GRCh38
rs867055048 Chr.5: 170109525-170109525 on Build GRCh38
rs1012360656 Chr.5: 170109222-170109222 on Build GRCh38
rs200357696 Chr.5: 170109215-170109215 on Build GRCh38
rs778213515 Chr.5: 170109555-170109555 on Build GRCh38
rs1011952980 Chr.5: 170109495-170109495 on Build GRCh38
rs571081281 Chr.5: 170109552-170109552 on Build GRCh38
rs1011016906 Chr.5: 170109516-170109516 on Build GRCh38
rs111876035 Chr.5: 170109597-170109597 on Build GRCh38
rs939010562 Chr.5: 170109431-170109431 on Build GRCh38
rs543346264 Chr.5: 170109224-170109224 on Build GRCh38
rs961021431 Chr.5: 170109542-170109543 on Build GRCh38
rs959189720 Chr.5: 170109204-170109204 on Build GRCh38
rs976150354 Chr.5: 170109306-170109306 on Build GRCh38
rs563255429 Chr.5: 170109283-170109283 on Build GRCh38
rs946590294 Chr.5: 170109381-170109381 on Build GRCh38
rs894853030 Chr.5: 170109473-170109473 on Build GRCh38
rs1042313630 Chr.5: 170109382-170109382 on Build GRCh38
rs550861326 Chr.5: 170109187-170109187 on Build GRCh38
rs1049268321 Chr.5: 170109429-170109429 on Build GRCh38
rs886060404 Chr.5: 170109318-170109318 on Build GRCh38
rs1004688536 Chr.5: 170109434-170109434 on Build GRCh38
rs550211890 Chr.5: 170109426-170109426 on Build GRCh38
rs576939349 Chr.5: 170109309-170109309 on Build GRCh38
rs528212394 Chr.5: 170109400-170109400 on Build GRCh38
rs775308176 Chr.5: 170109621-170109622 on Build GRCh38
rs965383447 Chr.5: 170109177-170109177 on Build GRCh38
rs533523592 Chr.5: 170109556-170109556 on Build GRCh38
rs3839285 Chr.5: 170109541-170109542 on Build GRCh38
rs757859974 Chr.5: 170109247-170109247 on Build GRCh38
rs386405656 Chr.5: 170109548-170109549 on Build GRCh38
rs916222243 Chr.5: 170109405-170109405 on Build GRCh38
rs765932799 Chr.5: 170109375-170109375 on Build GRCh38
rs548207515 Chr.5: 170109452-170109452 on Build GRCh38
rs561697271 Chr.5: 170109509-170109509 on Build GRCh38
rs998602713 Chr.5: 170109465-170109465 on Build GRCh38
rs1040332467 Chr.5: 170109435-170109435 on Build GRCh38
rs561616826 Chr.5: 170109305-170109305 on Build GRCh38
rs546888065 Chr.5: 170109625-170109625 on Build GRCh38
rs529428860 Chr.5: 170109532-170109532 on Build GRCh38
rs902449495 Chr.5: 170109406-170109406 on Build GRCh38
rs1035240161 Chr.5: 170109536-170109536 on Build GRCh38
rs983686401 Chr.5: 170109278-170109278 on Build GRCh38
rs201086598 Chr.5: 170109541-170109541 on Build GRCh38
rs771802654 Chr.5: 170109562-170109562 on Build GRCh38
rs753090561 Chr.5: 170109471-170109471 on Build GRCh38
rs1054762463 Chr.5: 170109402-170109402 on Build GRCh38
rs559357224 Chr.5: 170109354-170109354 on Build GRCh38
rs545680200 Chr.5: 170109313-170109313 on Build GRCh38
rs914691555 Chr.5: 170109624-170109624 on Build GRCh38
rs1040197567 Chr.5: 170109626-170109626 on Build GRCh38
rs45466695 Chr.5: 170109535-170109535 on Build GRCh38
rs976455348 Chr.5: 170109551-170109551 on Build GRCh38
rs570034406 Chr.5: 170109518-170109518 on Build GRCh38

View Full Product Details