Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176656790-176657283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN17
Entrez Gene ID
26262
Gene Name
tetraspanin 17
Gene Aliases
FBX23, FBXO23, TM4SF17
UniGene
Hs.532129
Gene Chromosome Location
Chr.5: 176647387-176659058 on Build GRCh38
Transcripts
NM_001006616.2 NM_012171.2 NM_130465.4 XM_005265876.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614379 c.739C>T p.L247F Chr.5: 176656808-176656808 on Build GRCh38
3340588 c.777C>T p.L259L Chr.5: 176656924-176656924 on Build GRCh38
1665996 c.747+1G>T p.? Chr.5: 176656817-176656817 on Build GRCh38
7193828 c.757A>G p.I253V Chr.5: 176656904-176656904 on Build GRCh38
6218052 c.721G>A p.V241I Chr.5: 176656790-176656790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182817482 Chr.5: 176657086-176657086 on Build GRCh38
rs750641909 Chr.5: 176656983-176656983 on Build GRCh38
rs772645367 Chr.5: 176656866-176656866 on Build GRCh38
rs780961599 Chr.5: 176656855-176656855 on Build GRCh38
rs914006607 Chr.5: 176657092-176657092 on Build GRCh38
rs973667693 Chr.5: 176657024-176657024 on Build GRCh38
rs760889135 Chr.5: 176656811-176656811 on Build GRCh38
rs751528425 Chr.5: 176657012-176657012 on Build GRCh38
rs368228156 Chr.5: 176656985-176656985 on Build GRCh38
rs758395604 Chr.5: 176656841-176656841 on Build GRCh38
rs756607290 Chr.5: 176656834-176656834 on Build GRCh38
rs761523681 Chr.5: 176656820-176656820 on Build GRCh38
rs946066482 Chr.5: 176657259-176657259 on Build GRCh38
rs548977433 Chr.5: 176657155-176657155 on Build GRCh38
rs973544096 Chr.5: 176657233-176657233 on Build GRCh38
rs112564143 Chr.5: 176656958-176656958 on Build GRCh38
rs779080904 Chr.5: 176656858-176656858 on Build GRCh38
rs1044103599 Chr.5: 176657154-176657154 on Build GRCh38
rs755886392 Chr.5: 176657104-176657104 on Build GRCh38
rs1051779331 Chr.5: 176656879-176656879 on Build GRCh38
rs1038400662 Chr.5: 176656854-176656854 on Build GRCh38
rs970840525 Chr.5: 176657209-176657209 on Build GRCh38
rs765901707 Chr.5: 176656933-176656933 on Build GRCh38
rs1000770271 Chr.5: 176656982-176656982 on Build GRCh38
rs374325652 Chr.5: 176656835-176656835 on Build GRCh38
rs537714468 Chr.5: 176657172-176657172 on Build GRCh38
rs78392471 Chr.5: 176657210-176657210 on Build GRCh38
rs777109974 Chr.5: 176656915-176656915 on Build GRCh38
rs764759797 Chr.5: 176656919-176656919 on Build GRCh38
rs201047148 Chr.5: 176656976-176656976 on Build GRCh38
rs200333069 Chr.5: 176656898-176656898 on Build GRCh38
rs745725587 Chr.5: 176657093-176657093 on Build GRCh38
rs745867963 Chr.5: 176656859-176656859 on Build GRCh38
rs566561020 Chr.5: 176657010-176657010 on Build GRCh38
rs754879483 Chr.5: 176656943-176656943 on Build GRCh38
rs200586493 Chr.5: 176656990-176656990 on Build GRCh38
rs987761137 Chr.5: 176657251-176657251 on Build GRCh38
rs34292197 Chr.5: 176657202-176657203 on Build GRCh38
rs761538357 Chr.5: 176656852-176656852 on Build GRCh38
rs959293491 Chr.5: 176656991-176656991 on Build GRCh38
rs976067001 Chr.5: 176657023-176657023 on Build GRCh38
rs1024590180 Chr.5: 176657208-176657208 on Build GRCh38
rs964413849 Chr.5: 176657240-176657240 on Build GRCh38
rs182442495 Chr.5: 176656851-176656851 on Build GRCh38
rs530385037 Chr.5: 176657141-176657141 on Build GRCh38
rs1001076636 Chr.5: 176657152-176657152 on Build GRCh38
rs4868677 Chr.5: 176656996-176656996 on Build GRCh38
rs75345917 Chr.5: 176656993-176656993 on Build GRCh38
rs752449443 Chr.5: 176656973-176656973 on Build GRCh38
rs778943343 Chr.5: 176656981-176656981 on Build GRCh38
rs767761958 Chr.5: 176656965-176656965 on Build GRCh38
rs772037181 Chr.5: 176656859-176656859 on Build GRCh38
rs960899080 Chr.5: 176656946-176656946 on Build GRCh38
rs761575243 Chr.5: 176656917-176656917 on Build GRCh38
rs748461525 Chr.5: 176656995-176656995 on Build GRCh38
rs187776418 Chr.5: 176657165-176657165 on Build GRCh38
rs4868678 Chr.5: 176657159-176657159 on Build GRCh38
rs189563674 Chr.5: 176657043-176657043 on Build GRCh38
rs61753280 Chr.5: 176656924-176656924 on Build GRCh38
rs895030155 Chr.5: 176657158-176657158 on Build GRCh38
rs775916485 Chr.5: 176656880-176656880 on Build GRCh38
rs772786406 Chr.5: 176656921-176656921 on Build GRCh38
rs551109009 Chr.5: 176657101-176657101 on Build GRCh38
rs922150313 Chr.5: 176657032-176657032 on Build GRCh38
rs772677973 Chr.5: 176657258-176657258 on Build GRCh38
rs554466133 Chr.5: 176657214-176657214 on Build GRCh38
rs778456478 Chr.5: 176657215-176657215 on Build GRCh38
rs573903435 Chr.5: 176657015-176657015 on Build GRCh38
rs1020777116 Chr.5: 176656975-176656975 on Build GRCh38
rs747564387 Chr.5: 176656992-176656992 on Build GRCh38
rs890505458 Chr.5: 176656964-176656964 on Build GRCh38
rs964981382 Chr.5: 176657021-176657021 on Build GRCh38
rs747511389 Chr.5: 176656889-176656889 on Build GRCh38
rs563047570 Chr.5: 176657133-176657133 on Build GRCh38
rs62402583 Chr.5: 176657234-176657234 on Build GRCh38
rs776490167 Chr.5: 176656816-176656816 on Build GRCh38
rs751519664 Chr.5: 176656937-176656937 on Build GRCh38
rs532280960 Chr.5: 176657022-176657022 on Build GRCh38
rs201230552 Chr.5: 176656857-176656857 on Build GRCh38
rs934489892 Chr.5: 176657069-176657069 on Build GRCh38
rs540849895 Chr.5: 176657042-176657042 on Build GRCh38
rs200164200 Chr.5: 176656852-176656852 on Build GRCh38
rs963172956 Chr.5: 176657231-176657231 on Build GRCh38
rs762664932 Chr.5: 176657006-176657006 on Build GRCh38
rs764538224 Chr.5: 176656813-176656813 on Build GRCh38
rs772879206 Chr.5: 176656929-176656929 on Build GRCh38
rs1051731962 Chr.5: 176657078-176657078 on Build GRCh38
rs932213325 Chr.5: 176657241-176657241 on Build GRCh38
rs187034910 Chr.5: 176657011-176657011 on Build GRCh38

View Full Product Details