Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29792073-29792574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCG4
Entrez Gene ID
54435
Gene Name
HLA complex group 4 (non-protein coding)
Gene Aliases
HCG4P10, HCGIV-10, HCGIV.9
UniGene
Hs.660431
Gene Chromosome Location
Chr.6: 29791031-29793073 on Build GRCh38
Transcripts
NR_002139.2 NR_132323.1

Gene Symbol
LOC554223
Entrez Gene ID
554223
Gene Name
histocompatibility antigen-related
Gene Aliases
-
UniGene
Hs.60856
Gene Chromosome Location
Chr.6: 29791906-29797807 on Build GRCh38
Transcripts
NR_002139.2 NR_132323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316448 c.193C>A p.P65T Chr.6: 29792200-29792200 on Build GRCh38
7219627 c.412G>A p.D138N Chr.6: 29792550-29792550 on Build GRCh38
7247801 c.279C>T p.V93V Chr.6: 29792286-29792286 on Build GRCh38
4596044 c.402G>C p.R134R Chr.6: 29792540-29792540 on Build GRCh38
4593329 c.101G>A p.R34Q Chr.6: 29792108-29792108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777227696 Chr.6: 29792500-29792500 on Build GRCh38
rs45479291 Chr.6: 29792252-29792252 on Build GRCh38
rs749771955 Chr.6: 29792532-29792532 on Build GRCh38
rs979565332 Chr.6: 29792166-29792166 on Build GRCh38
rs747649407 Chr.6: 29792354-29792354 on Build GRCh38
rs1032475637 Chr.6: 29792376-29792376 on Build GRCh38
rs766787798 Chr.6: 29792203-29792203 on Build GRCh38
rs760051201 Chr.6: 29792313-29792313 on Build GRCh38
rs779825685 Chr.6: 29792256-29792256 on Build GRCh38
rs770525496 Chr.6: 29792391-29792391 on Build GRCh38
rs767773208 Chr.6: 29792544-29792544 on Build GRCh38
rs747667845 Chr.6: 29792268-29792268 on Build GRCh38
rs886388139 Chr.6: 29792274-29792274 on Build GRCh38
rs773505681 Chr.6: 29792192-29792192 on Build GRCh38
rs200069678 Chr.6: 29792514-29792514 on Build GRCh38
rs776991645 Chr.6: 29792395-29792395 on Build GRCh38
rs925458263 Chr.6: 29792169-29792169 on Build GRCh38
rs778157296 Chr.6: 29792247-29792247 on Build GRCh38
rs773754438 Chr.6: 29792165-29792165 on Build GRCh38
rs941913320 Chr.6: 29792497-29792497 on Build GRCh38
rs930810473 Chr.6: 29792205-29792205 on Build GRCh38
rs753579239 Chr.6: 29792337-29792337 on Build GRCh38
rs758358734 Chr.6: 29792480-29792480 on Build GRCh38
rs7764777 Chr.6: 29792372-29792372 on Build GRCh38
rs922956820 Chr.6: 29792228-29792228 on Build GRCh38
rs115645599 Chr.6: 29792340-29792340 on Build GRCh38
rs543704267 Chr.6: 29792366-29792366 on Build GRCh38
rs757441775 Chr.6: 29792331-29792331 on Build GRCh38
rs761082849 Chr.6: 29792200-29792200 on Build GRCh38
rs983494728 Chr.6: 29792484-29792484 on Build GRCh38
rs746235180 Chr.6: 29792398-29792398 on Build GRCh38
rs183041424 Chr.6: 29792479-29792479 on Build GRCh38
rs749200579 Chr.6: 29792260-29792260 on Build GRCh38
rs772649467 Chr.6: 29792245-29792246 on Build GRCh38
rs746240494 Chr.6: 29792517-29792517 on Build GRCh38
rs765669205 Chr.6: 29792315-29792315 on Build GRCh38
rs753651090 Chr.6: 29792456-29792456 on Build GRCh38
rs755149489 Chr.6: 29792113-29792113 on Build GRCh38
rs61737605 Chr.6: 29792540-29792540 on Build GRCh38
rs966853196 Chr.6: 29792153-29792153 on Build GRCh38
rs542620054 Chr.6: 29792373-29792373 on Build GRCh38
rs748501351 Chr.6: 29792230-29792230 on Build GRCh38
rs1611209 Chr.6: 29792146-29792146 on Build GRCh38
rs758720386 Chr.6: 29792137-29792137 on Build GRCh38
rs771380476 Chr.6: 29792299-29792299 on Build GRCh38
rs979681408 Chr.6: 29792374-29792374 on Build GRCh38
rs888207510 Chr.6: 29792226-29792226 on Build GRCh38
rs1737006 Chr.6: 29792334-29792334 on Build GRCh38
rs13211705 Chr.6: 29792320-29792320 on Build GRCh38
rs764319473 Chr.6: 29792422-29792422 on Build GRCh38
rs762142366 Chr.6: 29792102-29792102 on Build GRCh38
rs1043947295 Chr.6: 29792473-29792473 on Build GRCh38
rs770324120 Chr.6: 29792518-29792518 on Build GRCh38
rs767819505 Chr.6: 29792103-29792103 on Build GRCh38
rs746509116 Chr.6: 29792504-29792504 on Build GRCh38
rs1610718 Chr.6: 29792411-29792411 on Build GRCh38
rs1010846680 Chr.6: 29792470-29792470 on Build GRCh38
rs770894489 Chr.6: 29792094-29792094 on Build GRCh38
rs769291425 Chr.6: 29792539-29792539 on Build GRCh38
rs776690590 Chr.6: 29792095-29792095 on Build GRCh38
rs761413564 Chr.6: 29792178-29792178 on Build GRCh38
rs777937013 Chr.6: 29792537-29792559 on Build GRCh38
rs775556236 Chr.6: 29792144-29792144 on Build GRCh38
rs190797234 Chr.6: 29792191-29792191 on Build GRCh38
rs551267656 Chr.6: 29792107-29792107 on Build GRCh38
rs769885958 Chr.6: 29792097-29792097 on Build GRCh38
rs978072819 Chr.6: 29792154-29792154 on Build GRCh38
rs770414084 Chr.6: 29792406-29792406 on Build GRCh38
rs922090180 Chr.6: 29792204-29792204 on Build GRCh38
rs907565065 Chr.6: 29792486-29792486 on Build GRCh38
rs1036746745 Chr.6: 29792405-29792405 on Build GRCh38
rs1033651151 Chr.6: 29792101-29792101 on Build GRCh38
rs908061398 Chr.6: 29792385-29792385 on Build GRCh38
rs937194161 Chr.6: 29792543-29792543 on Build GRCh38
rs768507038 Chr.6: 29792262-29792262 on Build GRCh38
rs114179071 Chr.6: 29792355-29792355 on Build GRCh38
rs763641080 Chr.6: 29792100-29792100 on Build GRCh38
rs1055451736 Chr.6: 29792293-29792293 on Build GRCh38
rs530677491 Chr.6: 29792553-29792553 on Build GRCh38
rs778877513 Chr.6: 29792343-29792343 on Build GRCh38
rs750816705 Chr.6: 29792105-29792105 on Build GRCh38
rs1020978981 Chr.6: 29792360-29792360 on Build GRCh38
rs768262278 Chr.6: 29792156-29792156 on Build GRCh38
rs764407587 Chr.6: 29792326-29792326 on Build GRCh38
rs765125231 Chr.6: 29792474-29792474 on Build GRCh38
rs754793815 Chr.6: 29792472-29792472 on Build GRCh38
rs559726901 Chr.6: 29792408-29792408 on Build GRCh38
rs1610645 Chr.6: 29792219-29792219 on Build GRCh38
rs374283668 Chr.6: 29792342-29792342 on Build GRCh38
rs776119010 Chr.6: 29792529-29792529 on Build GRCh38
rs774797738 Chr.6: 29792098-29792098 on Build GRCh38
rs776886396 Chr.6: 29792307-29792307 on Build GRCh38
rs750231734 Chr.6: 29792248-29792248 on Build GRCh38
rs564322364 Chr.6: 29792238-29792238 on Build GRCh38
rs778325707 Chr.6: 29792266-29792266 on Build GRCh38
rs762027844 Chr.6: 29792430-29792430 on Build GRCh38
rs772912434 Chr.6: 29792279-29792279 on Build GRCh38
rs911919208 Chr.6: 29792163-29792163 on Build GRCh38
rs769561589 Chr.6: 29792141-29792141 on Build GRCh38
rs767645075 Chr.6: 29792447-29792447 on Build GRCh38
rs577856641 Chr.6: 29792356-29792356 on Build GRCh38
rs752858836 Chr.6: 29792229-29792229 on Build GRCh38
rs760911980 Chr.6: 29792287-29792287 on Build GRCh38
rs771696419 Chr.6: 29792277-29792277 on Build GRCh38
rs1611208 Chr.6: 29792108-29792108 on Build GRCh38
rs376123818 Chr.6: 29792367-29792367 on Build GRCh38
rs1055020736 Chr.6: 29792435-29792435 on Build GRCh38
rs1019042096 Chr.6: 29792498-29792498 on Build GRCh38
rs200560631 Chr.6: 29792530-29792530 on Build GRCh38
rs778976135 Chr.6: 29792116-29792116 on Build GRCh38
rs1611207 Chr.6: 29792099-29792099 on Build GRCh38
rs781096884 Chr.6: 29792140-29792140 on Build GRCh38
rs1034218982 Chr.6: 29792352-29792352 on Build GRCh38
rs964911298 Chr.6: 29792417-29792417 on Build GRCh38
rs563590890 Chr.6: 29792369-29792369 on Build GRCh38
rs748434926 Chr.6: 29792133-29792133 on Build GRCh38
rs754405079 Chr.6: 29792212-29792212 on Build GRCh38
rs1037615547 Chr.6: 29792507-29792507 on Build GRCh38
rs1022157529 Chr.6: 29792318-29792318 on Build GRCh38
rs752768857 Chr.6: 29792317-29792317 on Build GRCh38
rs759516635 Chr.6: 29792213-29792213 on Build GRCh38
rs746628615 Chr.6: 29792390-29792390 on Build GRCh38
rs552949222 Chr.6: 29792329-29792329 on Build GRCh38
rs756544323 Chr.6: 29792106-29792106 on Build GRCh38

View Full Product Details