Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31507241-31507759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICB
Entrez Gene ID
4277
Gene Name
MHC class I polypeptide-related sequence B
Gene Aliases
PERB11.2
UniGene
Hs.731446
Gene Chromosome Location
Chr.6: 31494881-31511124 on Build GRCh38
Transcripts
NM_001289160.1 NM_001289161.1 NM_005931.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403600 c.931C>T p.P311S Chr.6: 31507438-31507438 on Build GRCh38
3761885 c.870C>T p.H290H Chr.6: 31507278-31507278 on Build GRCh38
483855 c.968T>A p.I323N Chr.6: 31507475-31507475 on Build GRCh38
3830100 c.951G>A p.M317I Chr.6: 31507458-31507458 on Build GRCh38
741390 c.996G>A p.K332K Chr.6: 31507503-31507503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375735072 Chr.6: 31507518-31507518 on Build GRCh38
rs747784533 Chr.6: 31507559-31507559 on Build GRCh38
rs368616699 Chr.6: 31507426-31507426 on Build GRCh38
rs568721471 Chr.6: 31507597-31507597 on Build GRCh38
rs374212282 Chr.6: 31507526-31507526 on Build GRCh38
rs761418200 Chr.6: 31507351-31507351 on Build GRCh38
rs11509487 Chr.6: 31507315-31507316 on Build GRCh38
rs200882372 Chr.6: 31507345-31507345 on Build GRCh38
rs772593233 Chr.6: 31507567-31507567 on Build GRCh38
rs200730915 Chr.6: 31507548-31507548 on Build GRCh38
rs777683189 Chr.6: 31507517-31507517 on Build GRCh38
rs370386306 Chr.6: 31507319-31507319 on Build GRCh38
rs758668810 Chr.6: 31507516-31507516 on Build GRCh38
rs368095298 Chr.6: 31507397-31507397 on Build GRCh38
rs1024282587 Chr.6: 31507625-31507625 on Build GRCh38
rs45519735 Chr.6: 31507278-31507278 on Build GRCh38
rs766632217 Chr.6: 31507386-31507386 on Build GRCh38
rs115231984 Chr.6: 31507736-31507736 on Build GRCh38
rs755350585 Chr.6: 31507497-31507497 on Build GRCh38
rs35849126 Chr.6: 31507665-31507666 on Build GRCh38
rs755987354 Chr.6: 31507525-31507525 on Build GRCh38
rs756455541 Chr.6: 31507370-31507370 on Build GRCh38
rs752936258 Chr.6: 31507515-31507515 on Build GRCh38
rs768916248 Chr.6: 31507271-31507271 on Build GRCh38
rs566742548 Chr.6: 31507604-31507604 on Build GRCh38
rs776435141 Chr.6: 31507288-31507288 on Build GRCh38
rs201731965 Chr.6: 31507464-31507464 on Build GRCh38
rs770843869 Chr.6: 31507280-31507280 on Build GRCh38
rs771797650 Chr.6: 31507560-31507560 on Build GRCh38
rs754141865 Chr.6: 31507495-31507495 on Build GRCh38
rs775139123 Chr.6: 31507432-31507432 on Build GRCh38
rs759143175 Chr.6: 31507469-31507470 on Build GRCh38
rs3132468 Chr.6: 31507709-31507709 on Build GRCh38
rs866950989 Chr.6: 31507450-31507450 on Build GRCh38
rs550543205 Chr.6: 31507456-31507456 on Build GRCh38
rs914984278 Chr.6: 31507724-31507724 on Build GRCh38
rs775715352 Chr.6: 31507394-31507394 on Build GRCh38
rs375872171 Chr.6: 31507420-31507420 on Build GRCh38
rs371638435 Chr.6: 31507270-31507270 on Build GRCh38
rs376855573 Chr.6: 31507269-31507269 on Build GRCh38
rs765401600 Chr.6: 31507504-31507504 on Build GRCh38
rs138352176 Chr.6: 31507568-31507568 on Build GRCh38
rs181443790 Chr.6: 31507614-31507614 on Build GRCh38
rs769806725 Chr.6: 31507427-31507427 on Build GRCh38
rs561660266 Chr.6: 31507387-31507387 on Build GRCh38
rs773620107 Chr.6: 31507469-31507474 on Build GRCh38
rs3131638 Chr.6: 31507350-31507350 on Build GRCh38
rs749151497 Chr.6: 31507437-31507437 on Build GRCh38
rs762450313 Chr.6: 31507469-31507471 on Build GRCh38
rs768073779 Chr.6: 31507354-31507354 on Build GRCh38
rs977599161 Chr.6: 31507383-31507383 on Build GRCh38
rs750930133 Chr.6: 31507355-31507355 on Build GRCh38
rs1031939575 Chr.6: 31507676-31507676 on Build GRCh38
rs377052867 Chr.6: 31507460-31507460 on Build GRCh38
rs995362095 Chr.6: 31507286-31507286 on Build GRCh38
rs774707886 Chr.6: 31507440-31507441 on Build GRCh38
rs117481357 Chr.6: 31507718-31507718 on Build GRCh38
rs528089914 Chr.6: 31507310-31507310 on Build GRCh38
rs924803269 Chr.6: 31507393-31507393 on Build GRCh38
rs890376882 Chr.6: 31507444-31507444 on Build GRCh38
rs766976414 Chr.6: 31507477-31507477 on Build GRCh38
rs978047955 Chr.6: 31507666-31507666 on Build GRCh38
rs3130615 Chr.6: 31507636-31507636 on Build GRCh38
rs138314754 Chr.6: 31507307-31507308 on Build GRCh38
rs749021734 Chr.6: 31507541-31507541 on Build GRCh38
rs772297798 Chr.6: 31507468-31507469 on Build GRCh38
rs781134713 Chr.6: 31507419-31507419 on Build GRCh38
rs41273040 Chr.6: 31507279-31507279 on Build GRCh38
rs769499370 Chr.6: 31507314-31507314 on Build GRCh38
rs567693124 Chr.6: 31507683-31507683 on Build GRCh38
rs372919851 Chr.6: 31507440-31507440 on Build GRCh38
rs774100611 Chr.6: 31507558-31507558 on Build GRCh38
rs762659841 Chr.6: 31507343-31507343 on Build GRCh38
rs1027709719 Chr.6: 31507306-31507306 on Build GRCh38
rs772831299 Chr.6: 31507462-31507462 on Build GRCh38
rs958790674 Chr.6: 31507519-31507519 on Build GRCh38
rs45470602 Chr.6: 31507406-31507406 on Build GRCh38

View Full Product Details