Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106521003-106521525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM1
Entrez Gene ID
202
Gene Name
absent in melanoma 1
Gene Aliases
CRYBG1, ST4
UniGene
Hs.724262
Gene Chromosome Location
Chr.6: 106360808-106570449 on Build GRCh38
Transcripts
NM_001624.3 XM_005266839.3 XM_017010333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
739786 c.2590A>G p.T864A Chr.6: 106521022-106521022 on Build GRCh38
1071770 c.2688G>A p.L896L Chr.6: 106521120-106521120 on Build GRCh38
6547537 c.2692G>T p.D898Y Chr.6: 106521124-106521124 on Build GRCh38
1071769 c.2581G>A p.A861T Chr.6: 106521013-106521013 on Build GRCh38
739785 c.2691C>G p.P897P Chr.6: 106521123-106521123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187710226 Chr.6: 106521482-106521482 on Build GRCh38
rs775150386 Chr.6: 106521269-106521269 on Build GRCh38
rs1032636068 Chr.6: 106521247-106521247 on Build GRCh38
rs200461385 Chr.6: 106521123-106521123 on Build GRCh38
rs780076876 Chr.6: 106521099-106521099 on Build GRCh38
rs771571468 Chr.6: 106521363-106521363 on Build GRCh38
rs777224294 Chr.6: 106521042-106521042 on Build GRCh38
rs763975227 Chr.6: 106521155-106521155 on Build GRCh38
rs201797268 Chr.6: 106521329-106521329 on Build GRCh38
rs146778804 Chr.6: 106521152-106521152 on Build GRCh38
rs999044727 Chr.6: 106521160-106521160 on Build GRCh38
rs768892501 Chr.6: 106521115-106521115 on Build GRCh38
rs769481114 Chr.6: 106521500-106521500 on Build GRCh38
rs751728606 Chr.6: 106521196-106521196 on Build GRCh38
rs931861110 Chr.6: 106521383-106521383 on Build GRCh38
rs139997199 Chr.6: 106521404-106521404 on Build GRCh38
rs751257287 Chr.6: 106521080-106521080 on Build GRCh38
rs144529082 Chr.6: 106521327-106521327 on Build GRCh38
rs374238219 Chr.6: 106521128-106521128 on Build GRCh38
rs773226061 Chr.6: 106521502-106521502 on Build GRCh38
rs761878814 Chr.6: 106521293-106521293 on Build GRCh38
rs142617024 Chr.6: 106521376-106521376 on Build GRCh38
rs539874404 Chr.6: 106521413-106521413 on Build GRCh38
rs752973425 Chr.6: 106521328-106521328 on Build GRCh38
rs533984443 Chr.6: 106521245-106521245 on Build GRCh38
rs761018814 Chr.6: 106521415-106521415 on Build GRCh38
rs937805774 Chr.6: 106521348-106521348 on Build GRCh38
rs778938706 Chr.6: 106521086-106521086 on Build GRCh38
rs778358007 Chr.6: 106521231-106521231 on Build GRCh38
rs367761608 Chr.6: 106521345-106521345 on Build GRCh38
rs747292072 Chr.6: 106521034-106521034 on Build GRCh38
rs746635528 Chr.6: 106521478-106521478 on Build GRCh38
rs756577686 Chr.6: 106521224-106521224 on Build GRCh38
rs772228590 Chr.6: 106521028-106521028 on Build GRCh38
rs894077902 Chr.6: 106521496-106521496 on Build GRCh38
rs370122396 Chr.6: 106521122-106521122 on Build GRCh38
rs755294260 Chr.6: 106521209-106521209 on Build GRCh38
rs747271353 Chr.6: 106521110-106521110 on Build GRCh38
rs776424472 Chr.6: 106521283-106521283 on Build GRCh38
rs944543786 Chr.6: 106521403-106521403 on Build GRCh38
rs761637731 Chr.6: 106521386-106521386 on Build GRCh38
rs138198128 Chr.6: 106521307-106521307 on Build GRCh38
rs554973603 Chr.6: 106521159-106521159 on Build GRCh38
rs770475403 Chr.6: 106521124-106521124 on Build GRCh38
rs745483795 Chr.6: 106521260-106521260 on Build GRCh38
rs544344817 Chr.6: 106521378-106521378 on Build GRCh38
rs371480971 Chr.6: 106521405-106521405 on Build GRCh38
rs759942742 Chr.6: 106521443-106521443 on Build GRCh38
rs373317464 Chr.6: 106521077-106521077 on Build GRCh38
rs149596949 Chr.6: 106521318-106521318 on Build GRCh38
rs768407460 Chr.6: 106521479-106521479 on Build GRCh38
rs772891918 Chr.6: 106521061-106521061 on Build GRCh38
rs771661623 Chr.6: 106521262-106521262 on Build GRCh38
rs765412508 Chr.6: 106521132-106521133 on Build GRCh38
rs990986216 Chr.6: 106521306-106521306 on Build GRCh38
rs375366785 Chr.6: 106521332-106521332 on Build GRCh38
rs779611046 Chr.6: 106521461-106521461 on Build GRCh38
rs1008705878 Chr.6: 106521170-106521170 on Build GRCh38
rs760392136 Chr.6: 106521280-106521280 on Build GRCh38
rs766542926 Chr.6: 106521165-106521165 on Build GRCh38
rs779307555 Chr.6: 106521352-106521352 on Build GRCh38
rs199825949 Chr.6: 106521424-106521424 on Build GRCh38
rs780731550 Chr.6: 106521480-106521480 on Build GRCh38
rs762472106 Chr.6: 106521059-106521059 on Build GRCh38
rs1054596985 Chr.6: 106521418-106521418 on Build GRCh38
rs377595303 Chr.6: 106521151-106521151 on Build GRCh38
rs201230039 Chr.6: 106521029-106521029 on Build GRCh38
rs965816836 Chr.6: 106521273-106521273 on Build GRCh38
rs756083117 Chr.6: 106521085-106521085 on Build GRCh38
rs762330813 Chr.6: 106521136-106521136 on Build GRCh38
rs146033137 Chr.6: 106521391-106521391 on Build GRCh38
rs375860758 Chr.6: 106521154-106521154 on Build GRCh38
rs369377315 Chr.6: 106521487-106521487 on Build GRCh38
rs372779090 Chr.6: 106521244-106521244 on Build GRCh38
rs765900077 Chr.6: 106521069-106521069 on Build GRCh38
rs139327699 Chr.6: 106521375-106521375 on Build GRCh38
rs561072991 Chr.6: 106521382-106521382 on Build GRCh38
rs142990443 Chr.6: 106521156-106521156 on Build GRCh38
rs748476178 Chr.6: 106521213-106521213 on Build GRCh38
rs140407254 Chr.6: 106521322-106521322 on Build GRCh38
rs199971648 Chr.6: 106521084-106521084 on Build GRCh38
rs773862291 Chr.6: 106521146-106521146 on Build GRCh38
rs267600753 Chr.6: 106521199-106521199 on Build GRCh38
rs773950741 Chr.6: 106521257-106521257 on Build GRCh38
rs780341653 Chr.6: 106521033-106521033 on Build GRCh38
rs375834494 Chr.6: 106521446-106521446 on Build GRCh38
rs745340305 Chr.6: 106521358-106521358 on Build GRCh38
rs949074343 Chr.6: 106521082-106521082 on Build GRCh38
rs752822054 Chr.6: 106521227-106521229 on Build GRCh38
rs754861208 Chr.6: 106521279-106521279 on Build GRCh38
rs142050085 Chr.6: 106521431-106521431 on Build GRCh38
rs773056665 Chr.6: 106521398-106521398 on Build GRCh38
rs769426664 Chr.6: 106521060-106521060 on Build GRCh38
rs769073910 Chr.6: 106521038-106521038 on Build GRCh38
rs575356547 Chr.6: 106521335-106521335 on Build GRCh38
rs970514934 Chr.6: 106521316-106521316 on Build GRCh38
rs201285247 Chr.6: 106521138-106521138 on Build GRCh38
rs202235693 Chr.6: 106521083-106521083 on Build GRCh38
rs150414602 Chr.6: 106521210-106521210 on Build GRCh38
rs566268314 Chr.6: 106521097-106521097 on Build GRCh38
rs146905899 Chr.6: 106521068-106521068 on Build GRCh38
rs757894683 Chr.6: 106521452-106521452 on Build GRCh38

View Full Product Details