Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2917203-2917667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD11
Entrez Gene ID
84433
Gene Name
caspase recruitment domain family member 11
Gene Aliases
BENTA, BIMP3, CARMA1, IMD11, PPBL
UniGene
Hs.648101
Gene Chromosome Location
Chr.7: 2906075-3043945 on Build GRCh38
Transcripts
NM_001324281.1 NM_032415.5 XM_011515586.2 XM_011515587.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5710537 c.2677A>T p.S893C Chr.7: 2917316-2917316 on Build GRCh38
5997546 c.2631G>A p.A877A Chr.7: 2917362-2917362 on Build GRCh38
6912913 c.2698C>T p.L900F Chr.7: 2917295-2917295 on Build GRCh38
5019288 c.2622A>G p.P874P Chr.7: 2917371-2917371 on Build GRCh38
1716026 c.2663G>A p.R888H Chr.7: 2917330-2917330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377702889 Chr.7: 2917238-2917238 on Build GRCh38
rs1042785865 Chr.7: 2917594-2917594 on Build GRCh38
rs201282256 Chr.7: 2917267-2917267 on Build GRCh38
rs927214222 Chr.7: 2917495-2917495 on Build GRCh38
rs980008889 Chr.7: 2917494-2917494 on Build GRCh38
rs151053853 Chr.7: 2917332-2917332 on Build GRCh38
rs1000297229 Chr.7: 2917557-2917557 on Build GRCh38
rs527714292 Chr.7: 2917456-2917456 on Build GRCh38
rs3735123 Chr.7: 2917284-2917284 on Build GRCh38
rs1015218915 Chr.7: 2917521-2917521 on Build GRCh38
rs375163437 Chr.7: 2917266-2917266 on Build GRCh38
rs370020664 Chr.7: 2917243-2917243 on Build GRCh38
rs375613504 Chr.7: 2917583-2917583 on Build GRCh38
rs756219246 Chr.7: 2917425-2917425 on Build GRCh38
rs750486472 Chr.7: 2917322-2917322 on Build GRCh38
rs952495417 Chr.7: 2917502-2917502 on Build GRCh38
rs953684131 Chr.7: 2917380-2917380 on Build GRCh38
rs775771988 Chr.7: 2917362-2917362 on Build GRCh38
rs1050675470 Chr.7: 2917596-2917596 on Build GRCh38
rs988999113 Chr.7: 2917453-2917453 on Build GRCh38
rs773308753 Chr.7: 2917399-2917402 on Build GRCh38
rs774944371 Chr.7: 2917590-2917590 on Build GRCh38
rs770706374 Chr.7: 2917350-2917350 on Build GRCh38
rs191837050 Chr.7: 2917289-2917289 on Build GRCh38
rs3735124 Chr.7: 2917371-2917371 on Build GRCh38
rs543253971 Chr.7: 2917363-2917363 on Build GRCh38
rs996347748 Chr.7: 2917480-2917481 on Build GRCh38
rs201256077 Chr.7: 2917286-2917286 on Build GRCh38
rs111660799 Chr.7: 2917562-2917562 on Build GRCh38
rs556715404 Chr.7: 2917239-2917239 on Build GRCh38
rs746650253 Chr.7: 2917347-2917347 on Build GRCh38
rs984779462 Chr.7: 2917533-2917533 on Build GRCh38
rs762078325 Chr.7: 2917408-2917408 on Build GRCh38
rs570201130 Chr.7: 2917592-2917592 on Build GRCh38
rs902761185 Chr.7: 2917229-2917229 on Build GRCh38
rs772298133 Chr.7: 2917284-2917284 on Build GRCh38
rs946746705 Chr.7: 2917423-2917423 on Build GRCh38
rs146545469 Chr.7: 2917323-2917323 on Build GRCh38
rs6971582 Chr.7: 2917553-2917553 on Build GRCh38
rs981964732 Chr.7: 2917544-2917544 on Build GRCh38
rs539279952 Chr.7: 2917265-2917265 on Build GRCh38
rs397967116 Chr.7: 2917483-2917484 on Build GRCh38
rs927870120 Chr.7: 2917547-2917547 on Build GRCh38
rs930572452 Chr.7: 2917517-2917517 on Build GRCh38
rs777569549 Chr.7: 2917346-2917346 on Build GRCh38
rs1019143750 Chr.7: 2917462-2917462 on Build GRCh38
rs1033487626 Chr.7: 2917563-2917563 on Build GRCh38
rs907912831 Chr.7: 2917537-2917537 on Build GRCh38
rs866973330 Chr.7: 2917552-2917552 on Build GRCh38
rs923914665 Chr.7: 2917623-2917623 on Build GRCh38
rs3735122 Chr.7: 2917250-2917250 on Build GRCh38
rs142828146 Chr.7: 2917331-2917331 on Build GRCh38
rs369243746 Chr.7: 2917271-2917271 on Build GRCh38
rs764369699 Chr.7: 2917377-2917377 on Build GRCh38
rs971638154 Chr.7: 2917489-2917489 on Build GRCh38
rs748079791 Chr.7: 2917334-2917334 on Build GRCh38
rs201489315 Chr.7: 2917397-2917397 on Build GRCh38
rs558286041 Chr.7: 2917277-2917277 on Build GRCh38
rs761775078 Chr.7: 2917619-2917619 on Build GRCh38
rs753142060 Chr.7: 2917273-2917273 on Build GRCh38
rs567469370 Chr.7: 2917535-2917535 on Build GRCh38
rs886506846 Chr.7: 2917296-2917296 on Build GRCh38
rs1009103943 Chr.7: 2917231-2917231 on Build GRCh38
rs559324674 Chr.7: 2917643-2917643 on Build GRCh38
rs775406780 Chr.7: 2917237-2917237 on Build GRCh38
rs772771059 Chr.7: 2917253-2917253 on Build GRCh38
rs538987998 Chr.7: 2917617-2917617 on Build GRCh38
rs780059657 Chr.7: 2917433-2917433 on Build GRCh38
rs186809292 Chr.7: 2917434-2917434 on Build GRCh38
rs59298103 Chr.7: 2917479-2917480 on Build GRCh38
rs372718712 Chr.7: 2917278-2917278 on Build GRCh38
rs767702969 Chr.7: 2917418-2917418 on Build GRCh38
rs41467845 Chr.7: 2917420-2917420 on Build GRCh38
rs140097633 Chr.7: 2917352-2917352 on Build GRCh38
rs547379115 Chr.7: 2917532-2917532 on Build GRCh38
rs955849261 Chr.7: 2917232-2917232 on Build GRCh38
rs781316312 Chr.7: 2917299-2917299 on Build GRCh38
rs1024137570 Chr.7: 2917464-2917464 on Build GRCh38

View Full Product Details