Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5475951-5476442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL18
Entrez Gene ID
80028
Gene Name
F-box and leucine rich repeat protein 18
Gene Aliases
Fbl18
UniGene
Hs.623974
Gene Chromosome Location
Chr.7: 5454448-5513813 on Build GRCh38
Transcripts
NM_024963.5 XM_011515531.2

SNP Detail

SNP ID Chromosome Location
rs937825412 Chr.7: 5476183-5476183 on Build GRCh38
rs965722664 Chr.7: 5476104-5476104 on Build GRCh38
rs879367717 Chr.7: 5476061-5476061 on Build GRCh38
rs777846744 Chr.7: 5476186-5476186 on Build GRCh38
rs975711594 Chr.7: 5476123-5476123 on Build GRCh38
rs543509391 Chr.7: 5476102-5476102 on Build GRCh38
rs528837677 Chr.7: 5476143-5476143 on Build GRCh38
rs752835874 Chr.7: 5476210-5476210 on Build GRCh38
rs925171281 Chr.7: 5476182-5476182 on Build GRCh38
rs1036287041 Chr.7: 5476208-5476208 on Build GRCh38
rs561790586 Chr.7: 5476105-5476105 on Build GRCh38
rs534669859 Chr.7: 5476365-5476365 on Build GRCh38
rs141258633 Chr.7: 5476340-5476340 on Build GRCh38
rs998891230 Chr.7: 5476410-5476410 on Build GRCh38
rs958753628 Chr.7: 5476211-5476211 on Build GRCh38
rs567750326 Chr.7: 5476364-5476364 on Build GRCh38
rs1026320519 Chr.7: 5476338-5476338 on Build GRCh38
rs975919529 Chr.7: 5476281-5476281 on Build GRCh38
rs1057265501 Chr.7: 5476350-5476350 on Build GRCh38
rs887451751 Chr.7: 5476170-5476170 on Build GRCh38
rs11762179 Chr.7: 5476038-5476038 on Build GRCh38
rs908869086 Chr.7: 5476327-5476327 on Build GRCh38
rs560077228 Chr.7: 5476375-5476375 on Build GRCh38
rs897973394 Chr.7: 5476196-5476196 on Build GRCh38
rs756004922 Chr.7: 5476148-5476148 on Build GRCh38
rs563174679 Chr.7: 5476329-5476329 on Build GRCh38
rs897251146 Chr.7: 5476064-5476064 on Build GRCh38
rs750010577 Chr.7: 5475971-5475971 on Build GRCh38
rs541033380 Chr.7: 5476169-5476169 on Build GRCh38
rs1037036953 Chr.7: 5476272-5476272 on Build GRCh38
rs553342580 Chr.7: 5476003-5476003 on Build GRCh38
rs954325523 Chr.7: 5476096-5476096 on Build GRCh38
rs985757198 Chr.7: 5476099-5476099 on Build GRCh38
rs147283128 Chr.7: 5476095-5476095 on Build GRCh38
rs201525464 Chr.7: 5476257-5476258 on Build GRCh38
rs7789129 Chr.7: 5475993-5475993 on Build GRCh38
rs796377513 Chr.7: 5476298-5476298 on Build GRCh38
rs898053962 Chr.7: 5476335-5476335 on Build GRCh38
rs966338912 Chr.7: 5476367-5476367 on Build GRCh38
rs559380123 Chr.7: 5476179-5476179 on Build GRCh38
rs1032110383 Chr.7: 5476421-5476421 on Build GRCh38
rs530632045 Chr.7: 5476330-5476330 on Build GRCh38
rs991423742 Chr.7: 5476070-5476070 on Build GRCh38
rs945096313 Chr.7: 5475991-5475991 on Build GRCh38
rs751293705 Chr.7: 5476084-5476084 on Build GRCh38
rs377473049 Chr.7: 5476150-5476150 on Build GRCh38
rs551137313 Chr.7: 5476213-5476213 on Build GRCh38
rs544821205 Chr.7: 5476129-5476129 on Build GRCh38
rs972443574 Chr.7: 5476333-5476333 on Build GRCh38
rs1055440314 Chr.7: 5476023-5476023 on Build GRCh38
rs931738770 Chr.7: 5476158-5476158 on Build GRCh38
rs968678390 Chr.7: 5476403-5476403 on Build GRCh38
rs1043545374 Chr.7: 5475992-5475992 on Build GRCh38
rs1003064995 Chr.7: 5476207-5476207 on Build GRCh38
rs1027469403 Chr.7: 5476311-5476311 on Build GRCh38
rs889981386 Chr.7: 5476323-5476323 on Build GRCh38
rs956727228 Chr.7: 5476275-5476275 on Build GRCh38
rs991541407 Chr.7: 5476212-5476212 on Build GRCh38
rs533308551 Chr.7: 5476185-5476185 on Build GRCh38
rs557616089 Chr.7: 5476048-5476048 on Build GRCh38
rs756167927 Chr.7: 5476299-5476299 on Build GRCh38
rs371600464 Chr.7: 5476039-5476039 on Build GRCh38
rs923910124 Chr.7: 5476274-5476274 on Build GRCh38
rs995550745 Chr.7: 5476085-5476085 on Build GRCh38
rs937957832 Chr.7: 5476021-5476021 on Build GRCh38
rs1055089611 Chr.7: 5476200-5476200 on Build GRCh38
rs140503477 Chr.7: 5476336-5476336 on Build GRCh38

View Full Product Details