Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77196455-77196962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC146
Entrez Gene ID
57639
Gene Name
coiled-coil domain containing 146
Gene Aliases
-
UniGene
Hs.113940
Gene Chromosome Location
Chr.7: 77122617-77295204 on Build GRCh38
Transcripts
NM_020879.2 NM_006682.2

Gene Symbol
FGL2
Entrez Gene ID
10875
Gene Name
fibrinogen like 2
Gene Aliases
T49, pT49
UniGene
Hs.520989
Gene Chromosome Location
Chr.7: 77193371-77199833 on Build GRCh38
Transcripts
NM_020879.2 NM_006682.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5774731 c.865A>T p.I289F Chr.7: 77196734-77196734 on Build GRCh38
1698775 c.859G>A p.D287N Chr.7: 77196740-77196740 on Build GRCh38
5737968 c.1054G>A p.D352N Chr.7: 77196545-77196545 on Build GRCh38
7349426 c.933delA p.E311fs*43 Chr.7: 77196666-77196666 on Build GRCh38
3029103 c.652G>A p.A218T Chr.7: 77196947-77196947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548765525 Chr.7: 77196858-77196858 on Build GRCh38
rs780583784 Chr.7: 77196939-77196939 on Build GRCh38
rs777003440 Chr.7: 77196591-77196591 on Build GRCh38
rs1016961575 Chr.7: 77196817-77196817 on Build GRCh38
rs147689299 Chr.7: 77196933-77196933 on Build GRCh38
rs191569909 Chr.7: 77196487-77196487 on Build GRCh38
rs763765559 Chr.7: 77196588-77196588 on Build GRCh38
rs752637532 Chr.7: 77196727-77196727 on Build GRCh38
rs760659595 Chr.7: 77196903-77196903 on Build GRCh38
rs762055560 Chr.7: 77196597-77196597 on Build GRCh38
rs772704744 Chr.7: 77196676-77196676 on Build GRCh38
rs1048619829 Chr.7: 77196748-77196748 on Build GRCh38
rs777734007 Chr.7: 77196834-77196834 on Build GRCh38
rs867952756 Chr.7: 77196545-77196545 on Build GRCh38
rs771291448 Chr.7: 77196544-77196544 on Build GRCh38
rs549177681 Chr.7: 77196553-77196553 on Build GRCh38
rs775844765 Chr.7: 77196680-77196680 on Build GRCh38
rs762149169 Chr.7: 77196704-77196704 on Build GRCh38
rs747588219 Chr.7: 77196506-77196506 on Build GRCh38
rs1013266693 Chr.7: 77196895-77196895 on Build GRCh38
rs1053397439 Chr.7: 77196708-77196708 on Build GRCh38
rs188953293 Chr.7: 77196503-77196503 on Build GRCh38
rs147938006 Chr.7: 77196546-77196546 on Build GRCh38
rs752263777 Chr.7: 77196626-77196626 on Build GRCh38
rs775217486 Chr.7: 77196882-77196882 on Build GRCh38
rs371989577 Chr.7: 77196922-77196922 on Build GRCh38
rs751263824 Chr.7: 77196605-77196605 on Build GRCh38
rs748206225 Chr.7: 77196843-77196843 on Build GRCh38
rs756977540 Chr.7: 77196800-77196800 on Build GRCh38
rs371430811 Chr.7: 77196735-77196735 on Build GRCh38
rs1005562886 Chr.7: 77196783-77196783 on Build GRCh38
rs776808846 Chr.7: 77196694-77196694 on Build GRCh38
rs773321059 Chr.7: 77196856-77196856 on Build GRCh38
rs777392979 Chr.7: 77196521-77196521 on Build GRCh38
rs767324365 Chr.7: 77196613-77196613 on Build GRCh38
rs925148838 Chr.7: 77196500-77196500 on Build GRCh38
rs772021463 Chr.7: 77196741-77196741 on Build GRCh38
rs780495796 Chr.7: 77196830-77196830 on Build GRCh38
rs756815151 Chr.7: 77196662-77196662 on Build GRCh38
rs781470633 Chr.7: 77196839-77196839 on Build GRCh38
rs764744247 Chr.7: 77196930-77196930 on Build GRCh38
rs761218758 Chr.7: 77196928-77196928 on Build GRCh38
rs753328837 Chr.7: 77196798-77196798 on Build GRCh38
rs746412322 Chr.7: 77196556-77196556 on Build GRCh38
rs770119287 Chr.7: 77196855-77196855 on Build GRCh38
rs768858175 Chr.7: 77196691-77196691 on Build GRCh38
rs762871890 Chr.7: 77196495-77196497 on Build GRCh38
rs900095022 Chr.7: 77196690-77196690 on Build GRCh38
rs569344238 Chr.7: 77196568-77196568 on Build GRCh38
rs868604891 Chr.7: 77196658-77196658 on Build GRCh38
rs993060186 Chr.7: 77196523-77196523 on Build GRCh38
rs751564138 Chr.7: 77196934-77196935 on Build GRCh38
rs758025210 Chr.7: 77196937-77196937 on Build GRCh38
rs145870073 Chr.7: 77196543-77196543 on Build GRCh38
rs374306688 Chr.7: 77196829-77196829 on Build GRCh38
rs368142215 Chr.7: 77196838-77196838 on Build GRCh38
rs747458103 Chr.7: 77196689-77196689 on Build GRCh38
rs766228164 Chr.7: 77196601-77196601 on Build GRCh38
rs765354770 Chr.7: 77196712-77196712 on Build GRCh38
rs748971623 Chr.7: 77196652-77196652 on Build GRCh38
rs150013489 Chr.7: 77196636-77196636 on Build GRCh38
rs199775926 Chr.7: 77196606-77196606 on Build GRCh38
rs755801771 Chr.7: 77196511-77196511 on Build GRCh38
rs994124675 Chr.7: 77196870-77196870 on Build GRCh38
rs915669291 Chr.7: 77196528-77196528 on Build GRCh38
rs778428328 Chr.7: 77196822-77196822 on Build GRCh38
rs751515083 Chr.7: 77196499-77196499 on Build GRCh38
rs201777914 Chr.7: 77196825-77196825 on Build GRCh38
rs767286995 Chr.7: 77196723-77196723 on Build GRCh38
rs763617359 Chr.7: 77196740-77196740 on Build GRCh38
rs376169671 Chr.7: 77196717-77196717 on Build GRCh38
rs370416718 Chr.7: 77196668-77196668 on Build GRCh38
rs764464982 Chr.7: 77196574-77196574 on Build GRCh38
rs140799198 Chr.7: 77196501-77196501 on Build GRCh38
rs368742430 Chr.7: 77196719-77196719 on Build GRCh38
rs960459619 Chr.7: 77196869-77196869 on Build GRCh38
rs998400365 Chr.7: 77196738-77196738 on Build GRCh38
rs3093278 Chr.7: 77196669-77196669 on Build GRCh38

View Full Product Details