Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 9138429-9138927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R3B
Entrez Gene ID
79660
Gene Name
protein phosphatase 1 regulatory subunit 3B
Gene Aliases
GL, PPP1R4, PTG
UniGene
Hs.458513
Gene Chromosome Location
Chr.8: 9136254-9151642 on Build GRCh38
Transcripts
NM_001201329.1 NM_024607.3 XM_006716253.3 XM_017013835.1

SNP Detail

SNP ID Chromosome Location
rs187093130 Chr.8: 9138500-9138500 on Build GRCh38
rs915977664 Chr.8: 9138871-9138871 on Build GRCh38
rs745913774 Chr.8: 9138483-9138483 on Build GRCh38
rs1017217254 Chr.8: 9138603-9138603 on Build GRCh38
rs191146816 Chr.8: 9138522-9138522 on Build GRCh38
rs148178016 Chr.8: 9138874-9138874 on Build GRCh38
rs976199797 Chr.8: 9138455-9138455 on Build GRCh38
rs991254791 Chr.8: 9138857-9138857 on Build GRCh38
rs889465045 Chr.8: 9138775-9138775 on Build GRCh38
rs768835363 Chr.8: 9138664-9138664 on Build GRCh38
rs549755541 Chr.8: 9138520-9138520 on Build GRCh38
rs186755985 Chr.8: 9138688-9138688 on Build GRCh38
rs925276667 Chr.8: 9138682-9138682 on Build GRCh38
rs116475566 Chr.8: 9138648-9138648 on Build GRCh38
rs959484435 Chr.8: 9138855-9138855 on Build GRCh38
rs979450696 Chr.8: 9138491-9138491 on Build GRCh38
rs1032875387 Chr.8: 9138838-9138838 on Build GRCh38
rs968501184 Chr.8: 9138815-9138815 on Build GRCh38
rs330912 Chr.8: 9138784-9138784 on Build GRCh38
rs1020110612 Chr.8: 9138643-9138643 on Build GRCh38
rs764760340 Chr.8: 9138807-9138807 on Build GRCh38
rs1037162090 Chr.8: 9138556-9138556 on Build GRCh38
rs558076825 Chr.8: 9138700-9138700 on Build GRCh38
rs1048060918 Chr.8: 9138587-9138587 on Build GRCh38
rs867101246 Chr.8: 9138721-9138721 on Build GRCh38
rs996585055 Chr.8: 9138789-9138794 on Build GRCh38
rs950151531 Chr.8: 9138901-9138904 on Build GRCh38
rs931080960 Chr.8: 9138757-9138757 on Build GRCh38
rs749832378 Chr.8: 9138644-9138644 on Build GRCh38
rs193092593 Chr.8: 9138869-9138869 on Build GRCh38
rs370467786 Chr.8: 9138502-9138502 on Build GRCh38
rs137922958 Chr.8: 9138701-9138701 on Build GRCh38
rs11777327 Chr.8: 9138819-9138819 on Build GRCh38
rs1040712867 Chr.8: 9138794-9138794 on Build GRCh38
rs971402411 Chr.8: 9138886-9138886 on Build GRCh38
rs1010112144 Chr.8: 9138629-9138629 on Build GRCh38
rs768202900 Chr.8: 9138675-9138675 on Build GRCh38
rs535773881 Chr.8: 9138618-9138618 on Build GRCh38
rs1027906336 Chr.8: 9138634-9138634 on Build GRCh38
rs936062810 Chr.8: 9138695-9138695 on Build GRCh38
rs143851762 Chr.8: 9138726-9138726 on Build GRCh38
rs914022270 Chr.8: 9138470-9138470 on Build GRCh38
rs774788859 Chr.8: 9138699-9138699 on Build GRCh38
rs35755658 Chr.8: 9138454-9138454 on Build GRCh38
rs967644211 Chr.8: 9138657-9138657 on Build GRCh38
rs896935252 Chr.8: 9138563-9138563 on Build GRCh38
rs330911 Chr.8: 9138763-9138763 on Build GRCh38
rs1024243522 Chr.8: 9138528-9138528 on Build GRCh38
rs776302598 Chr.8: 9138779-9138779 on Build GRCh38
rs149458764 Chr.8: 9138702-9138702 on Build GRCh38
rs978080493 Chr.8: 9138836-9138836 on Build GRCh38
rs955153217 Chr.8: 9138797-9138797 on Build GRCh38
rs182153814 Chr.8: 9138589-9138589 on Build GRCh38
rs765261974 Chr.8: 9138755-9138756 on Build GRCh38
rs1006809406 Chr.8: 9138611-9138611 on Build GRCh38
rs73526121 Chr.8: 9138526-9138526 on Build GRCh38
rs752568231 Chr.8: 9138812-9138812 on Build GRCh38
rs979489909 Chr.8: 9138661-9138661 on Build GRCh38
rs774827604 Chr.8: 9138504-9138506 on Build GRCh38
rs576306096 Chr.8: 9138616-9138616 on Build GRCh38
rs972934800 Chr.8: 9138905-9138905 on Build GRCh38
rs1045783669 Chr.8: 9138730-9138730 on Build GRCh38
rs1010143458 Chr.8: 9138808-9138808 on Build GRCh38
rs548505854 Chr.8: 9138796-9138796 on Build GRCh38
rs1038170834 Chr.8: 9138765-9138765 on Build GRCh38
rs887051104 Chr.8: 9138566-9138566 on Build GRCh38
rs977610511 Chr.8: 9138670-9138670 on Build GRCh38
rs1024963576 Chr.8: 9138884-9138884 on Build GRCh38
rs926122594 Chr.8: 9138680-9138680 on Build GRCh38
rs918724810 Chr.8: 9138732-9138732 on Build GRCh38
rs901108443 Chr.8: 9138801-9138801 on Build GRCh38
rs532175600 Chr.8: 9138584-9138584 on Build GRCh38
rs1037191482 Chr.8: 9138729-9138729 on Build GRCh38
rs957786841 Chr.8: 9138852-9138852 on Build GRCh38
rs146723513 Chr.8: 9138495-9138496 on Build GRCh38

View Full Product Details