Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63038519-63039057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGH
Entrez Gene ID
8836
Gene Name
gamma-glutamyl hydrolase
Gene Aliases
GH
UniGene
Hs.78619
Gene Chromosome Location
Chr.8: 63015079-63039051 on Build GRCh38
Transcripts
NM_003878.2 XM_011517623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670693 c.82G>A p.G28S Chr.8: 63038687-63038687 on Build GRCh38
6421217 c.64G>T p.E22* Chr.8: 63038705-63038705 on Build GRCh38
7095041 c.49G>C p.G17R Chr.8: 63038720-63038720 on Build GRCh38
4423693 c.98A>T p.K33M Chr.8: 63038671-63038671 on Build GRCh38
4686860 c.74G>A p.R25K Chr.8: 63038695-63038695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954194407 Chr.8: 63038592-63038592 on Build GRCh38
rs748715651 Chr.8: 63038752-63038752 on Build GRCh38
rs976998115 Chr.8: 63038567-63038567 on Build GRCh38
rs944135091 Chr.8: 63038629-63038629 on Build GRCh38
rs894183581 Chr.8: 63039031-63039031 on Build GRCh38
rs971526611 Chr.8: 63039012-63039013 on Build GRCh38
rs546748531 Chr.8: 63038865-63038865 on Build GRCh38
rs945905875 Chr.8: 63038900-63038900 on Build GRCh38
rs965763282 Chr.8: 63038563-63038563 on Build GRCh38
rs751485283 Chr.8: 63038637-63038637 on Build GRCh38
rs140038759 Chr.8: 63038864-63038864 on Build GRCh38
rs559618319 Chr.8: 63038862-63038862 on Build GRCh38
rs867973444 Chr.8: 63038826-63038826 on Build GRCh38
rs909046374 Chr.8: 63038936-63038936 on Build GRCh38
rs931788688 Chr.8: 63038810-63038810 on Build GRCh38
rs755522328 Chr.8: 63038736-63038737 on Build GRCh38
rs563081883 Chr.8: 63038614-63038614 on Build GRCh38
rs750144213 Chr.8: 63038621-63038621 on Build GRCh38
rs768824343 Chr.8: 63038745-63038745 on Build GRCh38
rs752518189 Chr.8: 63038767-63038767 on Build GRCh38
rs747177333 Chr.8: 63038664-63038664 on Build GRCh38
rs940562912 Chr.8: 63038940-63038940 on Build GRCh38
rs567771154 Chr.8: 63038555-63038555 on Build GRCh38
rs1040167750 Chr.8: 63038632-63038632 on Build GRCh38
rs758611939 Chr.8: 63038734-63038734 on Build GRCh38
rs1049725699 Chr.8: 63038947-63038947 on Build GRCh38
rs1055500224 Chr.8: 63039020-63039020 on Build GRCh38
rs774186370 Chr.8: 63038761-63038761 on Build GRCh38
rs558675099 Chr.8: 63038824-63038824 on Build GRCh38
rs773716886 Chr.8: 63038756-63038756 on Build GRCh38
rs763479722 Chr.8: 63038757-63038757 on Build GRCh38
rs796447883 Chr.8: 63038607-63038607 on Build GRCh38
rs1028563576 Chr.8: 63038809-63038809 on Build GRCh38
rs914824586 Chr.8: 63038768-63038768 on Build GRCh38
rs1017207175 Chr.8: 63038979-63038979 on Build GRCh38
rs201285211 Chr.8: 63038811-63038811 on Build GRCh38
rs190945668 Chr.8: 63038557-63038557 on Build GRCh38
rs990016194 Chr.8: 63039028-63039028 on Build GRCh38
rs1021466586 Chr.8: 63038782-63038782 on Build GRCh38
rs369584231 Chr.8: 63038648-63038648 on Build GRCh38
rs944112799 Chr.8: 63038777-63038777 on Build GRCh38
rs949744003 Chr.8: 63038863-63038863 on Build GRCh38
rs899942393 Chr.8: 63038791-63038791 on Build GRCh38
rs754939974 Chr.8: 63038591-63038591 on Build GRCh38
rs984309426 Chr.8: 63038596-63038596 on Build GRCh38
rs946034846 Chr.8: 63038601-63038601 on Build GRCh38
rs926921936 Chr.8: 63039017-63039017 on Build GRCh38
rs927458756 Chr.8: 63038543-63038543 on Build GRCh38
rs1005769501 Chr.8: 63038959-63038959 on Build GRCh38
rs780309319 Chr.8: 63038742-63038742 on Build GRCh38
rs906883533 Chr.8: 63038828-63038828 on Build GRCh38
rs965745195 Chr.8: 63038868-63038868 on Build GRCh38
rs767556657 Chr.8: 63038804-63038805 on Build GRCh38
rs11545077 Chr.8: 63038678-63038678 on Build GRCh38
rs2736678 Chr.8: 63038842-63038842 on Build GRCh38
rs762708836 Chr.8: 63038695-63038695 on Build GRCh38
rs753615495 Chr.8: 63038774-63038774 on Build GRCh38
rs912534896 Chr.8: 63038867-63038867 on Build GRCh38
rs1014630480 Chr.8: 63038708-63038708 on Build GRCh38
rs769668813 Chr.8: 63038645-63038645 on Build GRCh38
rs554238614 Chr.8: 63038799-63038799 on Build GRCh38
rs891203369 Chr.8: 63038622-63038622 on Build GRCh38
rs1048890363 Chr.8: 63038902-63038902 on Build GRCh38
rs762056917 Chr.8: 63038679-63038679 on Build GRCh38
rs571549892 Chr.8: 63038583-63038583 on Build GRCh38
rs767324259 Chr.8: 63038723-63038723 on Build GRCh38
rs867923310 Chr.8: 63038732-63038732 on Build GRCh38
rs946336252 Chr.8: 63038771-63038771 on Build GRCh38
rs376225287 Chr.8: 63038763-63038763 on Build GRCh38
rs901278772 Chr.8: 63038910-63038911 on Build GRCh38
rs765668706 Chr.8: 63038690-63038690 on Build GRCh38
rs571386201 Chr.8: 63039005-63039005 on Build GRCh38
rs746110197 Chr.8: 63038660-63038660 on Build GRCh38
rs777554108 Chr.8: 63038726-63038726 on Build GRCh38
rs575389093 Chr.8: 63038998-63039003 on Build GRCh38
rs754564165 Chr.8: 63038718-63038718 on Build GRCh38
rs865790840 Chr.8: 63038823-63038823 on Build GRCh38
rs757089623 Chr.8: 63038776-63038776 on Build GRCh38
rs1800909 Chr.8: 63038753-63038753 on Build GRCh38
rs764777758 Chr.8: 63038620-63038620 on Build GRCh38
rs1040205937 Chr.8: 63038779-63038779 on Build GRCh38
rs887715478 Chr.8: 63038674-63038674 on Build GRCh38
rs757946225 Chr.8: 63038633-63038633 on Build GRCh38
rs147536505 Chr.8: 63038597-63038597 on Build GRCh38
rs879536623 Chr.8: 63039013-63039013 on Build GRCh38
rs754968695 Chr.8: 63038820-63038820 on Build GRCh38
rs531939120 Chr.8: 63038661-63038661 on Build GRCh38
rs549910927 Chr.8: 63038993-63038993 on Build GRCh38
rs907143105 Chr.8: 63038962-63038962 on Build GRCh38
rs776968253 Chr.8: 63038749-63038749 on Build GRCh38
rs958523338 Chr.8: 63038845-63038845 on Build GRCh38
rs961560956 Chr.8: 63038997-63038998 on Build GRCh38
rs576207255 Chr.8: 63038806-63038806 on Build GRCh38
rs772951871 Chr.8: 63038692-63038692 on Build GRCh38
rs912566113 Chr.8: 63038775-63038775 on Build GRCh38
rs957042113 Chr.8: 63039021-63039021 on Build GRCh38
rs1012152219 Chr.8: 63038852-63038852 on Build GRCh38
rs536747486 Chr.8: 63039003-63039004 on Build GRCh38
rs765922991 Chr.8: 63038698-63038698 on Build GRCh38
rs781259634 Chr.8: 63038644-63038644 on Build GRCh38
rs546095627 Chr.8: 63038606-63038606 on Build GRCh38
rs764081468 Chr.8: 63038772-63038772 on Build GRCh38
rs1041740983 Chr.8: 63038608-63038608 on Build GRCh38
rs765870198 Chr.8: 63038742-63038750 on Build GRCh38
rs4590435 Chr.8: 63038924-63038924 on Build GRCh38
rs998181375 Chr.8: 63038916-63038916 on Build GRCh38
rs1002952229 Chr.8: 63038985-63038985 on Build GRCh38
rs11545076 Chr.8: 63038892-63038892 on Build GRCh38
rs977359112 Chr.8: 63038773-63038773 on Build GRCh38
rs780221906 Chr.8: 63038783-63038783 on Build GRCh38
rs754877968 Chr.8: 63038641-63038641 on Build GRCh38
rs779734779 Chr.8: 63038634-63038634 on Build GRCh38
rs1011328880 Chr.8: 63039033-63039033 on Build GRCh38
rs766118226 Chr.8: 63038759-63038759 on Build GRCh38
rs1029029246 Chr.8: 63038589-63038589 on Build GRCh38
rs549972560 Chr.8: 63038579-63038579 on Build GRCh38
rs921344481 Chr.8: 63038888-63038888 on Build GRCh38
rs972910947 Chr.8: 63038635-63038635 on Build GRCh38
rs756079503 Chr.8: 63038769-63038769 on Build GRCh38
rs561087716 Chr.8: 63038871-63038871 on Build GRCh38
rs201836906 Chr.8: 63038724-63038724 on Build GRCh38
rs746019566 Chr.8: 63038730-63038730 on Build GRCh38
rs1036270228 Chr.8: 63038956-63038956 on Build GRCh38
rs971098934 Chr.8: 63038719-63038719 on Build GRCh38
rs547764450 Chr.8: 63038860-63038860 on Build GRCh38
rs1024039648 Chr.8: 63039012-63039012 on Build GRCh38
rs1020932271 Chr.8: 63038560-63038560 on Build GRCh38
rs182198219 Chr.8: 63038835-63038835 on Build GRCh38
rs889812120 Chr.8: 63038953-63038953 on Build GRCh38
rs1002574659 Chr.8: 63038834-63038834 on Build GRCh38

View Full Product Details