Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 89325190-89325706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SECISBP2
Entrez Gene ID
79048
Gene Name
SECIS binding protein 2
Gene Aliases
SBP2
UniGene
Hs.59804
Gene Chromosome Location
Chr.9: 89318473-89367117 on Build GRCh38
Transcripts
NM_001282688.1 NM_001282689.1 NM_001282690.1 NM_024077.4 XM_005252196.2 XM_006717282.2 XM_011519000.2 XM_011519001.1 XM_011519002.1 XM_011519003.1 XM_017015122.1 XM_017015123.1 XM_017015124.1 XM_017015125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110970 c.383G>A p.R128Q Chr.9: 89325627-89325627 on Build GRCh38
4480846 c.245C>T p.S82F Chr.9: 89325489-89325489 on Build GRCh38
1110968 c.278C>G p.S93C Chr.9: 89325522-89325522 on Build GRCh38
6013524 c.401C>T p.P134L Chr.9: 89325645-89325645 on Build GRCh38
1110969 c.382C>T p.R128* Chr.9: 89325626-89325626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964425061 Chr.9: 89325522-89325522 on Build GRCh38
rs935722518 Chr.9: 89325240-89325240 on Build GRCh38
rs906762524 Chr.9: 89325440-89325440 on Build GRCh38
rs369918420 Chr.9: 89325536-89325536 on Build GRCh38
rs772199609 Chr.9: 89325532-89325532 on Build GRCh38
rs772557373 Chr.9: 89325637-89325637 on Build GRCh38
rs373079221 Chr.9: 89325563-89325563 on Build GRCh38
rs367928222 Chr.9: 89325414-89325414 on Build GRCh38
rs529644965 Chr.9: 89325515-89325515 on Build GRCh38
rs746037464 Chr.9: 89325529-89325529 on Build GRCh38
rs757133015 Chr.9: 89325627-89325627 on Build GRCh38
rs751352358 Chr.9: 89325517-89325517 on Build GRCh38
rs761754027 Chr.9: 89325385-89325385 on Build GRCh38
rs773115168 Chr.9: 89325549-89325549 on Build GRCh38
rs1055993083 Chr.9: 89325665-89325665 on Build GRCh38
rs1044927396 Chr.9: 89325267-89325267 on Build GRCh38
rs879108630 Chr.9: 89325594-89325594 on Build GRCh38
rs371791200 Chr.9: 89325441-89325441 on Build GRCh38
rs747596372 Chr.9: 89325504-89325504 on Build GRCh38
rs368643206 Chr.9: 89325640-89325640 on Build GRCh38
rs551981883 Chr.9: 89325230-89325230 on Build GRCh38
rs71510294 Chr.9: 89325673-89325673 on Build GRCh38
rs751179748 Chr.9: 89325437-89325437 on Build GRCh38
rs755662391 Chr.9: 89325601-89325601 on Build GRCh38
rs754858358 Chr.9: 89325573-89325573 on Build GRCh38
rs376067869 Chr.9: 89325603-89325603 on Build GRCh38
rs767070826 Chr.9: 89325589-89325589 on Build GRCh38
rs771373162 Chr.9: 89325518-89325518 on Build GRCh38
rs757696851 Chr.9: 89325594-89325596 on Build GRCh38
rs752432903 Chr.9: 89325592-89325592 on Build GRCh38
rs760743400 Chr.9: 89325541-89325541 on Build GRCh38
rs921073040 Chr.9: 89325417-89325417 on Build GRCh38
rs757844295 Chr.9: 89325622-89325622 on Build GRCh38
rs776693254 Chr.9: 89325655-89325655 on Build GRCh38
rs768678703 Chr.9: 89325654-89325654 on Build GRCh38
rs952834034 Chr.9: 89325479-89325479 on Build GRCh38
rs550276253 Chr.9: 89325409-89325409 on Build GRCh38
rs144448321 Chr.9: 89325513-89325513 on Build GRCh38
rs1010279225 Chr.9: 89325374-89325376 on Build GRCh38
rs765235805 Chr.9: 89325420-89325421 on Build GRCh38
rs530980733 Chr.9: 89325343-89325343 on Build GRCh38
rs372635973 Chr.9: 89325543-89325543 on Build GRCh38
rs765148149 Chr.9: 89325390-89325390 on Build GRCh38
rs749859737 Chr.9: 89325516-89325516 on Build GRCh38
rs1055473408 Chr.9: 89325250-89325250 on Build GRCh38
rs779877417 Chr.9: 89325411-89325411 on Build GRCh38
rs1013921613 Chr.9: 89325299-89325299 on Build GRCh38
rs758782931 Chr.9: 89325504-89325519 on Build GRCh38
rs747284759 Chr.9: 89325651-89325651 on Build GRCh38
rs762905999 Chr.9: 89325556-89325556 on Build GRCh38
rs75102435 Chr.9: 89325271-89325271 on Build GRCh38
rs149232886 Chr.9: 89325458-89325458 on Build GRCh38
rs376444493 Chr.9: 89325528-89325528 on Build GRCh38
rs747514658 Chr.9: 89325589-89325590 on Build GRCh38
rs898791863 Chr.9: 89325234-89325234 on Build GRCh38
rs774611487 Chr.9: 89325524-89325524 on Build GRCh38
rs755750135 Chr.9: 89325512-89325512 on Build GRCh38
rs374983317 Chr.9: 89325410-89325410 on Build GRCh38
rs751321557 Chr.9: 89325568-89325568 on Build GRCh38
rs1036691762 Chr.9: 89325564-89325564 on Build GRCh38
rs755250278 Chr.9: 89325416-89325418 on Build GRCh38
rs750429765 Chr.9: 89325394-89325394 on Build GRCh38
rs766210796 Chr.9: 89325660-89325660 on Build GRCh38
rs922546563 Chr.9: 89325531-89325531 on Build GRCh38
rs995683897 Chr.9: 89325262-89325262 on Build GRCh38
rs778322437 Chr.9: 89325519-89325520 on Build GRCh38
rs906036177 Chr.9: 89325268-89325268 on Build GRCh38
rs754076788 Chr.9: 89325375-89325375 on Build GRCh38
rs780692869 Chr.9: 89325476-89325476 on Build GRCh38
rs1025787206 Chr.9: 89325474-89325474 on Build GRCh38
rs994354748 Chr.9: 89325454-89325454 on Build GRCh38
rs570788252 Chr.9: 89325256-89325256 on Build GRCh38
rs377237035 Chr.9: 89325337-89325337 on Build GRCh38
rs62638730 Chr.9: 89325666-89325666 on Build GRCh38
rs779280246 Chr.9: 89325626-89325626 on Build GRCh38
rs1025257788 Chr.9: 89325330-89325330 on Build GRCh38
rs777447200 Chr.9: 89325602-89325602 on Build GRCh38
rs199933213 Chr.9: 89325415-89325415 on Build GRCh38
rs752855637 Chr.9: 89325453-89325453 on Build GRCh38

View Full Product Details