Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100301989-100302495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INVS
Entrez Gene ID
27130
Gene Name
inversin
Gene Aliases
INV, NPH2, NPHP2
UniGene
Hs.558477
Gene Chromosome Location
Chr.9: 100099185-100302175 on Build GRCh38
Transcripts
NM_001318381.1 NM_001318382.1 NM_014425.4 NR_134606.1 NM_001161584.1 NM_017746.3 XM_011518798.1 XM_017014848.1

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
UniGene
Hs.494648
Gene Chromosome Location
Chr.9: 100302075-100352977 on Build GRCh38
Transcripts
NM_001318381.1 NM_001318382.1 NM_014425.4 NR_134606.1 NM_001161584.1 NM_017746.3 XM_011518798.1 XM_017014848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5875238 c.2695A>G p.I899V Chr.9: 100302238-100302238 on Build GRCh38
7279206 c.2763G>A p.G921G Chr.9: 100302218-100302218 on Build GRCh38
6114588 c.2676T>G p.H892Q Chr.9: 100302257-100302257 on Build GRCh38
1228983 c.2707T>C p.W903R Chr.9: 100302274-100302274 on Build GRCh38
5875240 c.2689G>A p.V897M Chr.9: 100302244-100302244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748189853 Chr.9: 100302159-100302161 on Build GRCh38
rs758466284 Chr.9: 100302319-100302320 on Build GRCh38
rs768618117 Chr.9: 100302344-100302344 on Build GRCh38
rs1033434697 Chr.9: 100302302-100302302 on Build GRCh38
rs756627177 Chr.9: 100302288-100302288 on Build GRCh38
rs925311972 Chr.9: 100302354-100302354 on Build GRCh38
rs759926008 Chr.9: 100302307-100302308 on Build GRCh38
rs193026865 Chr.9: 100302213-100302213 on Build GRCh38
rs751715524 Chr.9: 100302173-100302173 on Build GRCh38
rs35689659 Chr.9: 100302053-100302054 on Build GRCh38
rs549569443 Chr.9: 100302429-100302429 on Build GRCh38
rs557542255 Chr.9: 100302091-100302091 on Build GRCh38
rs374400521 Chr.9: 100302184-100302184 on Build GRCh38
rs762929771 Chr.9: 100302151-100302151 on Build GRCh38
rs560527106 Chr.9: 100302080-100302080 on Build GRCh38
rs758193340 Chr.9: 100302320-100302320 on Build GRCh38
rs765054405 Chr.9: 100302145-100302145 on Build GRCh38
rs941813872 Chr.9: 100302045-100302045 on Build GRCh38
rs773409439 Chr.9: 100302146-100302146 on Build GRCh38
rs931651556 Chr.9: 100302259-100302259 on Build GRCh38
rs373037476 Chr.9: 100302245-100302245 on Build GRCh38
rs746977498 Chr.9: 100302340-100302340 on Build GRCh38
rs748947632 Chr.9: 100302218-100302218 on Build GRCh38
rs907608984 Chr.9: 100302034-100302034 on Build GRCh38
rs779308268 Chr.9: 100302149-100302150 on Build GRCh38
rs182875991 Chr.9: 100302241-100302241 on Build GRCh38
rs375700935 Chr.9: 100302200-100302200 on Build GRCh38
rs974677127 Chr.9: 100302455-100302455 on Build GRCh38
rs773329485 Chr.9: 100302254-100302254 on Build GRCh38
rs41274953 Chr.9: 100302307-100302307 on Build GRCh38
rs756678341 Chr.9: 100302316-100302316 on Build GRCh38
rs375507071 Chr.9: 100302352-100302352 on Build GRCh38
rs778275379 Chr.9: 100302200-100302201 on Build GRCh38
rs983299516 Chr.9: 100302210-100302210 on Build GRCh38
rs886703262 Chr.9: 100302212-100302212 on Build GRCh38
rs1053736452 Chr.9: 100302118-100302118 on Build GRCh38
rs554677331 Chr.9: 100302042-100302042 on Build GRCh38
rs775141829 Chr.9: 100302327-100302328 on Build GRCh38
rs202112876 Chr.9: 100302314-100302314 on Build GRCh38
rs376447766 Chr.9: 100302181-100302181 on Build GRCh38
rs71498737 Chr.9: 100302054-100302055 on Build GRCh38
rs921632300 Chr.9: 100302057-100302057 on Build GRCh38
rs1049983400 Chr.9: 100302083-100302084 on Build GRCh38
rs7472 Chr.9: 100302248-100302248 on Build GRCh38
rs753265472 Chr.9: 100302286-100302286 on Build GRCh38
rs770593519 Chr.9: 100302231-100302231 on Build GRCh38
rs767939276 Chr.9: 100302284-100302284 on Build GRCh38
rs146292456 Chr.9: 100302109-100302110 on Build GRCh38
rs766678620 Chr.9: 100302264-100302264 on Build GRCh38
rs776900829 Chr.9: 100302280-100302280 on Build GRCh38
rs369754553 Chr.9: 100302217-100302217 on Build GRCh38
rs1035618687 Chr.9: 100302198-100302198 on Build GRCh38
rs771804501 Chr.9: 100302244-100302244 on Build GRCh38
rs1039090132 Chr.9: 100302085-100302085 on Build GRCh38
rs891255394 Chr.9: 100302084-100302084 on Build GRCh38
rs750614420 Chr.9: 100302176-100302176 on Build GRCh38
rs753210570 Chr.9: 100302208-100302208 on Build GRCh38
rs760597143 Chr.9: 100302100-100302100 on Build GRCh38
rs749420249 Chr.9: 100302347-100302347 on Build GRCh38
rs895713545 Chr.9: 100302378-100302378 on Build GRCh38
rs771306898 Chr.9: 100302234-100302235 on Build GRCh38
rs760004585 Chr.9: 100302280-100302280 on Build GRCh38
rs371643437 Chr.9: 100302147-100302147 on Build GRCh38
rs760592973 Chr.9: 100302246-100302246 on Build GRCh38
rs574415078 Chr.9: 100302066-100302066 on Build GRCh38
rs754066009 Chr.9: 100302140-100302141 on Build GRCh38
rs781773056 Chr.9: 100302202-100302202 on Build GRCh38
rs766640928 Chr.9: 100302171-100302171 on Build GRCh38
rs925291725 Chr.9: 100302027-100302027 on Build GRCh38
rs891860282 Chr.9: 100302139-100302140 on Build GRCh38
rs774732540 Chr.9: 100302353-100302353 on Build GRCh38
rs745693527 Chr.9: 100302243-100302243 on Build GRCh38
rs530887406 Chr.9: 100302341-100302341 on Build GRCh38
rs371096677 Chr.9: 100302318-100302318 on Build GRCh38
rs773206028 Chr.9: 100302346-100302346 on Build GRCh38
rs535918870 Chr.9: 100302363-100302363 on Build GRCh38
rs34169614 Chr.9: 100302317-100302317 on Build GRCh38
rs189132127 Chr.9: 100302090-100302090 on Build GRCh38
rs371133850 Chr.9: 100302143-100302144 on Build GRCh38
rs763136487 Chr.9: 100302258-100302258 on Build GRCh38
rs369973072 Chr.9: 100302183-100302183 on Build GRCh38
rs747529174 Chr.9: 100302233-100302233 on Build GRCh38
rs779820090 Chr.9: 100302327-100302327 on Build GRCh38
rs377185856 Chr.9: 100302269-100302269 on Build GRCh38

View Full Product Details