Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104098254-104098767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMC2
Entrez Gene ID
10592
Gene Name
structural maintenance of chromosomes 2
Gene Aliases
CAP-E, CAPE, SMC-2, SMC2L1
UniGene
Hs.119023
Gene Chromosome Location
Chr.9: 104093808-104141419 on Build GRCh38
Transcripts
NM_001042550.1 NM_001042551.1 NM_001265602.1 NM_006444.2 XM_006716933.3 XM_011518148.2 XM_011518149.2 XM_011518150.2 XM_011518151.2 XM_011518152.2 XM_011518153.1 XM_017014206.1 XM_017014207.1 XM_017014208.1 XM_017014209.1 XM_017014210.1 XM_017014211.1 XM_017014212.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6785174 c.400G>A p.V134I Chr.9: 104098527-104098527 on Build GRCh38
6785199 c.407T>G p.L136R Chr.9: 104098534-104098534 on Build GRCh38
3929706 c.377G>C p.R126T Chr.9: 104098504-104098504 on Build GRCh38
3745731 c.333T>C p.G111G Chr.9: 104098460-104098460 on Build GRCh38
6550316 c.385G>T p.D129Y Chr.9: 104098512-104098512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049652255 Chr.9: 104098511-104098511 on Build GRCh38
rs377071933 Chr.9: 104098565-104098565 on Build GRCh38
rs773917860 Chr.9: 104098694-104098694 on Build GRCh38
rs778733809 Chr.9: 104098454-104098454 on Build GRCh38
rs368963452 Chr.9: 104098481-104098481 on Build GRCh38
rs757983427 Chr.9: 104098472-104098472 on Build GRCh38
rs780819035 Chr.9: 104098508-104098508 on Build GRCh38
rs1013548505 Chr.9: 104098611-104098611 on Build GRCh38
rs764348994 Chr.9: 104098424-104098424 on Build GRCh38
rs772523355 Chr.9: 104098343-104098343 on Build GRCh38
rs568501472 Chr.9: 104098312-104098312 on Build GRCh38
rs376791512 Chr.9: 104098453-104098453 on Build GRCh38
rs747879470 Chr.9: 104098614-104098614 on Build GRCh38
rs778183578 Chr.9: 104098315-104098315 on Build GRCh38
rs1016355743 Chr.9: 104098289-104098289 on Build GRCh38
rs199689144 Chr.9: 104098420-104098420 on Build GRCh38
rs544194442 Chr.9: 104098579-104098579 on Build GRCh38
rs993447417 Chr.9: 104098583-104098583 on Build GRCh38
rs774567646 Chr.9: 104098522-104098522 on Build GRCh38
rs1006087229 Chr.9: 104098551-104098551 on Build GRCh38
rs1008551755 Chr.9: 104098545-104098545 on Build GRCh38
rs779678324 Chr.9: 104098474-104098474 on Build GRCh38
rs961182355 Chr.9: 104098300-104098300 on Build GRCh38
rs982763880 Chr.9: 104098730-104098730 on Build GRCh38
rs373195767 Chr.9: 104098517-104098517 on Build GRCh38
rs749896885 Chr.9: 104098465-104098465 on Build GRCh38
rs952869231 Chr.9: 104098681-104098681 on Build GRCh38
rs541559030 Chr.9: 104098437-104098437 on Build GRCh38
rs367624269 Chr.9: 104098409-104098409 on Build GRCh38
rs767275140 Chr.9: 104098738-104098738 on Build GRCh38
rs902497353 Chr.9: 104098421-104098421 on Build GRCh38
rs79656676 Chr.9: 104098455-104098455 on Build GRCh38
rs372132522 Chr.9: 104098442-104098442 on Build GRCh38
rs960310252 Chr.9: 104098389-104098389 on Build GRCh38
rs769575161 Chr.9: 104098615-104098615 on Build GRCh38
rs775009187 Chr.9: 104098581-104098581 on Build GRCh38
rs772127542 Chr.9: 104098548-104098548 on Build GRCh38
rs114913341 Chr.9: 104098708-104098708 on Build GRCh38
rs368804202 Chr.9: 104098410-104098410 on Build GRCh38
rs991077605 Chr.9: 104098384-104098384 on Build GRCh38
rs370657190 Chr.9: 104098589-104098589 on Build GRCh38
rs764914282 Chr.9: 104098598-104098598 on Build GRCh38
rs149445715 Chr.9: 104098302-104098302 on Build GRCh38
rs115415227 Chr.9: 104098599-104098599 on Build GRCh38
rs767948559 Chr.9: 104098413-104098413 on Build GRCh38
rs1037747940 Chr.9: 104098559-104098559 on Build GRCh38
rs748751115 Chr.9: 104098676-104098676 on Build GRCh38
rs200585797 Chr.9: 104098398-104098398 on Build GRCh38
rs751021508 Chr.9: 104098586-104098591 on Build GRCh38
rs554183025 Chr.9: 104098518-104098518 on Build GRCh38
rs956412881 Chr.9: 104098695-104098695 on Build GRCh38
rs761306214 Chr.9: 104098423-104098423 on Build GRCh38
rs2274724 Chr.9: 104098287-104098287 on Build GRCh38
rs775579576 Chr.9: 104098396-104098396 on Build GRCh38
rs759631833 Chr.9: 104098411-104098411 on Build GRCh38
rs201849086 Chr.9: 104098592-104098592 on Build GRCh38
rs761525066 Chr.9: 104098704-104098704 on Build GRCh38
rs751120592 Chr.9: 104098601-104098601 on Build GRCh38
rs771357122 Chr.9: 104098575-104098575 on Build GRCh38
rs529518385 Chr.9: 104098576-104098576 on Build GRCh38
rs781210735 Chr.9: 104098610-104098610 on Build GRCh38
rs530633123 Chr.9: 104098505-104098505 on Build GRCh38
rs750164954 Chr.9: 104098600-104098600 on Build GRCh38
rs552938226 Chr.9: 104098641-104098641 on Build GRCh38
rs776547198 Chr.9: 104098402-104098402 on Build GRCh38
rs746903710 Chr.9: 104098495-104098495 on Build GRCh38
rs375955893 Chr.9: 104098408-104098408 on Build GRCh38
rs532833355 Chr.9: 104098609-104098609 on Build GRCh38
rs1023226193 Chr.9: 104098660-104098660 on Build GRCh38
rs143439504 Chr.9: 104098526-104098526 on Build GRCh38
rs113313889 Chr.9: 104098658-104098658 on Build GRCh38
rs10991107 Chr.9: 104098688-104098688 on Build GRCh38
rs566131393 Chr.9: 104098651-104098651 on Build GRCh38
rs61755310 Chr.9: 104098498-104098498 on Build GRCh38
rs916729118 Chr.9: 104098702-104098702 on Build GRCh38

View Full Product Details