Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105362731-105363161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A1
Entrez Gene ID
23446
Gene Name
solute carrier family 44 member 1
Gene Aliases
CD92, CDW92, CHTL1, CTL1
UniGene
Hs.573495
Gene Chromosome Location
Chr.9: 105244608-105438504 on Build GRCh38
Transcripts
NM_001286730.1 NM_080546.4 XM_005251855.4 XM_006717027.3 XM_006717028.3 XM_006717029.2 XM_017014559.1 XM_017014560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6884453 c.1014C>T p.P338P Chr.9: 105362934-105362934 on Build GRCh38
4727392 c.958T>C p.L320L Chr.9: 105362878-105362878 on Build GRCh38
1103557 c.1079G>A p.G360D Chr.9: 105362999-105362999 on Build GRCh38
6782292 c.1087G>A p.G363S Chr.9: 105363007-105363007 on Build GRCh38
6782291 c.1082C>G p.T361S Chr.9: 105363002-105363002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551917120 Chr.9: 105362785-105362787 on Build GRCh38
rs755921134 Chr.9: 105362789-105362789 on Build GRCh38
rs905145114 Chr.9: 105362915-105362915 on Build GRCh38
rs888838615 Chr.9: 105363071-105363071 on Build GRCh38
rs972382366 Chr.9: 105363096-105363096 on Build GRCh38
rs1031572496 Chr.9: 105362767-105362767 on Build GRCh38
rs964333796 Chr.9: 105363131-105363131 on Build GRCh38
rs193178204 Chr.9: 105363087-105363087 on Build GRCh38
rs1010991515 Chr.9: 105363017-105363017 on Build GRCh38
rs1010605439 Chr.9: 105362793-105362793 on Build GRCh38
rs766236604 Chr.9: 105362806-105362807 on Build GRCh38
rs953054559 Chr.9: 105362815-105362815 on Build GRCh38
rs1045057821 Chr.9: 105362894-105362894 on Build GRCh38
rs771542290 Chr.9: 105363023-105363023 on Build GRCh38
rs1003725685 Chr.9: 105362758-105362758 on Build GRCh38
rs771467103 Chr.9: 105363001-105363001 on Build GRCh38
rs753184710 Chr.9: 105362956-105362956 on Build GRCh38
rs966343119 Chr.9: 105362851-105362851 on Build GRCh38
rs141625866 Chr.9: 105362943-105362943 on Build GRCh38
rs1026195022 Chr.9: 105363132-105363132 on Build GRCh38
rs763860457 Chr.9: 105362794-105362794 on Build GRCh38
rs145083133 Chr.9: 105362857-105362857 on Build GRCh38
rs138697456 Chr.9: 105363002-105363002 on Build GRCh38
rs371768444 Chr.9: 105363020-105363020 on Build GRCh38
rs139580101 Chr.9: 105362874-105362874 on Build GRCh38
rs770733177 Chr.9: 105362871-105362871 on Build GRCh38
rs906812826 Chr.9: 105362931-105362931 on Build GRCh38
rs776971940 Chr.9: 105363124-105363124 on Build GRCh38
rs766558103 Chr.9: 105362937-105362937 on Build GRCh38
rs778237402 Chr.9: 105362980-105362980 on Build GRCh38
rs150556036 Chr.9: 105362817-105362817 on Build GRCh38
rs570006425 Chr.9: 105362821-105362821 on Build GRCh38
rs1022019716 Chr.9: 105363070-105363070 on Build GRCh38
rs751670520 Chr.9: 105362829-105362829 on Build GRCh38
rs763889810 Chr.9: 105362905-105362905 on Build GRCh38
rs750576469 Chr.9: 105362804-105362804 on Build GRCh38
rs756596917 Chr.9: 105362961-105362961 on Build GRCh38
rs781199771 Chr.9: 105362951-105362951 on Build GRCh38
rs775490518 Chr.9: 105362887-105362887 on Build GRCh38
rs763121195 Chr.9: 105362932-105362932 on Build GRCh38
rs368894258 Chr.9: 105362852-105362852 on Build GRCh38
rs774872299 Chr.9: 105363073-105363073 on Build GRCh38
rs139026951 Chr.9: 105362863-105362863 on Build GRCh38
rs752622142 Chr.9: 105362784-105362784 on Build GRCh38
rs147994302 Chr.9: 105362928-105362928 on Build GRCh38
rs748663384 Chr.9: 105362856-105362856 on Build GRCh38
rs955437682 Chr.9: 105363042-105363042 on Build GRCh38
rs951860324 Chr.9: 105363136-105363136 on Build GRCh38
rs759503510 Chr.9: 105362775-105362775 on Build GRCh38
rs1020352154 Chr.9: 105363037-105363037 on Build GRCh38
rs774875168 Chr.9: 105363043-105363043 on Build GRCh38
rs142817213 Chr.9: 105363006-105363006 on Build GRCh38
rs966029022 Chr.9: 105363095-105363095 on Build GRCh38
rs778006877 Chr.9: 105362858-105362858 on Build GRCh38
rs767456441 Chr.9: 105362776-105362776 on Build GRCh38
rs749635231 Chr.9: 105362993-105362993 on Build GRCh38
rs911826695 Chr.9: 105362836-105362836 on Build GRCh38
rs145882369 Chr.9: 105362945-105362945 on Build GRCh38
rs755092832 Chr.9: 105362842-105362842 on Build GRCh38
rs760611501 Chr.9: 105363055-105363055 on Build GRCh38
rs760532547 Chr.9: 105362895-105362895 on Build GRCh38
rs143807755 Chr.9: 105362886-105362886 on Build GRCh38

View Full Product Details