Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70201806-70202310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGAT2L6
Entrez Gene ID
347516
Gene Name
diacylglycerol O-acyltransferase 2 like 6
Gene Aliases
DC3
UniGene
Hs.454355
Gene Chromosome Location
Chr.X: 70177483-70205703 on Build GRCh38
Transcripts
NM_198512.2

SNP Detail

SNP ID Chromosome Location
rs768722303 Chr.X: 70202079-70202079 on Build GRCh38
rs771887078 Chr.X: 70201866-70201866 on Build GRCh38
rs372836202 Chr.X: 70202057-70202057 on Build GRCh38
rs759342762 Chr.X: 70202095-70202095 on Build GRCh38
rs780102567 Chr.X: 70201905-70201905 on Build GRCh38
rs762086023 Chr.X: 70202147-70202147 on Build GRCh38
rs754945430 Chr.X: 70201932-70201932 on Build GRCh38
rs746922269 Chr.X: 70201954-70201954 on Build GRCh38
rs368517131 Chr.X: 70201950-70201950 on Build GRCh38
rs766300465 Chr.X: 70201913-70201913 on Build GRCh38
rs781632348 Chr.X: 70201990-70201990 on Build GRCh38
rs145626540 Chr.X: 70202039-70202039 on Build GRCh38
rs371876539 Chr.X: 70201958-70201958 on Build GRCh38
rs775292059 Chr.X: 70202212-70202212 on Build GRCh38
rs767179787 Chr.X: 70201897-70201897 on Build GRCh38
rs375346876 Chr.X: 70201900-70201900 on Build GRCh38
rs138262705 Chr.X: 70202048-70202048 on Build GRCh38
rs971969935 Chr.X: 70201904-70201904 on Build GRCh38
rs749546854 Chr.X: 70202222-70202222 on Build GRCh38
rs1023037256 Chr.X: 70201874-70201874 on Build GRCh38
rs778818580 Chr.X: 70201933-70201933 on Build GRCh38
rs767995320 Chr.X: 70201880-70201880 on Build GRCh38
rs771682108 Chr.X: 70202049-70202049 on Build GRCh38
rs777068649 Chr.X: 70201850-70201850 on Build GRCh38
rs1034838290 Chr.X: 70201887-70201887 on Build GRCh38
rs936552863 Chr.X: 70202255-70202255 on Build GRCh38
rs945438064 Chr.X: 70201886-70201886 on Build GRCh38
rs773690172 Chr.X: 70202197-70202197 on Build GRCh38
rs989434405 Chr.X: 70202273-70202273 on Build GRCh38
rs971753383 Chr.X: 70202194-70202194 on Build GRCh38
rs141887110 Chr.X: 70201998-70201998 on Build GRCh38
rs961563567 Chr.X: 70202192-70202192 on Build GRCh38
rs1053371691 Chr.X: 70202145-70202145 on Build GRCh38
rs905057868 Chr.X: 70201882-70201882 on Build GRCh38
rs913727591 Chr.X: 70202284-70202284 on Build GRCh38
rs934942996 Chr.X: 70202142-70202142 on Build GRCh38
rs1046553979 Chr.X: 70201894-70201894 on Build GRCh38
rs368849374 Chr.X: 70201876-70201876 on Build GRCh38
rs752789423 Chr.X: 70202109-70202109 on Build GRCh38
rs370823085 Chr.X: 70201863-70201863 on Build GRCh38
rs904170550 Chr.X: 70202067-70202067 on Build GRCh38
rs768993529 Chr.X: 70201871-70201880 on Build GRCh38
rs954378002 Chr.X: 70202213-70202213 on Build GRCh38
rs773688896 Chr.X: 70202040-70202040 on Build GRCh38
rs768821779 Chr.X: 70202232-70202232 on Build GRCh38
rs1035271785 Chr.X: 70202276-70202276 on Build GRCh38
rs760451314 Chr.X: 70202076-70202076 on Build GRCh38
rs773920428 Chr.X: 70201885-70201885 on Build GRCh38
rs771530576 Chr.X: 70201845-70201845 on Build GRCh38
rs760504681 Chr.X: 70201906-70201906 on Build GRCh38
rs146765066 Chr.X: 70202083-70202083 on Build GRCh38
rs753659944 Chr.X: 70201924-70201924 on Build GRCh38
rs757750414 Chr.X: 70201970-70201970 on Build GRCh38
rs761419482 Chr.X: 70201892-70201892 on Build GRCh38
rs778176074 Chr.X: 70201955-70201955 on Build GRCh38
rs770368808 Chr.X: 70202030-70202030 on Build GRCh38
rs770181043 Chr.X: 70202156-70202156 on Build GRCh38
rs1033914340 Chr.X: 70202027-70202027 on Build GRCh38
rs765265420 Chr.X: 70202096-70202096 on Build GRCh38
rs779266204 Chr.X: 70201877-70201878 on Build GRCh38

View Full Product Details