Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65021724-65022230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2
Entrez Gene ID
402
Gene Name
ADP ribosylation factor like GTPase 2
Gene Aliases
ARFL2
UniGene
Hs.502836
Gene Chromosome Location
Chr.11: 65014113-65022185 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604809 c.478G>A p.V160I Chr.11: 65021778-65021778 on Build GRCh38
4972050 c.500C>T p.P167L Chr.11: 65021800-65021800 on Build GRCh38
4776288 c.498G>A p.L166L Chr.11: 65021798-65021798 on Build GRCh38
6134067 c.479T>C p.V160A Chr.11: 65021779-65021779 on Build GRCh38
2164895 c.483C>T p.T161T Chr.11: 65021783-65021783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920059567 Chr.11: 65021943-65021943 on Build GRCh38
rs777046100 Chr.11: 65021757-65021757 on Build GRCh38
rs761289343 Chr.11: 65022170-65022170 on Build GRCh38
rs567351380 Chr.11: 65022103-65022103 on Build GRCh38
rs969899509 Chr.11: 65021919-65021919 on Build GRCh38
rs934648767 Chr.11: 65021954-65021954 on Build GRCh38
rs532902659 Chr.11: 65021992-65021992 on Build GRCh38
rs370945888 Chr.11: 65021860-65021860 on Build GRCh38
rs752879055 Chr.11: 65021774-65021774 on Build GRCh38
rs536231079 Chr.11: 65022106-65022106 on Build GRCh38
rs186387718 Chr.11: 65022102-65022102 on Build GRCh38
rs182274655 Chr.11: 65022143-65022143 on Build GRCh38
rs948841563 Chr.11: 65022002-65022002 on Build GRCh38
rs765325857 Chr.11: 65021762-65021762 on Build GRCh38
rs947615055 Chr.11: 65021923-65021923 on Build GRCh38
rs546242758 Chr.11: 65021807-65021807 on Build GRCh38
rs11540368 Chr.11: 65021898-65021898 on Build GRCh38
rs771769117 Chr.11: 65021793-65021793 on Build GRCh38
rs752241565 Chr.11: 65021870-65021870 on Build GRCh38
rs962543637 Chr.11: 65022033-65022033 on Build GRCh38
rs759590910 Chr.11: 65021759-65021759 on Build GRCh38
rs1044892977 Chr.11: 65022006-65022006 on Build GRCh38
rs186939062 Chr.11: 65021778-65021778 on Build GRCh38
rs897776010 Chr.11: 65022058-65022058 on Build GRCh38
rs145706691 Chr.11: 65022146-65022146 on Build GRCh38
rs766906687 Chr.11: 65021756-65021756 on Build GRCh38
rs148156544 Chr.11: 65021835-65021835 on Build GRCh38
rs760492829 Chr.11: 65021857-65021857 on Build GRCh38
rs767163012 Chr.11: 65021777-65021777 on Build GRCh38
rs1030914739 Chr.11: 65022101-65022101 on Build GRCh38
rs931693788 Chr.11: 65022025-65022025 on Build GRCh38
rs1050199311 Chr.11: 65022026-65022026 on Build GRCh38
rs373752961 Chr.11: 65022171-65022171 on Build GRCh38
rs931501116 Chr.11: 65021955-65021955 on Build GRCh38
rs992602059 Chr.11: 65021894-65021894 on Build GRCh38
rs1015394725 Chr.11: 65022029-65022029 on Build GRCh38
rs760069227 Chr.11: 65021829-65021829 on Build GRCh38
rs903647631 Chr.11: 65022039-65022039 on Build GRCh38
rs200574613 Chr.11: 65021808-65021808 on Build GRCh38
rs924892727 Chr.11: 65021952-65021953 on Build GRCh38
rs777076110 Chr.11: 65021822-65021822 on Build GRCh38
rs772807691 Chr.11: 65021801-65021801 on Build GRCh38
rs771288868 Chr.11: 65021887-65021887 on Build GRCh38
rs372463350 Chr.11: 65021836-65021836 on Build GRCh38
rs920170451 Chr.11: 65021786-65021786 on Build GRCh38
rs769801595 Chr.11: 65021834-65021834 on Build GRCh38
rs956707938 Chr.11: 65021746-65021746 on Build GRCh38
rs546693176 Chr.11: 65021993-65021993 on Build GRCh38
rs550659216 Chr.11: 65022018-65022018 on Build GRCh38
rs373523646 Chr.11: 65021874-65021874 on Build GRCh38
rs1056957 Chr.11: 65022090-65022090 on Build GRCh38
rs61736632 Chr.11: 65021750-65021750 on Build GRCh38
rs1030977007 Chr.11: 65021745-65021745 on Build GRCh38
rs1000707187 Chr.11: 65022093-65022093 on Build GRCh38
rs764369522 Chr.11: 65021765-65021765 on Build GRCh38
rs141724442 Chr.11: 65021780-65021780 on Build GRCh38
rs1056955 Chr.11: 65022087-65022087 on Build GRCh38
rs75332988 Chr.11: 65021787-65021787 on Build GRCh38
rs974368053 Chr.11: 65022041-65022041 on Build GRCh38
rs553084517 Chr.11: 65022183-65022183 on Build GRCh38
rs774717693 Chr.11: 65021868-65021869 on Build GRCh38
rs181106139 Chr.11: 65021937-65021937 on Build GRCh38
rs369145929 Chr.11: 65021783-65021783 on Build GRCh38
rs1040485865 Chr.11: 65022013-65022013 on Build GRCh38
rs11825104 Chr.11: 65022109-65022109 on Build GRCh38
rs531231757 Chr.11: 65021921-65021921 on Build GRCh38
rs2271540 Chr.11: 65021968-65021968 on Build GRCh38
rs781545281 Chr.11: 65021891-65021891 on Build GRCh38
rs541806031 Chr.11: 65021883-65021883 on Build GRCh38
rs146832019 Chr.11: 65021743-65021743 on Build GRCh38
rs1057176161 Chr.11: 65021956-65021956 on Build GRCh38
rs754969281 Chr.11: 65021866-65021866 on Build GRCh38
rs566554615 Chr.11: 65022009-65022009 on Build GRCh38
rs535477031 Chr.11: 65022097-65022097 on Build GRCh38
rs530953384 Chr.11: 65021918-65021918 on Build GRCh38
rs778507212 Chr.11: 65021869-65021869 on Build GRCh38
rs758628032 Chr.11: 65021775-65021775 on Build GRCh38
rs1007326672 Chr.11: 65022121-65022121 on Build GRCh38
rs557682309 Chr.11: 65022129-65022129 on Build GRCh38
rs982958134 Chr.11: 65021963-65021963 on Build GRCh38
rs375730001 Chr.11: 65021855-65021855 on Build GRCh38

View Full Product Details