Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66858588-66859106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRFN4
Entrez Gene ID
78999
Gene Name
leucine rich repeat and fibronectin type III domain containing 4
Gene Aliases
FIGLER6, SALM3, SALM3.
UniGene
Hs.209979
Gene Chromosome Location
Chr.11: 66857087-66860475 on Build GRCh38
Transcripts
NM_024036.4 XM_005274239.3 NM_000920.3 NM_001040716.1 NM_022172.2 XM_005274031.4 XM_005274032.4 XM_006718578.3 XM_011545086.2 XM_017017868.1 XM_017017869.1 XM_017017870.1 XM_017017871.1 XM_017017872.1

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
UniGene
Hs.89890
Gene Chromosome Location
Chr.11: 66848522-66958418 on Build GRCh38
Transcripts
NM_024036.4 XM_005274239.3 NM_000920.3 NM_001040716.1 NM_022172.2 XM_005274031.4 XM_005274032.4 XM_006718578.3 XM_011545086.2 XM_017017868.1 XM_017017869.1 XM_017017870.1 XM_017017871.1 XM_017017872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6461993 c.1328A>C p.E443A Chr.11: 66859072-66859072 on Build GRCh38
6431819 c.936G>T p.M312I Chr.11: 66858680-66858680 on Build GRCh38
1239347 c.1156C>T p.P386S Chr.11: 66858900-66858900 on Build GRCh38
5446948 c.853C>T p.R285C Chr.11: 66858597-66858597 on Build GRCh38
6702885 c.1300A>G p.M434V Chr.11: 66859044-66859044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776530207 Chr.11: 66858729-66858729 on Build GRCh38
rs144041059 Chr.11: 66858959-66858959 on Build GRCh38
rs965801825 Chr.11: 66858624-66858624 on Build GRCh38
rs139704723 Chr.11: 66859074-66859074 on Build GRCh38
rs765840797 Chr.11: 66858994-66858994 on Build GRCh38
rs952871230 Chr.11: 66858731-66858731 on Build GRCh38
rs144355365 Chr.11: 66859086-66859086 on Build GRCh38
rs946568424 Chr.11: 66858845-66858845 on Build GRCh38
rs780815118 Chr.11: 66859013-66859014 on Build GRCh38
rs560082761 Chr.11: 66858698-66858698 on Build GRCh38
rs751754935 Chr.11: 66859024-66859024 on Build GRCh38
rs766244836 Chr.11: 66858640-66858640 on Build GRCh38
rs772966034 Chr.11: 66858989-66858989 on Build GRCh38
rs768571190 Chr.11: 66858979-66858979 on Build GRCh38
rs992499038 Chr.11: 66859083-66859083 on Build GRCh38
rs777782158 Chr.11: 66858857-66858857 on Build GRCh38
rs758678940 Chr.11: 66858952-66858952 on Build GRCh38
rs756654010 Chr.11: 66858862-66858862 on Build GRCh38
rs771914453 Chr.11: 66858988-66858988 on Build GRCh38
rs753688182 Chr.11: 66858645-66858645 on Build GRCh38
rs375297266 Chr.11: 66858987-66858987 on Build GRCh38
rs767165983 Chr.11: 66858843-66858843 on Build GRCh38
rs758408482 Chr.11: 66858737-66858737 on Build GRCh38
rs866747977 Chr.11: 66858718-66858718 on Build GRCh38
rs746648364 Chr.11: 66859069-66859069 on Build GRCh38
rs751550136 Chr.11: 66858611-66858611 on Build GRCh38
rs777341660 Chr.11: 66858677-66858677 on Build GRCh38
rs372294508 Chr.11: 66858791-66858791 on Build GRCh38
rs61731780 Chr.11: 66858814-66858814 on Build GRCh38
rs199739517 Chr.11: 66858767-66858767 on Build GRCh38
rs775261146 Chr.11: 66858905-66858905 on Build GRCh38
rs747078428 Chr.11: 66858897-66858897 on Build GRCh38
rs746446085 Chr.11: 66858679-66858679 on Build GRCh38
rs765027667 Chr.11: 66858800-66858800 on Build GRCh38
rs967763201 Chr.11: 66859055-66859055 on Build GRCh38
rs1056561916 Chr.11: 66858652-66858652 on Build GRCh38
rs767216993 Chr.11: 66858921-66858921 on Build GRCh38
rs372192216 Chr.11: 66858949-66858949 on Build GRCh38
rs979342953 Chr.11: 66858651-66858651 on Build GRCh38
rs367902341 Chr.11: 66858881-66858881 on Build GRCh38
rs776067349 Chr.11: 66859073-66859073 on Build GRCh38
rs779202810 Chr.11: 66858889-66858889 on Build GRCh38
rs868551638 Chr.11: 66858744-66858744 on Build GRCh38
rs201413613 Chr.11: 66858770-66858770 on Build GRCh38
rs759478441 Chr.11: 66858796-66858796 on Build GRCh38
rs749555267 Chr.11: 66858704-66858704 on Build GRCh38
rs775955953 Chr.11: 66859081-66859081 on Build GRCh38
rs546519818 Chr.11: 66858689-66858689 on Build GRCh38
rs753128001 Chr.11: 66859019-66859019 on Build GRCh38
rs375098656 Chr.11: 66858944-66858944 on Build GRCh38
rs757104711 Chr.11: 66858998-66858999 on Build GRCh38
rs770259072 Chr.11: 66858702-66858702 on Build GRCh38
rs1018574768 Chr.11: 66858780-66858780 on Build GRCh38
rs781763774 Chr.11: 66858690-66858690 on Build GRCh38
rs201895033 Chr.11: 66858904-66858904 on Build GRCh38
rs746869072 Chr.11: 66858979-66858979 on Build GRCh38
rs200734989 Chr.11: 66858935-66858935 on Build GRCh38
rs752372693 Chr.11: 66858742-66858742 on Build GRCh38
rs769166809 Chr.11: 66858980-66858980 on Build GRCh38
rs895890272 Chr.11: 66858723-66858723 on Build GRCh38
rs149152305 Chr.11: 66858911-66858911 on Build GRCh38
rs751232106 Chr.11: 66858832-66858832 on Build GRCh38
rs374346244 Chr.11: 66858835-66858835 on Build GRCh38
rs768356861 Chr.11: 66858914-66858914 on Build GRCh38
rs747993632 Chr.11: 66859049-66859049 on Build GRCh38
rs773239672 Chr.11: 66858794-66858794 on Build GRCh38
rs902402677 Chr.11: 66858646-66858646 on Build GRCh38
rs755265013 Chr.11: 66858882-66858882 on Build GRCh38
rs775414771 Chr.11: 66858810-66858810 on Build GRCh38
rs763849195 Chr.11: 66858823-66858823 on Build GRCh38
rs776142846 Chr.11: 66859007-66859007 on Build GRCh38
rs765116208 Chr.11: 66858730-66858730 on Build GRCh38
rs778985492 Chr.11: 66859046-66859046 on Build GRCh38
rs765774004 Chr.11: 66859079-66859079 on Build GRCh38
rs754740182 Chr.11: 66858660-66858660 on Build GRCh38
rs781135899 Chr.11: 66859031-66859031 on Build GRCh38
rs768977845 Chr.11: 66858779-66858779 on Build GRCh38
rs760606629 Chr.11: 66858618-66858618 on Build GRCh38
rs61731779 Chr.11: 66858879-66858879 on Build GRCh38
rs749213323 Chr.11: 66858969-66858969 on Build GRCh38
rs376308544 Chr.11: 66858771-66858771 on Build GRCh38
rs766367685 Chr.11: 66858722-66858722 on Build GRCh38
rs770895637 Chr.11: 66858960-66858960 on Build GRCh38
rs368557087 Chr.11: 66858786-66858786 on Build GRCh38
rs374715016 Chr.11: 66858607-66858607 on Build GRCh38
rs768690295 Chr.11: 66858710-66858710 on Build GRCh38
rs758084352 Chr.11: 66858743-66858743 on Build GRCh38
rs188956583 Chr.11: 66859032-66859032 on Build GRCh38
rs199906653 Chr.11: 66858950-66858950 on Build GRCh38
rs760570067 Chr.11: 66858924-66858924 on Build GRCh38
rs146377143 Chr.11: 66858990-66858990 on Build GRCh38
rs61742217 Chr.11: 66858852-66858852 on Build GRCh38
rs568932527 Chr.11: 66858953-66858953 on Build GRCh38
rs762653204 Chr.11: 66858821-66858821 on Build GRCh38
rs746399305 Chr.11: 66858768-66858768 on Build GRCh38
rs765724677 Chr.11: 66859018-66859018 on Build GRCh38
rs774498707 Chr.11: 66858715-66858715 on Build GRCh38
rs753798727 Chr.11: 66859070-66859070 on Build GRCh38
rs1004972521 Chr.11: 66858775-66858775 on Build GRCh38
rs550761110 Chr.11: 66858896-66858896 on Build GRCh38
rs531056189 Chr.11: 66858793-66858793 on Build GRCh38
rs3741194 Chr.11: 66858763-66858763 on Build GRCh38
rs1044990835 Chr.11: 66858740-66858740 on Build GRCh38
rs771967835 Chr.11: 66859058-66859058 on Build GRCh38
rs757458564 Chr.11: 66859025-66859025 on Build GRCh38
rs930826260 Chr.11: 66858945-66858945 on Build GRCh38
rs748403576 Chr.11: 66858892-66858892 on Build GRCh38
rs1005700699 Chr.11: 66858732-66858732 on Build GRCh38
rs764849455 Chr.11: 66858668-66858668 on Build GRCh38
rs367785957 Chr.11: 66858671-66858671 on Build GRCh38
rs374693774 Chr.11: 66858920-66858920 on Build GRCh38
rs759978534 Chr.11: 66859010-66859010 on Build GRCh38
rs764349967 Chr.11: 66859023-66859023 on Build GRCh38
rs566421079 Chr.11: 66858927-66858927 on Build GRCh38
rs763641039 Chr.11: 66858758-66858758 on Build GRCh38
rs774246521 Chr.11: 66858915-66858915 on Build GRCh38
rs369067794 Chr.11: 66858925-66858925 on Build GRCh38
rs894803860 Chr.11: 66858669-66858669 on Build GRCh38
rs775668753 Chr.11: 66858703-66858703 on Build GRCh38
rs532858753 Chr.11: 66858963-66858963 on Build GRCh38
rs749886260 Chr.11: 66858853-66858853 on Build GRCh38
rs752420546 Chr.11: 66858670-66858670 on Build GRCh38
rs974985014 Chr.11: 66858626-66858626 on Build GRCh38
rs758732336 Chr.11: 66859020-66859020 on Build GRCh38

View Full Product Details