Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43363823-43364324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN29
Entrez Gene ID
146050
Gene Name
zinc finger and SCAN domain containing 29
Gene Aliases
ZNF690, Zfp690
UniGene
Hs.418287
Gene Chromosome Location
Chr.15: 43358172-43371032 on Build GRCh38
Transcripts
NM_152455.3 XM_006720401.3 XM_011521266.2 XM_017021949.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5566414 c.1688G>A p.R563H Chr.15: 43363917-43363917 on Build GRCh38
4929045 c.1447A>C p.T483P Chr.15: 43364158-43364158 on Build GRCh38
1493437 c.1339G>A p.E447K Chr.15: 43364266-43364266 on Build GRCh38
188138 c.1643delC p.P548fs*5 Chr.15: 43363962-43363962 on Build GRCh38
7288851 c.1687C>T p.R563C Chr.15: 43363918-43363918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753745948 Chr.15: 43364275-43364275 on Build GRCh38
rs770540121 Chr.15: 43363932-43363932 on Build GRCh38
rs753603052 Chr.15: 43364113-43364113 on Build GRCh38
rs532162092 Chr.15: 43364011-43364011 on Build GRCh38
rs151260867 Chr.15: 43363964-43363964 on Build GRCh38
rs866244005 Chr.15: 43363878-43363878 on Build GRCh38
rs780062361 Chr.15: 43364278-43364279 on Build GRCh38
rs761074357 Chr.15: 43363966-43363966 on Build GRCh38
rs761631253 Chr.15: 43364276-43364276 on Build GRCh38
rs1056689022 Chr.15: 43364138-43364138 on Build GRCh38
rs748495124 Chr.15: 43364180-43364180 on Build GRCh38
rs2899082 Chr.15: 43363845-43363845 on Build GRCh38
rs79105877 Chr.15: 43364242-43364242 on Build GRCh38
rs148746894 Chr.15: 43363920-43363920 on Build GRCh38
rs756044509 Chr.15: 43364015-43364015 on Build GRCh38
rs540103257 Chr.15: 43363973-43363973 on Build GRCh38
rs757457895 Chr.15: 43363871-43363871 on Build GRCh38
rs750297466 Chr.15: 43364295-43364295 on Build GRCh38
rs201083890 Chr.15: 43363965-43363965 on Build GRCh38
rs750234483 Chr.15: 43363980-43363982 on Build GRCh38
rs981338831 Chr.15: 43364231-43364231 on Build GRCh38
rs765439900 Chr.15: 43363994-43363994 on Build GRCh38
rs201902651 Chr.15: 43364081-43364081 on Build GRCh38
rs576666978 Chr.15: 43364247-43364247 on Build GRCh38
rs765335050 Chr.15: 43363873-43363873 on Build GRCh38
rs769999057 Chr.15: 43364079-43364079 on Build GRCh38
rs766414519 Chr.15: 43364092-43364092 on Build GRCh38
rs147749773 Chr.15: 43364097-43364097 on Build GRCh38
rs746564574 Chr.15: 43364165-43364165 on Build GRCh38
rs749095806 Chr.15: 43364023-43364023 on Build GRCh38
rs767392497 Chr.15: 43364010-43364010 on Build GRCh38
rs756930650 Chr.15: 43364034-43364034 on Build GRCh38
rs139243405 Chr.15: 43364019-43364019 on Build GRCh38
rs142610145 Chr.15: 43364221-43364221 on Build GRCh38
rs202186966 Chr.15: 43364236-43364236 on Build GRCh38
rs747492950 Chr.15: 43364056-43364056 on Build GRCh38
rs781560238 Chr.15: 43364174-43364174 on Build GRCh38
rs939213299 Chr.15: 43364134-43364134 on Build GRCh38
rs761663803 Chr.15: 43363983-43363983 on Build GRCh38
rs764305921 Chr.15: 43364283-43364283 on Build GRCh38
rs1025700275 Chr.15: 43364220-43364220 on Build GRCh38
rs371535992 Chr.15: 43364014-43364014 on Build GRCh38
rs964630299 Chr.15: 43364270-43364270 on Build GRCh38
rs769098912 Chr.15: 43364059-43364059 on Build GRCh38
rs758228781 Chr.15: 43364232-43364232 on Build GRCh38
rs112956834 Chr.15: 43363925-43363925 on Build GRCh38
rs770086195 Chr.15: 43364196-43364196 on Build GRCh38
rs143986040 Chr.15: 43363918-43363918 on Build GRCh38
rs905095067 Chr.15: 43364132-43364132 on Build GRCh38
rs745319191 Chr.15: 43364040-43364040 on Build GRCh38
rs778438562 Chr.15: 43364037-43364037 on Build GRCh38
rs771201858 Chr.15: 43364227-43364227 on Build GRCh38
rs3917222 Chr.15: 43364088-43364088 on Build GRCh38
rs772644607 Chr.15: 43363960-43363960 on Build GRCh38
rs750133880 Chr.15: 43364125-43364125 on Build GRCh38
rs201009870 Chr.15: 43364104-43364104 on Build GRCh38
rs1020399956 Chr.15: 43363868-43363868 on Build GRCh38
rs560406082 Chr.15: 43363982-43363982 on Build GRCh38
rs773604219 Chr.15: 43364219-43364219 on Build GRCh38
rs781110783 Chr.15: 43363909-43363909 on Build GRCh38
rs370272155 Chr.15: 43363968-43363968 on Build GRCh38
rs778664376 Chr.15: 43364116-43364116 on Build GRCh38
rs750431845 Chr.15: 43363998-43363998 on Build GRCh38
rs146772052 Chr.15: 43364262-43364262 on Build GRCh38
rs1055723180 Chr.15: 43364282-43364282 on Build GRCh38
rs532183967 Chr.15: 43363886-43363886 on Build GRCh38
rs762085365 Chr.15: 43364066-43364066 on Build GRCh38
rs779694314 Chr.15: 43364156-43364156 on Build GRCh38
rs750555852 Chr.15: 43363893-43363893 on Build GRCh38
rs758167367 Chr.15: 43364131-43364131 on Build GRCh38
rs773899606 Chr.15: 43363944-43363944 on Build GRCh38
rs758485935 Chr.15: 43364002-43364002 on Build GRCh38
rs780669763 Chr.15: 43364053-43364053 on Build GRCh38
rs754901870 Chr.15: 43363901-43363901 on Build GRCh38
rs764882023 Chr.15: 43363948-43363948 on Build GRCh38
rs997717739 Chr.15: 43363897-43363897 on Build GRCh38
rs1035798249 Chr.15: 43363924-43363924 on Build GRCh38
rs769137066 Chr.15: 43364167-43364167 on Build GRCh38
rs757019551 Chr.15: 43364115-43364115 on Build GRCh38
rs762977866 Chr.15: 43364083-43364083 on Build GRCh38
rs369750925 Chr.15: 43364188-43364188 on Build GRCh38
rs750350792 Chr.15: 43364047-43364047 on Build GRCh38
rs984379697 Chr.15: 43363992-43363992 on Build GRCh38
rs946353904 Chr.15: 43364121-43364121 on Build GRCh38
rs758116202 Chr.15: 43364297-43364297 on Build GRCh38
rs935058745 Chr.15: 43364069-43364069 on Build GRCh38
rs145699954 Chr.15: 43364065-43364065 on Build GRCh38
rs747879860 Chr.15: 43363910-43363910 on Build GRCh38
rs377268526 Chr.15: 43364095-43364095 on Build GRCh38
rs755923103 Chr.15: 43363917-43363917 on Build GRCh38
rs745438331 Chr.15: 43363946-43363946 on Build GRCh38

View Full Product Details