Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41464191-41464698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT32
Entrez Gene ID
3882
Gene Name
keratin 32
Gene Aliases
HA2, HKA2, KRTHA2, hHa2
UniGene
Hs.41752
Gene Chromosome Location
Chr.17: 41459513-41469252 on Build GRCh38
Transcripts
NM_002278.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5386338 c.719C>T p.S240F Chr.17: 41464433-41464433 on Build GRCh38
3517385 c.843G>A p.R281R Chr.17: 41464309-41464309 on Build GRCh38
3517386 c.772C>T p.P258S Chr.17: 41464380-41464380 on Build GRCh38
979181 c.779A>G p.D260G Chr.17: 41464373-41464373 on Build GRCh38
5202382 c.723delT p.R242fs*20 Chr.17: 41464429-41464429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146792525 Chr.17: 41464389-41464389 on Build GRCh38
rs769005536 Chr.17: 41464250-41464250 on Build GRCh38
rs899061409 Chr.17: 41464273-41464273 on Build GRCh38
rs117761077 Chr.17: 41464665-41464665 on Build GRCh38
rs758977562 Chr.17: 41464291-41464291 on Build GRCh38
rs966983925 Chr.17: 41464514-41464514 on Build GRCh38
rs202139624 Chr.17: 41464399-41464399 on Build GRCh38
rs140575243 Chr.17: 41464453-41464453 on Build GRCh38
rs562951037 Chr.17: 41464604-41464604 on Build GRCh38
rs761238628 Chr.17: 41464282-41464282 on Build GRCh38
rs764395615 Chr.17: 41464489-41464489 on Build GRCh38
rs111388261 Chr.17: 41464336-41464336 on Build GRCh38
rs376557140 Chr.17: 41464378-41464378 on Build GRCh38
rs374113287 Chr.17: 41464218-41464220 on Build GRCh38
rs765403077 Chr.17: 41464398-41464398 on Build GRCh38
rs778185516 Chr.17: 41464346-41464346 on Build GRCh38
rs1022434783 Chr.17: 41464507-41464507 on Build GRCh38
rs555643784 Chr.17: 41464538-41464538 on Build GRCh38
rs72830046 Chr.17: 41464313-41464313 on Build GRCh38
rs921031229 Chr.17: 41464553-41464553 on Build GRCh38
rs945460908 Chr.17: 41464223-41464223 on Build GRCh38
rs764897371 Chr.17: 41464296-41464296 on Build GRCh38
rs190006017 Chr.17: 41464379-41464379 on Build GRCh38
rs1011169657 Chr.17: 41464541-41464541 on Build GRCh38
rs757317631 Chr.17: 41464252-41464252 on Build GRCh38
rs774203140 Chr.17: 41464384-41464384 on Build GRCh38
rs73983451 Chr.17: 41464314-41464314 on Build GRCh38
rs369299043 Chr.17: 41464268-41464268 on Build GRCh38
rs892753456 Chr.17: 41464518-41464518 on Build GRCh38
rs1011079637 Chr.17: 41464494-41464494 on Build GRCh38
rs573683336 Chr.17: 41464532-41464532 on Build GRCh38
rs779820459 Chr.17: 41464466-41464467 on Build GRCh38
rs182948314 Chr.17: 41464621-41464621 on Build GRCh38
rs201967748 Chr.17: 41464408-41464408 on Build GRCh38
rs1001662815 Chr.17: 41464496-41464496 on Build GRCh38
rs751511511 Chr.17: 41464312-41464312 on Build GRCh38
rs752490341 Chr.17: 41464214-41464214 on Build GRCh38
rs909712552 Chr.17: 41464625-41464625 on Build GRCh38
rs749270808 Chr.17: 41464249-41464249 on Build GRCh38
rs748545659 Chr.17: 41464265-41464265 on Build GRCh38
rs758144413 Chr.17: 41464219-41464219 on Build GRCh38
rs375825183 Chr.17: 41464259-41464259 on Build GRCh38
rs375993058 Chr.17: 41464325-41464325 on Build GRCh38
rs761717124 Chr.17: 41464436-41464436 on Build GRCh38
rs747275881 Chr.17: 41464347-41464347 on Build GRCh38
rs1032697503 Chr.17: 41464515-41464515 on Build GRCh38
rs773349020 Chr.17: 41464438-41464438 on Build GRCh38
rs574301036 Chr.17: 41464544-41464544 on Build GRCh38
rs764110812 Chr.17: 41464409-41464409 on Build GRCh38
rs1021207928 Chr.17: 41464574-41464574 on Build GRCh38
rs372388716 Chr.17: 41464284-41464284 on Build GRCh38
rs773827921 Chr.17: 41464432-41464432 on Build GRCh38
rs905594131 Chr.17: 41464482-41464482 on Build GRCh38
rs748732566 Chr.17: 41464423-41464423 on Build GRCh38
rs368638057 Chr.17: 41464437-41464437 on Build GRCh38
rs187233165 Chr.17: 41464626-41464626 on Build GRCh38
rs34593860 Chr.17: 41464559-41464559 on Build GRCh38
rs116221624 Chr.17: 41464305-41464305 on Build GRCh38
rs769355585 Chr.17: 41464416-41464416 on Build GRCh38
rs147094229 Chr.17: 41464380-41464380 on Build GRCh38
rs141728288 Chr.17: 41464335-41464335 on Build GRCh38
rs145720725 Chr.17: 41464428-41464428 on Build GRCh38
rs543082865 Chr.17: 41464521-41464521 on Build GRCh38
rs745401786 Chr.17: 41464363-41464363 on Build GRCh38
rs770539388 Chr.17: 41464261-41464261 on Build GRCh38
rs2604959 Chr.17: 41464658-41464658 on Build GRCh38
rs534514452 Chr.17: 41464536-41464536 on Build GRCh38
rs528431706 Chr.17: 41464321-41464321 on Build GRCh38
rs202198782 Chr.17: 41464220-41464220 on Build GRCh38
rs758248194 Chr.17: 41464466-41464466 on Build GRCh38
rs886281934 Chr.17: 41464460-41464460 on Build GRCh38
rs568254576 Chr.17: 41464351-41464351 on Build GRCh38
rs544689749 Chr.17: 41464592-41464592 on Build GRCh38
rs773631568 Chr.17: 41464269-41464269 on Build GRCh38
rs144180024 Chr.17: 41464299-41464299 on Build GRCh38
rs144107966 Chr.17: 41464410-41464410 on Build GRCh38
rs905561480 Chr.17: 41464330-41464330 on Build GRCh38
rs79177667 Chr.17: 41464246-41464246 on Build GRCh38
rs776314677 Chr.17: 41464285-41464285 on Build GRCh38
rs777787635 Chr.17: 41464401-41464401 on Build GRCh38
rs2604958 Chr.17: 41464481-41464481 on Build GRCh38
rs368488025 Chr.17: 41464493-41464493 on Build GRCh38
rs149150620 Chr.17: 41464306-41464306 on Build GRCh38
rs201036046 Chr.17: 41464492-41464492 on Build GRCh38
rs751486516 Chr.17: 41464233-41464233 on Build GRCh38
rs767834759 Chr.17: 41464673-41464673 on Build GRCh38
rs755148997 Chr.17: 41464393-41464393 on Build GRCh38
rs374696277 Chr.17: 41464375-41464375 on Build GRCh38
rs779045478 Chr.17: 41464262-41464263 on Build GRCh38
rs949880395 Chr.17: 41464475-41464475 on Build GRCh38
rs751705894 Chr.17: 41464491-41464491 on Build GRCh38
rs776960040 Chr.17: 41464359-41464359 on Build GRCh38
rs774996977 Chr.17: 41464418-41464418 on Build GRCh38
rs750623362 Chr.17: 41464412-41464413 on Build GRCh38
rs756218572 Chr.17: 41464234-41464234 on Build GRCh38
rs760676982 Chr.17: 41464439-41464439 on Build GRCh38

View Full Product Details