Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41465543-41466061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT32
Entrez Gene ID
3882
Gene Name
keratin 32
Gene Aliases
HA2, HKA2, KRTHA2, hHa2
UniGene
Hs.41752
Gene Chromosome Location
Chr.17: 41459513-41469252 on Build GRCh38
Transcripts
NM_002278.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1166017 c.636C>A p.C212* Chr.17: 41465845-41465845 on Build GRCh38
6386463 c.670A>G p.K224E Chr.17: 41465811-41465811 on Build GRCh38
4502558 c.616C>G p.L206V Chr.17: 41465865-41465865 on Build GRCh38
3517387 c.591C>G p.A197A Chr.17: 41465890-41465890 on Build GRCh38
4924626 c.595A>G p.I199V Chr.17: 41465886-41465886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544111243 Chr.17: 41465720-41465720 on Build GRCh38
rs538728586 Chr.17: 41465643-41465643 on Build GRCh38
rs777612920 Chr.17: 41465907-41465907 on Build GRCh38
rs764046225 Chr.17: 41465762-41465762 on Build GRCh38
rs371356129 Chr.17: 41465730-41465730 on Build GRCh38
rs780866147 Chr.17: 41465916-41465916 on Build GRCh38
rs779378843 Chr.17: 41465697-41465697 on Build GRCh38
rs527436785 Chr.17: 41465873-41465873 on Build GRCh38
rs990623065 Chr.17: 41465608-41465608 on Build GRCh38
rs868088959 Chr.17: 41465802-41465802 on Build GRCh38
rs147177232 Chr.17: 41465715-41465715 on Build GRCh38
rs780499585 Chr.17: 41465731-41465731 on Build GRCh38
rs778272970 Chr.17: 41465832-41465832 on Build GRCh38
rs760377550 Chr.17: 41466000-41466000 on Build GRCh38
rs751869334 Chr.17: 41465779-41465779 on Build GRCh38
rs1049268986 Chr.17: 41465671-41465671 on Build GRCh38
rs1037006570 Chr.17: 41465869-41465869 on Build GRCh38
rs554215078 Chr.17: 41465651-41465651 on Build GRCh38
rs923370739 Chr.17: 41465786-41465786 on Build GRCh38
rs776745683 Chr.17: 41465860-41465860 on Build GRCh38
rs759904226 Chr.17: 41465958-41465958 on Build GRCh38
rs983738609 Chr.17: 41466037-41466037 on Build GRCh38
rs375223295 Chr.17: 41465784-41465784 on Build GRCh38
rs745783879 Chr.17: 41465861-41465861 on Build GRCh38
rs748226139 Chr.17: 41465925-41465925 on Build GRCh38
rs544903437 Chr.17: 41465800-41465800 on Build GRCh38
rs200655946 Chr.17: 41465922-41465922 on Build GRCh38
rs943742605 Chr.17: 41465592-41465592 on Build GRCh38
rs115611881 Chr.17: 41465742-41465742 on Build GRCh38
rs761068532 Chr.17: 41465888-41465888 on Build GRCh38
rs373985240 Chr.17: 41465874-41465874 on Build GRCh38
rs979435975 Chr.17: 41465668-41465668 on Build GRCh38
rs2071561 Chr.17: 41465816-41465816 on Build GRCh38
rs199750321 Chr.17: 41465756-41465756 on Build GRCh38
rs566160324 Chr.17: 41466038-41466038 on Build GRCh38
rs745612757 Chr.17: 41465921-41465921 on Build GRCh38
rs1002930043 Chr.17: 41465698-41465698 on Build GRCh38
rs539204747 Chr.17: 41466017-41466017 on Build GRCh38
rs896793988 Chr.17: 41465725-41465725 on Build GRCh38
rs73983464 Chr.17: 41465620-41465620 on Build GRCh38
rs117928429 Chr.17: 41465794-41465794 on Build GRCh38
rs942276527 Chr.17: 41465711-41465711 on Build GRCh38
rs571998013 Chr.17: 41465987-41465987 on Build GRCh38
rs774430230 Chr.17: 41465913-41465913 on Build GRCh38
rs547712755 Chr.17: 41466033-41466033 on Build GRCh38
rs940889132 Chr.17: 41465808-41465808 on Build GRCh38
rs144482548 Chr.17: 41465890-41465890 on Build GRCh38
rs760478878 Chr.17: 41465746-41465746 on Build GRCh38
rs769718581 Chr.17: 41465863-41465863 on Build GRCh38
rs369663094 Chr.17: 41465650-41465650 on Build GRCh38
rs772109125 Chr.17: 41465926-41465926 on Build GRCh38
rs757182041 Chr.17: 41465909-41465909 on Build GRCh38
rs766529604 Chr.17: 41465948-41465948 on Build GRCh38
rs1036499092 Chr.17: 41465814-41465814 on Build GRCh38
rs932244841 Chr.17: 41465655-41465655 on Build GRCh38
rs772149700 Chr.17: 41465959-41465959 on Build GRCh38
rs570984984 Chr.17: 41465721-41465721 on Build GRCh38
rs909491659 Chr.17: 41465916-41465931 on Build GRCh38
rs1031341935 Chr.17: 41465937-41465937 on Build GRCh38
rs746743648 Chr.17: 41465886-41465887 on Build GRCh38
rs756423581 Chr.17: 41465796-41465796 on Build GRCh38
rs746820307 Chr.17: 41465840-41465840 on Build GRCh38
rs1056094042 Chr.17: 41465887-41465887 on Build GRCh38
rs758019971 Chr.17: 41465977-41465977 on Build GRCh38
rs774301975 Chr.17: 41465866-41465866 on Build GRCh38
rs754584511 Chr.17: 41465817-41465817 on Build GRCh38
rs888035367 Chr.17: 41465704-41465704 on Build GRCh38
rs369806846 Chr.17: 41465934-41465934 on Build GRCh38
rs747636371 Chr.17: 41465836-41465836 on Build GRCh38
rs556752923 Chr.17: 41465591-41465591 on Build GRCh38
rs137911171 Chr.17: 41465692-41465692 on Build GRCh38
rs143521620 Chr.17: 41465702-41465702 on Build GRCh38
rs775453547 Chr.17: 41465924-41465924 on Build GRCh38
rs752178888 Chr.17: 41465905-41465905 on Build GRCh38
rs760841408 Chr.17: 41466018-41466018 on Build GRCh38
rs767662150 Chr.17: 41465790-41465790 on Build GRCh38
rs993958824 Chr.17: 41465729-41465729 on Build GRCh38
rs766726111 Chr.17: 41465889-41465889 on Build GRCh38
rs942308896 Chr.17: 41465825-41465825 on Build GRCh38
rs776658443 Chr.17: 41465755-41465755 on Build GRCh38
rs773152181 Chr.17: 41465745-41465745 on Build GRCh38
rs369737962 Chr.17: 41465822-41465822 on Build GRCh38
rs192768473 Chr.17: 41465683-41465683 on Build GRCh38
rs576831254 Chr.17: 41465741-41465741 on Build GRCh38
rs760874576 Chr.17: 41465938-41465938 on Build GRCh38
rs770823083 Chr.17: 41465856-41465856 on Build GRCh38
rs766647046 Chr.17: 41465639-41465639 on Build GRCh38
rs188735491 Chr.17: 41465898-41465898 on Build GRCh38
rs1030300951 Chr.17: 41465988-41465988 on Build GRCh38
rs771021253 Chr.17: 41465749-41465749 on Build GRCh38
rs373349636 Chr.17: 41465951-41465951 on Build GRCh38
rs753221708 Chr.17: 41465965-41465965 on Build GRCh38
rs967918512 Chr.17: 41465649-41465649 on Build GRCh38
rs771625588 Chr.17: 41465838-41465838 on Build GRCh38
rs200664830 Chr.17: 41465906-41465906 on Build GRCh38
rs965515263 Chr.17: 41465744-41465744 on Build GRCh38
rs547775362 Chr.17: 41465673-41465673 on Build GRCh38
rs111628156 Chr.17: 41465707-41465707 on Build GRCh38
rs181116160 Chr.17: 41465964-41465964 on Build GRCh38

View Full Product Details