Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132378801-132379319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTF1
Entrez Gene ID
7270
Gene Name
transcription termination factor 1
Gene Aliases
TTF-1, TTF-I
UniGene
Hs.54780
Gene Chromosome Location
Chr.9: 132375548-132406851 on Build GRCh38
Transcripts
NM_001205296.1 NM_007344.3 NR_134525.1 XM_006717273.3 XM_017015105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297918 c.2412T>C p.F804F Chr.9: 132379111-132379111 on Build GRCh38
3655110 c.2461C>T p.P821S Chr.9: 132379062-132379062 on Build GRCh38
6571177 c.2421G>A p.L807L Chr.9: 132379102-132379102 on Build GRCh38
4737265 c.2397C>T p.Y799Y Chr.9: 132379126-132379126 on Build GRCh38
5904345 c.2437C>T p.P813S Chr.9: 132379086-132379086 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766759691 Chr.9: 132379181-132379181 on Build GRCh38
rs894841960 Chr.9: 132378864-132378864 on Build GRCh38
rs777121873 Chr.9: 132379039-132379039 on Build GRCh38
rs748033634 Chr.9: 132379034-132379034 on Build GRCh38
rs1026267808 Chr.9: 132378955-132378955 on Build GRCh38
rs754709623 Chr.9: 132379022-132379022 on Build GRCh38
rs549590378 Chr.9: 132379068-132379068 on Build GRCh38
rs946593507 Chr.9: 132378834-132378834 on Build GRCh38
rs545617880 Chr.9: 132378877-132378877 on Build GRCh38
rs1016657770 Chr.9: 132378886-132378886 on Build GRCh38
rs753615668 Chr.9: 132379019-132379019 on Build GRCh38
rs1050946717 Chr.9: 132378956-132378956 on Build GRCh38
rs958761276 Chr.9: 132379207-132379207 on Build GRCh38
rs750911734 Chr.9: 132379081-132379081 on Build GRCh38
rs749730122 Chr.9: 132379189-132379189 on Build GRCh38
rs939173082 Chr.9: 132379156-132379156 on Build GRCh38
rs780536688 Chr.9: 132379187-132379187 on Build GRCh38
rs890811880 Chr.9: 132378967-132378967 on Build GRCh38
rs115471853 Chr.9: 132379002-132379002 on Build GRCh38
rs200153166 Chr.9: 132379126-132379126 on Build GRCh38
rs962736102 Chr.9: 132378908-132378908 on Build GRCh38
rs895043204 Chr.9: 132379234-132379234 on Build GRCh38
rs902366565 Chr.9: 132379147-132379147 on Build GRCh38
rs914666424 Chr.9: 132379077-132379077 on Build GRCh38
rs760913419 Chr.9: 132379135-132379135 on Build GRCh38
rs772099166 Chr.9: 132379195-132379195 on Build GRCh38
rs751468421 Chr.9: 132379150-132379150 on Build GRCh38
rs942349188 Chr.9: 132378986-132378986 on Build GRCh38
rs1003453953 Chr.9: 132378829-132378829 on Build GRCh38
rs995706964 Chr.9: 132379059-132379059 on Build GRCh38
rs778565156 Chr.9: 132379186-132379187 on Build GRCh38
rs778683454 Chr.9: 132379025-132379025 on Build GRCh38
rs770244496 Chr.9: 132379080-132379080 on Build GRCh38
rs753268627 Chr.9: 132379126-132379127 on Build GRCh38
rs774287892 Chr.9: 132379120-132379120 on Build GRCh38
rs533635957 Chr.9: 132378873-132378873 on Build GRCh38
rs762230562 Chr.9: 132379192-132379192 on Build GRCh38
rs955565321 Chr.9: 132378983-132378983 on Build GRCh38
rs756723081 Chr.9: 132379160-132379160 on Build GRCh38
rs182214241 Chr.9: 132378906-132378906 on Build GRCh38
rs746567394 Chr.9: 132379063-132379063 on Build GRCh38
rs758374673 Chr.9: 132379036-132379036 on Build GRCh38
rs776367998 Chr.9: 132379074-132379074 on Build GRCh38
rs762009394 Chr.9: 132379122-132379122 on Build GRCh38
rs546962163 Chr.9: 132379176-132379176 on Build GRCh38
rs570788115 Chr.9: 132378910-132378913 on Build GRCh38
rs969215628 Chr.9: 132379260-132379260 on Build GRCh38
rs753696937 Chr.9: 132379182-132379183 on Build GRCh38
rs759122026 Chr.9: 132378941-132378941 on Build GRCh38
rs148200795 Chr.9: 132379139-132379139 on Build GRCh38
rs12347290 Chr.9: 132379190-132379190 on Build GRCh38
rs898238507 Chr.9: 132379046-132379046 on Build GRCh38
rs774862489 Chr.9: 132379191-132379191 on Build GRCh38
rs754842011 Chr.9: 132379185-132379185 on Build GRCh38
rs188447352 Chr.9: 132379229-132379229 on Build GRCh38
rs758235058 Chr.9: 132379140-132379140 on Build GRCh38
rs371815734 Chr.9: 132379137-132379137 on Build GRCh38
rs1034880402 Chr.9: 132378840-132378840 on Build GRCh38
rs763532821 Chr.9: 132379083-132379083 on Build GRCh38
rs780884275 Chr.9: 132379173-132379173 on Build GRCh38
rs1013314938 Chr.9: 132379243-132379243 on Build GRCh38
rs185996276 Chr.9: 132379125-132379125 on Build GRCh38
rs557571516 Chr.9: 132379254-132379254 on Build GRCh38
rs769247792 Chr.9: 132379109-132379109 on Build GRCh38
rs777826895 Chr.9: 132379141-132379141 on Build GRCh38
rs769621052 Chr.9: 132379185-132379185 on Build GRCh38

View Full Product Details