Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132386470-132386937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTF1
Entrez Gene ID
7270
Gene Name
transcription termination factor 1
Gene Aliases
TTF-1, TTF-I
UniGene
Hs.54780
Gene Chromosome Location
Chr.9: 132375548-132406851 on Build GRCh38
Transcripts
NM_001205296.1 NM_007344.3 NR_134525.1 XM_006717273.3 XM_017015105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5351578 c.2335G>A p.D779N Chr.9: 132386599-132386599 on Build GRCh38
7264399 c.2355G>T p.W785C Chr.9: 132386579-132386579 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547274842 Chr.9: 132386711-132386711 on Build GRCh38
rs1046843671 Chr.9: 132386713-132386713 on Build GRCh38
rs762350570 Chr.9: 132386829-132386829 on Build GRCh38
rs576105919 Chr.9: 132386893-132386893 on Build GRCh38
rs573595956 Chr.9: 132386886-132386886 on Build GRCh38
rs45610937 Chr.9: 132386626-132386626 on Build GRCh38
rs117329298 Chr.9: 132386870-132386870 on Build GRCh38
rs944167187 Chr.9: 132386718-132386718 on Build GRCh38
rs555844755 Chr.9: 132386818-132386818 on Build GRCh38
rs761023120 Chr.9: 132386743-132386743 on Build GRCh38
rs373530334 Chr.9: 132386616-132386616 on Build GRCh38
rs779592905 Chr.9: 132386548-132386548 on Build GRCh38
rs568229393 Chr.9: 132386533-132386533 on Build GRCh38
rs187972773 Chr.9: 132386830-132386830 on Build GRCh38
rs963695751 Chr.9: 132386606-132386606 on Build GRCh38
rs184476114 Chr.9: 132386889-132386889 on Build GRCh38
rs953029042 Chr.9: 132386649-132386649 on Build GRCh38
rs369241191 Chr.9: 132386538-132386538 on Build GRCh38
rs762730803 Chr.9: 132386816-132386816 on Build GRCh38
rs772613041 Chr.9: 132386643-132386643 on Build GRCh38
rs371098655 Chr.9: 132386666-132386666 on Build GRCh38
rs1027035015 Chr.9: 132386863-132386864 on Build GRCh38
rs375403741 Chr.9: 132386518-132386518 on Build GRCh38
rs374473942 Chr.9: 132386687-132386687 on Build GRCh38
rs193182993 Chr.9: 132386838-132386838 on Build GRCh38
rs773533059 Chr.9: 132386727-132386727 on Build GRCh38
rs749994737 Chr.9: 132386600-132386600 on Build GRCh38
rs551505460 Chr.9: 132386742-132386742 on Build GRCh38
rs749749786 Chr.9: 132386572-132386572 on Build GRCh38
rs919629796 Chr.9: 132386904-132386904 on Build GRCh38
rs1008175947 Chr.9: 132386815-132386815 on Build GRCh38
rs201197439 Chr.9: 132386532-132386532 on Build GRCh38
rs369763860 Chr.9: 132386560-132386560 on Build GRCh38
rs578203981 Chr.9: 132386543-132386543 on Build GRCh38
rs908724647 Chr.9: 132386898-132386898 on Build GRCh38
rs1019525570 Chr.9: 132386820-132386820 on Build GRCh38
rs998930232 Chr.9: 132386800-132386800 on Build GRCh38
rs779176727 Chr.9: 132386624-132386624 on Build GRCh38
rs768179916 Chr.9: 132386676-132386676 on Build GRCh38
rs539279082 Chr.9: 132386726-132386726 on Build GRCh38
rs531397182 Chr.9: 132386686-132386686 on Build GRCh38
rs751861164 Chr.9: 132386507-132386508 on Build GRCh38
rs751478212 Chr.9: 132386587-132386587 on Build GRCh38
rs529179150 Chr.9: 132386700-132386700 on Build GRCh38
rs530893039 Chr.9: 132386596-132386596 on Build GRCh38
rs748566303 Chr.9: 132386681-132386681 on Build GRCh38
rs561347154 Chr.9: 132386688-132386688 on Build GRCh38
rs757494195 Chr.9: 132386628-132386629 on Build GRCh38
rs916445393 Chr.9: 132386672-132386672 on Build GRCh38
rs967465585 Chr.9: 132386599-132386599 on Build GRCh38
rs757168717 Chr.9: 132386588-132386588 on Build GRCh38
rs201487107 Chr.9: 132386528-132386528 on Build GRCh38
rs1166300 Chr.9: 132386857-132386857 on Build GRCh38
rs779908674 Chr.9: 132386612-132386612 on Build GRCh38
rs1011490307 Chr.9: 132386575-132386575 on Build GRCh38
rs748904820 Chr.9: 132386615-132386615 on Build GRCh38
rs183432023 Chr.9: 132386757-132386757 on Build GRCh38
rs772376251 Chr.9: 132386698-132386698 on Build GRCh38
rs574426172 Chr.9: 132386496-132386496 on Build GRCh38
rs553715668 Chr.9: 132386848-132386848 on Build GRCh38
rs141141589 Chr.9: 132386605-132386605 on Build GRCh38
rs781423651 Chr.9: 132386645-132386646 on Build GRCh38
rs1011773243 Chr.9: 132386739-132386739 on Build GRCh38
rs912763582 Chr.9: 132386717-132386717 on Build GRCh38
rs75318823 Chr.9: 132386772-132386772 on Build GRCh38
rs771121467 Chr.9: 132386756-132386756 on Build GRCh38
rs75009763 Chr.9: 132386723-132386723 on Build GRCh38
rs1036151486 Chr.9: 132386805-132386805 on Build GRCh38

View Full Product Details