Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 13705833-13706334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMSB4Y
Entrez Gene ID
9087
Gene Name
thymosin beta 4, Y-linked
Gene Aliases
TB4Y
UniGene
Hs.159201
Gene Chromosome Location
Chr.Y: 13703567-13706024 on Build GRCh38
Transcripts
NM_004202.2

SNP Detail

SNP ID Chromosome Location
rs1039756838 Chr.Y: 13706166-13706166 on Build GRCh38
rs899827960 Chr.Y: 13706282-13706282 on Build GRCh38
rs998480600 Chr.Y: 13706286-13706286 on Build GRCh38
rs941355751 Chr.Y: 13705988-13705988 on Build GRCh38

View Full Product Details