Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42210261-42210752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT5B
Entrez Gene ID
6777
Gene Name
signal transducer and activator of transcription 5B
Gene Aliases
STAT5
UniGene
Hs.595276
Gene Chromosome Location
Chr.17: 42199177-42276460 on Build GRCh38
Transcripts
NM_012448.3 XM_017024977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849413 c.1713G>T p.W571C Chr.17: 42210465-42210465 on Build GRCh38
1383340 c.1749delA p.K583fs*16 Chr.17: 42210429-42210429 on Build GRCh38
6964498 c.1735G>A p.E579K Chr.17: 42210443-42210443 on Build GRCh38
1383339 c.1749_1750insA p.H584fs*8 Chr.17: 42210428-42210429 on Build GRCh38
4066533 c.1806G>T p.Q602H Chr.17: 42210271-42210271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189255705 Chr.17: 42210345-42210345 on Build GRCh38
rs768206896 Chr.17: 42210545-42210545 on Build GRCh38
rs749943637 Chr.17: 42210342-42210343 on Build GRCh38
rs748094482 Chr.17: 42210394-42210394 on Build GRCh38
rs192118598 Chr.17: 42210724-42210724 on Build GRCh38
rs753962351 Chr.17: 42210615-42210615 on Build GRCh38
rs967077155 Chr.17: 42210654-42210654 on Build GRCh38
rs764099461 Chr.17: 42210322-42210322 on Build GRCh38
rs561888537 Chr.17: 42210488-42210488 on Build GRCh38
rs562985404 Chr.17: 42210593-42210593 on Build GRCh38
rs931080846 Chr.17: 42210518-42210518 on Build GRCh38
rs893618560 Chr.17: 42210618-42210618 on Build GRCh38
rs931292108 Chr.17: 42210731-42210731 on Build GRCh38
rs757748627 Chr.17: 42210344-42210344 on Build GRCh38
rs964645095 Chr.17: 42210692-42210692 on Build GRCh38
rs758036191 Chr.17: 42210381-42210382 on Build GRCh38
rs547788769 Chr.17: 42210676-42210676 on Build GRCh38
rs774346811 Chr.17: 42210512-42210512 on Build GRCh38
rs375841379 Chr.17: 42210379-42210379 on Build GRCh38
rs368084521 Chr.17: 42210347-42210347 on Build GRCh38
rs765053192 Chr.17: 42210307-42210308 on Build GRCh38
rs920743191 Chr.17: 42210517-42210517 on Build GRCh38
rs777976212 Chr.17: 42210727-42210727 on Build GRCh38
rs1046759740 Chr.17: 42210554-42210554 on Build GRCh38
rs574003292 Chr.17: 42210511-42210511 on Build GRCh38
rs763332867 Chr.17: 42210450-42210450 on Build GRCh38
rs766608698 Chr.17: 42210600-42210600 on Build GRCh38
rs931422984 Chr.17: 42210315-42210315 on Build GRCh38
rs771960501 Chr.17: 42210414-42210414 on Build GRCh38
rs886782508 Chr.17: 42210576-42210576 on Build GRCh38
rs572365056 Chr.17: 42210423-42210423 on Build GRCh38
rs752917778 Chr.17: 42210304-42210304 on Build GRCh38
rs919883933 Chr.17: 42210289-42210289 on Build GRCh38
rs746737564 Chr.17: 42210426-42210426 on Build GRCh38
rs781517078 Chr.17: 42210478-42210479 on Build GRCh38
rs75130700 Chr.17: 42210453-42210453 on Build GRCh38
rs369496943 Chr.17: 42210378-42210378 on Build GRCh38
rs184941547 Chr.17: 42210725-42210725 on Build GRCh38
rs142538226 Chr.17: 42210626-42210626 on Build GRCh38
rs748907014 Chr.17: 42210375-42210375 on Build GRCh38
rs371550229 Chr.17: 42210368-42210368 on Build GRCh38
rs775637267 Chr.17: 42210462-42210462 on Build GRCh38
rs763027339 Chr.17: 42210305-42210305 on Build GRCh38
rs905133298 Chr.17: 42210609-42210609 on Build GRCh38
rs869067755 Chr.17: 42210414-42210414 on Build GRCh38
rs750877988 Chr.17: 42210349-42210349 on Build GRCh38
rs374070270 Chr.17: 42210433-42210433 on Build GRCh38
rs9897531 Chr.17: 42210690-42210690 on Build GRCh38
rs755310509 Chr.17: 42210625-42210625 on Build GRCh38
rs762372449 Chr.17: 42210514-42210514 on Build GRCh38
rs1032604378 Chr.17: 42210616-42210616 on Build GRCh38
rs1047173285 Chr.17: 42210369-42210369 on Build GRCh38
rs112265774 Chr.17: 42210513-42210513 on Build GRCh38
rs759915585 Chr.17: 42210657-42210657 on Build GRCh38
rs538817503 Chr.17: 42210361-42210361 on Build GRCh38
rs544346977 Chr.17: 42210559-42210559 on Build GRCh38
rs752068991 Chr.17: 42210330-42210330 on Build GRCh38

View Full Product Details