Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42762513-42763016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAMP2
Entrez Gene ID
10266
Gene Name
receptor activity modifying protein 2
Gene Aliases
-
UniGene
Hs.514193
Gene Chromosome Location
Chr.17: 42761194-42763041 on Build GRCh38
Transcripts
NM_005854.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464874 c.349G>A p.E117K Chr.17: 42762673-42762673 on Build GRCh38
6762505 c.420delC p.P142fs*8 Chr.17: 42762744-42762744 on Build GRCh38
6762504 c.334C>T p.P112S Chr.17: 42762658-42762658 on Build GRCh38
5852585 c.497G>A p.W166* Chr.17: 42762821-42762821 on Build GRCh38
4424016 c.318G>T p.L106F Chr.17: 42762642-42762642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778047581 Chr.17: 42762664-42762664 on Build GRCh38
rs543455351 Chr.17: 42762780-42762780 on Build GRCh38
rs942985749 Chr.17: 42762953-42762953 on Build GRCh38
rs763939532 Chr.17: 42762610-42762610 on Build GRCh38
rs1037476643 Chr.17: 42762983-42762983 on Build GRCh38
rs150879757 Chr.17: 42762566-42762567 on Build GRCh38
rs368416415 Chr.17: 42762890-42762890 on Build GRCh38
rs774324010 Chr.17: 42762832-42762832 on Build GRCh38
rs375082267 Chr.17: 42762768-42762768 on Build GRCh38
rs762862599 Chr.17: 42762591-42762591 on Build GRCh38
rs769135756 Chr.17: 42762686-42762686 on Build GRCh38
rs759449482 Chr.17: 42762569-42762569 on Build GRCh38
rs759612918 Chr.17: 42762859-42762859 on Build GRCh38
rs779036418 Chr.17: 42762590-42762590 on Build GRCh38
rs770604025 Chr.17: 42762678-42762678 on Build GRCh38
rs201333261 Chr.17: 42762827-42762827 on Build GRCh38
rs750675555 Chr.17: 42762611-42762611 on Build GRCh38
rs201318951 Chr.17: 42762872-42762872 on Build GRCh38
rs760868836 Chr.17: 42762719-42762719 on Build GRCh38
rs755482043 Chr.17: 42762616-42762616 on Build GRCh38
rs150494057 Chr.17: 42762612-42762612 on Build GRCh38
rs763990662 Chr.17: 42762712-42762712 on Build GRCh38
rs747370685 Chr.17: 42762762-42762762 on Build GRCh38
rs751383102 Chr.17: 42762713-42762713 on Build GRCh38
rs370655794 Chr.17: 42762760-42762760 on Build GRCh38
rs148106984 Chr.17: 42762814-42762814 on Build GRCh38
rs779456989 Chr.17: 42762627-42762627 on Build GRCh38
rs370708803 Chr.17: 42762860-42762860 on Build GRCh38
rs780919908 Chr.17: 42762788-42762788 on Build GRCh38
rs529577821 Chr.17: 42762901-42762901 on Build GRCh38
rs545776585 Chr.17: 42762700-42762700 on Build GRCh38
rs577781935 Chr.17: 42762546-42762546 on Build GRCh38
rs776150642 Chr.17: 42762568-42762568 on Build GRCh38
rs202235735 Chr.17: 42762858-42762858 on Build GRCh38
rs745621671 Chr.17: 42762636-42762637 on Build GRCh38
rs769349889 Chr.17: 42762804-42762804 on Build GRCh38
rs910203866 Chr.17: 42762931-42762931 on Build GRCh38
rs778820854 Chr.17: 42762745-42762745 on Build GRCh38
rs755706645 Chr.17: 42762900-42762900 on Build GRCh38
rs575263689 Chr.17: 42762652-42762652 on Build GRCh38
rs374042252 Chr.17: 42762571-42762572 on Build GRCh38
rs867318232 Chr.17: 42762778-42762778 on Build GRCh38
rs753918093 Chr.17: 42762728-42762728 on Build GRCh38
rs960248221 Chr.17: 42762608-42762608 on Build GRCh38
rs375600296 Chr.17: 42762861-42762861 on Build GRCh38
rs746678610 Chr.17: 42762670-42762670 on Build GRCh38
rs1018500137 Chr.17: 42762883-42762883 on Build GRCh38
rs370523361 Chr.17: 42762553-42762553 on Build GRCh38
rs569552830 Chr.17: 42762988-42762988 on Build GRCh38
rs745468412 Chr.17: 42762792-42762792 on Build GRCh38
rs926002503 Chr.17: 42762741-42762741 on Build GRCh38
rs918347190 Chr.17: 42762575-42762575 on Build GRCh38
rs770537957 Chr.17: 42762776-42762776 on Build GRCh38
rs775597852 Chr.17: 42762688-42762688 on Build GRCh38
rs765189917 Chr.17: 42762587-42762587 on Build GRCh38
rs899341750 Chr.17: 42762566-42762566 on Build GRCh38
rs771763881 Chr.17: 42762554-42762554 on Build GRCh38
rs377604752 Chr.17: 42762744-42762744 on Build GRCh38
rs200795379 Chr.17: 42762567-42762567 on Build GRCh38
rs772460185 Chr.17: 42762661-42762661 on Build GRCh38
rs767171231 Chr.17: 42762856-42762856 on Build GRCh38
rs372584074 Chr.17: 42762724-42762724 on Build GRCh38
rs749294029 Chr.17: 42762821-42762821 on Build GRCh38
rs781343629 Chr.17: 42762885-42762885 on Build GRCh38
rs780052665 Chr.17: 42762613-42762613 on Build GRCh38
rs748772498 Chr.17: 42762903-42762903 on Build GRCh38
rs752483328 Chr.17: 42762590-42762590 on Build GRCh38
rs929765630 Chr.17: 42762582-42762582 on Build GRCh38
rs753944631 Chr.17: 42762614-42762614 on Build GRCh38
rs200498109 Chr.17: 42762758-42762758 on Build GRCh38
rs772876647 Chr.17: 42762562-42762562 on Build GRCh38
rs761649763 Chr.17: 42762850-42762850 on Build GRCh38
rs751961997 Chr.17: 42762865-42762865 on Build GRCh38
rs551269561 Chr.17: 42762977-42762977 on Build GRCh38
rs753208055 Chr.17: 42762757-42762757 on Build GRCh38

View Full Product Details