Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 77841024-77841540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGT1
Entrez Gene ID
84061
Gene Name
magnesium transporter 1
Gene Aliases
IAP, MRX95, OST3B, PRO0756, XMEN, bA217H1.1
UniGene
Hs.323562
Gene Chromosome Location
Chr.X: 77826364-77895568 on Build GRCh38
Transcripts
NM_032121.5

SNP Detail

SNP ID Chromosome Location
rs928666724 Chr.X: 77841115-77841115 on Build GRCh38
rs782328903 Chr.X: 77841410-77841410 on Build GRCh38
rs782465970 Chr.X: 77841367-77841367 on Build GRCh38
rs782735712 Chr.X: 77841371-77841371 on Build GRCh38
rs782748807 Chr.X: 77841275-77841275 on Build GRCh38
rs781924673 Chr.X: 77841238-77841238 on Build GRCh38
rs990341178 Chr.X: 77841489-77841489 on Build GRCh38
rs1055661852 Chr.X: 77841185-77841185 on Build GRCh38
rs782176437 Chr.X: 77841327-77841327 on Build GRCh38
rs782168431 Chr.X: 77841438-77841439 on Build GRCh38
rs374143885 Chr.X: 77841267-77841267 on Build GRCh38
rs782105908 Chr.X: 77841123-77841123 on Build GRCh38
rs782031975 Chr.X: 77841212-77841212 on Build GRCh38
rs782814223 Chr.X: 77841348-77841348 on Build GRCh38
rs112279802 Chr.X: 77841286-77841286 on Build GRCh38
rs137852222 Chr.X: 77841311-77841311 on Build GRCh38
rs888811681 Chr.X: 77841438-77841438 on Build GRCh38
rs941882099 Chr.X: 77841298-77841298 on Build GRCh38
rs1007211300 Chr.X: 77841445-77841445 on Build GRCh38
rs782261573 Chr.X: 77841206-77841206 on Build GRCh38
rs377681364 Chr.X: 77841356-77841356 on Build GRCh38
rs1030076714 Chr.X: 77841361-77841361 on Build GRCh38
rs191651295 Chr.X: 77841112-77841112 on Build GRCh38
rs946087919 Chr.X: 77841096-77841096 on Build GRCh38
rs34302237 Chr.X: 77841195-77841196 on Build GRCh38
rs782114288 Chr.X: 77841508-77841508 on Build GRCh38
rs782315868 Chr.X: 77841228-77841228 on Build GRCh38
rs111841915 Chr.X: 77841476-77841476 on Build GRCh38
rs913327997 Chr.X: 77841066-77841066 on Build GRCh38
rs782007672 Chr.X: 77841278-77841278 on Build GRCh38
rs140857469 Chr.X: 77841316-77841316 on Build GRCh38
rs782372856 Chr.X: 77841210-77841210 on Build GRCh38
rs782732066 Chr.X: 77841129-77841129 on Build GRCh38
rs782142705 Chr.X: 77841224-77841224 on Build GRCh38
rs997716028 Chr.X: 77841349-77841349 on Build GRCh38
rs782124755 Chr.X: 77841333-77841334 on Build GRCh38
rs957578007 Chr.X: 77841471-77841471 on Build GRCh38
rs782120629 Chr.X: 77841287-77841287 on Build GRCh38
rs1018554474 Chr.X: 77841470-77841470 on Build GRCh38
rs183799734 Chr.X: 77841183-77841183 on Build GRCh38

View Full Product Details