Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110034243-110034832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRIP1
Entrez Gene ID
85369
Gene Name
striatin interacting protein 1
Gene Aliases
FAM40A, FAR11A
UniGene
Hs.584996
Gene Chromosome Location
Chr.1: 110031577-110054641 on Build GRCh38
Transcripts
NM_001270768.1 NM_033088.3 NR_073071.1 XM_017002629.1 XM_017002630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391556 c.142G>T p.A48S Chr.1: 110034779-110034779 on Build GRCh38
7415297 c.83C>T p.A28V Chr.1: 110034720-110034720 on Build GRCh38
6381983 c.85G>A p.A29T Chr.1: 110034722-110034722 on Build GRCh38
6391555 c.142G>T p.A48S Chr.1: 110034779-110034779 on Build GRCh38
7415296 c.83C>T p.A28V Chr.1: 110034720-110034720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964242996 Chr.1: 110034703-110034703 on Build GRCh38
rs926280253 Chr.1: 110034553-110034553 on Build GRCh38
rs945503362 Chr.1: 110034584-110034584 on Build GRCh38
rs1032892841 Chr.1: 110034332-110034338 on Build GRCh38
rs773264962 Chr.1: 110034645-110034645 on Build GRCh38
rs962429659 Chr.1: 110034710-110034710 on Build GRCh38
rs982683890 Chr.1: 110034520-110034520 on Build GRCh38
rs899142228 Chr.1: 110034649-110034649 on Build GRCh38
rs1000115039 Chr.1: 110034329-110034329 on Build GRCh38
rs370286507 Chr.1: 110034655-110034655 on Build GRCh38
rs895931130 Chr.1: 110034282-110034282 on Build GRCh38
rs72976670 Chr.1: 110034270-110034270 on Build GRCh38
rs905701037 Chr.1: 110034598-110034598 on Build GRCh38
rs890479169 Chr.1: 110034435-110034435 on Build GRCh38
rs762144036 Chr.1: 110034786-110034786 on Build GRCh38
rs574938927 Chr.1: 110034672-110034672 on Build GRCh38
rs759100551 Chr.1: 110034669-110034669 on Build GRCh38
rs35894520 Chr.1: 110034676-110034676 on Build GRCh38
rs773940350 Chr.1: 110034296-110034296 on Build GRCh38
rs575581066 Chr.1: 110034597-110034597 on Build GRCh38
rs373423235 Chr.1: 110034428-110034428 on Build GRCh38
rs764749204 Chr.1: 110034708-110034708 on Build GRCh38
rs542853573 Chr.1: 110034791-110034791 on Build GRCh38
rs1024756460 Chr.1: 110034758-110034758 on Build GRCh38
rs983883240 Chr.1: 110034538-110034538 on Build GRCh38
rs950194051 Chr.1: 110034561-110034561 on Build GRCh38
rs1010140525 Chr.1: 110034652-110034652 on Build GRCh38
rs553113096 Chr.1: 110034491-110034491 on Build GRCh38
rs905666919 Chr.1: 110034326-110034326 on Build GRCh38
rs890772640 Chr.1: 110034673-110034673 on Build GRCh38
rs1007055556 Chr.1: 110034697-110034697 on Build GRCh38
rs550243521 Chr.1: 110034276-110034276 on Build GRCh38
rs985197632 Chr.1: 110034475-110034475 on Build GRCh38
rs2453937 Chr.1: 110034359-110034359 on Build GRCh38
rs533293624 Chr.1: 110034335-110034335 on Build GRCh38
rs763446096 Chr.1: 110034743-110034752 on Build GRCh38
rs752275144 Chr.1: 110034394-110034394 on Build GRCh38
rs951262147 Chr.1: 110034501-110034501 on Build GRCh38
rs192260434 Chr.1: 110034607-110034607 on Build GRCh38
rs1003971327 Chr.1: 110034722-110034722 on Build GRCh38
rs536454251 Chr.1: 110034488-110034488 on Build GRCh38
rs1017707733 Chr.1: 110034486-110034486 on Build GRCh38
rs892611441 Chr.1: 110034361-110034361 on Build GRCh38
rs914085327 Chr.1: 110034560-110034560 on Build GRCh38
rs749387777 Chr.1: 110034603-110034603 on Build GRCh38
rs764060858 Chr.1: 110034528-110034528 on Build GRCh38
rs567094071 Chr.1: 110034529-110034529 on Build GRCh38
rs936458397 Chr.1: 110034555-110034555 on Build GRCh38
rs533938774 Chr.1: 110034557-110034557 on Build GRCh38
rs759230165 Chr.1: 110034305-110034305 on Build GRCh38
rs1017945615 Chr.1: 110034707-110034707 on Build GRCh38
rs566986042 Chr.1: 110034400-110034400 on Build GRCh38
rs188420033 Chr.1: 110034595-110034595 on Build GRCh38
rs1009851260 Chr.1: 110034380-110034380 on Build GRCh38
rs779214552 Chr.1: 110034622-110034622 on Build GRCh38
rs752434543 Chr.1: 110034715-110034715 on Build GRCh38
rs1027764192 Chr.1: 110034760-110034760 on Build GRCh38
rs1010354007 Chr.1: 110034351-110034351 on Build GRCh38
rs770561705 Chr.1: 110034665-110034665 on Build GRCh38
rs1044651175 Chr.1: 110034309-110034309 on Build GRCh38
rs917796697 Chr.1: 110034522-110034522 on Build GRCh38
rs973892270 Chr.1: 110034492-110034492 on Build GRCh38
rs750861765 Chr.1: 110034677-110034679 on Build GRCh38
rs57032702 Chr.1: 110034626-110034626 on Build GRCh38
rs980439651 Chr.1: 110034533-110034533 on Build GRCh38
rs970660163 Chr.1: 110034759-110034759 on Build GRCh38
rs1026841694 Chr.1: 110034497-110034497 on Build GRCh38
rs747853731 Chr.1: 110034623-110034623 on Build GRCh38
rs1030558704 Chr.1: 110034430-110034430 on Build GRCh38
rs753755654 Chr.1: 110034668-110034668 on Build GRCh38
rs950147130 Chr.1: 110034302-110034302 on Build GRCh38
rs951251061 Chr.1: 110034689-110034689 on Build GRCh38
rs750923470 Chr.1: 110034811-110034811 on Build GRCh38
rs539318479 Chr.1: 110034559-110034559 on Build GRCh38
rs955328622 Chr.1: 110034366-110034366 on Build GRCh38
rs1007526816 Chr.1: 110034470-110034470 on Build GRCh38
rs140729310 Chr.1: 110034549-110034550 on Build GRCh38

View Full Product Details