Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50722555-50722960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIN
Entrez Gene ID
51199
Gene Name
ninein
Gene Aliases
SCKL7
UniGene
Hs.310429
Gene Chromosome Location
Chr.14: 50719763-50831121 on Build GRCh38
Transcripts
NM_020921.3 XM_011536817.1 XM_011536822.1

SNP Detail

SNP ID Chromosome Location
rs760314538 Chr.14: 50722675-50722675 on Build GRCh38
rs770221734 Chr.14: 50722707-50722707 on Build GRCh38
rs1038096955 Chr.14: 50722604-50722604 on Build GRCh38
rs546121965 Chr.14: 50722700-50722700 on Build GRCh38
rs964057273 Chr.14: 50722907-50722907 on Build GRCh38
rs756319594 Chr.14: 50722704-50722704 on Build GRCh38
rs548437962 Chr.14: 50722885-50722885 on Build GRCh38
rs993943683 Chr.14: 50722616-50722616 on Build GRCh38
rs927768618 Chr.14: 50722776-50722776 on Build GRCh38
rs765232134 Chr.14: 50722607-50722607 on Build GRCh38
rs767020882 Chr.14: 50722930-50722930 on Build GRCh38
rs777769271 Chr.14: 50722730-50722730 on Build GRCh38
rs763501855 Chr.14: 50722743-50722743 on Build GRCh38
rs1038427400 Chr.14: 50722892-50722892 on Build GRCh38
rs1020827712 Chr.14: 50722822-50722822 on Build GRCh38
rs374744725 Chr.14: 50722647-50722648 on Build GRCh38
rs896866207 Chr.14: 50722929-50722929 on Build GRCh38
rs867662108 Chr.14: 50722608-50722608 on Build GRCh38
rs890660582 Chr.14: 50722645-50722646 on Build GRCh38
rs866498610 Chr.14: 50722644-50722644 on Build GRCh38
rs531886809 Chr.14: 50722884-50722884 on Build GRCh38
rs931469071 Chr.14: 50722661-50722661 on Build GRCh38
rs957915747 Chr.14: 50722777-50722777 on Build GRCh38
rs1047193322 Chr.14: 50722680-50722680 on Build GRCh38
rs920149743 Chr.14: 50722622-50722622 on Build GRCh38
rs773386619 Chr.14: 50722729-50722729 on Build GRCh38
rs1046553163 Chr.14: 50722714-50722714 on Build GRCh38
rs990729066 Chr.14: 50722831-50722831 on Build GRCh38
rs939191102 Chr.14: 50722827-50722827 on Build GRCh38
rs149410483 Chr.14: 50722850-50722850 on Build GRCh38
rs887266520 Chr.14: 50722691-50722691 on Build GRCh38
rs1009892938 Chr.14: 50722797-50722797 on Build GRCh38
rs920181000 Chr.14: 50722931-50722931 on Build GRCh38
rs904882265 Chr.14: 50722717-50722717 on Build GRCh38

View Full Product Details