Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 33834773-33835339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC3
Entrez Gene ID
5983
Gene Name
replication factor C subunit 3
Gene Aliases
RFC38
UniGene
Hs.115474
Gene Chromosome Location
Chr.13: 33818049-33973946 on Build GRCh38
Transcripts
NM_002915.3 NM_181558.2 XM_011535172.2 XM_011535173.2 XM_011535174.2 XM_011535175.1 XM_017020680.1 XM_017020681.1 XM_017020682.1 XM_017020683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047165 c.855T>C p.C285C Chr.13: 33835193-33835193 on Build GRCh38
5376678 c.814C>T p.L272F Chr.13: 33835152-33835152 on Build GRCh38
1317898 c.810G>A p.R270R Chr.13: 33835148-33835148 on Build GRCh38
5376679 c.814C>T p.L272F Chr.13: 33835152-33835152 on Build GRCh38
1264034 c.876G>A p.M292I Chr.13: 33835214-33835214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777082937 Chr.13: 33835295-33835295 on Build GRCh38
rs931179279 Chr.13: 33834984-33834984 on Build GRCh38
rs915160624 Chr.13: 33835046-33835046 on Build GRCh38
rs545659711 Chr.13: 33835307-33835307 on Build GRCh38
rs980707895 Chr.13: 33835100-33835100 on Build GRCh38
rs756817004 Chr.13: 33835298-33835298 on Build GRCh38
rs766785883 Chr.13: 33834909-33834909 on Build GRCh38
rs772709697 Chr.13: 33835125-33835125 on Build GRCh38
rs571033858 Chr.13: 33835004-33835004 on Build GRCh38
rs779597368 Chr.13: 33835200-33835200 on Build GRCh38
rs765178534 Chr.13: 33835142-33835142 on Build GRCh38
rs919766137 Chr.13: 33834954-33834954 on Build GRCh38
rs561232918 Chr.13: 33834796-33834796 on Build GRCh38
rs755494831 Chr.13: 33835194-33835194 on Build GRCh38
rs112189970 Chr.13: 33835312-33835312 on Build GRCh38
rs761308126 Chr.13: 33834825-33834825 on Build GRCh38
rs761845498 Chr.13: 33835161-33835161 on Build GRCh38
rs749553215 Chr.13: 33834836-33834837 on Build GRCh38
rs1805375 Chr.13: 33835114-33835114 on Build GRCh38
rs776557773 Chr.13: 33835130-33835130 on Build GRCh38
rs776235365 Chr.13: 33835246-33835246 on Build GRCh38
rs954410201 Chr.13: 33834919-33834919 on Build GRCh38
rs771810424 Chr.13: 33835117-33835117 on Build GRCh38
rs769614237 Chr.13: 33835253-33835253 on Build GRCh38
rs748705179 Chr.13: 33835224-33835224 on Build GRCh38
rs745691597 Chr.13: 33835248-33835248 on Build GRCh38
rs757826136 Chr.13: 33835226-33835226 on Build GRCh38
rs764492227 Chr.13: 33835276-33835277 on Build GRCh38
rs759404833 Chr.13: 33835135-33835135 on Build GRCh38
rs750596331 Chr.13: 33835178-33835178 on Build GRCh38
rs765746004 Chr.13: 33835277-33835277 on Build GRCh38
rs775288883 Chr.13: 33835072-33835072 on Build GRCh38
rs374999480 Chr.13: 33834823-33834823 on Build GRCh38
rs775379812 Chr.13: 33835254-33835254 on Build GRCh38
rs755363742 Chr.13: 33835276-33835276 on Build GRCh38
rs372589710 Chr.13: 33835260-33835260 on Build GRCh38
rs754247660 Chr.13: 33834965-33834965 on Build GRCh38
rs3135627 Chr.13: 33834797-33834797 on Build GRCh38
rs1038605872 Chr.13: 33834813-33834813 on Build GRCh38
rs377335265 Chr.13: 33835266-33835266 on Build GRCh38
rs777403284 Chr.13: 33835233-33835233 on Build GRCh38
rs769289596 Chr.13: 33835112-33835112 on Build GRCh38
rs755141958 Chr.13: 33835202-33835202 on Build GRCh38
rs369435129 Chr.13: 33835126-33835126 on Build GRCh38
rs780722086 Chr.13: 33835287-33835287 on Build GRCh38
rs1047033110 Chr.13: 33835006-33835009 on Build GRCh38
rs373733404 Chr.13: 33835129-33835129 on Build GRCh38
rs761165009 Chr.13: 33835128-33835129 on Build GRCh38
rs780789577 Chr.13: 33835301-33835301 on Build GRCh38
rs780525457 Chr.13: 33835102-33835102 on Build GRCh38
rs956546386 Chr.13: 33835034-33835034 on Build GRCh38
rs756881076 Chr.13: 33835271-33835271 on Build GRCh38
rs144033374 Chr.13: 33835115-33835115 on Build GRCh38
rs567007265 Chr.13: 33834852-33834852 on Build GRCh38
rs3783269 Chr.13: 33834927-33834927 on Build GRCh38
rs370230014 Chr.13: 33835244-33835244 on Build GRCh38
rs745355588 Chr.13: 33835310-33835310 on Build GRCh38
rs55657632 Chr.13: 33834873-33834873 on Build GRCh38
rs775683456 Chr.13: 33835105-33835125 on Build GRCh38
rs868361632 Chr.13: 33834868-33834868 on Build GRCh38
rs375140332 Chr.13: 33835268-33835268 on Build GRCh38
rs754267210 Chr.13: 33835192-33835192 on Build GRCh38
rs1038981804 Chr.13: 33835201-33835201 on Build GRCh38
rs376806684 Chr.13: 33834888-33834888 on Build GRCh38
rs968302796 Chr.13: 33834798-33834798 on Build GRCh38
rs3135628 Chr.13: 33835105-33835105 on Build GRCh38
rs572180418 Chr.13: 33835286-33835286 on Build GRCh38
rs774371943 Chr.13: 33835155-33835155 on Build GRCh38
rs746641947 Chr.13: 33835234-33835234 on Build GRCh38
rs1028666456 Chr.13: 33834882-33834882 on Build GRCh38
rs376913448 Chr.13: 33835190-33835190 on Build GRCh38
rs1030815289 Chr.13: 33834938-33834938 on Build GRCh38
rs373482833 Chr.13: 33835265-33835265 on Build GRCh38
rs756411029 Chr.13: 33835099-33835099 on Build GRCh38
rs969715998 Chr.13: 33835048-33835048 on Build GRCh38
rs745564914 Chr.13: 33835302-33835302 on Build GRCh38
rs1008589926 Chr.13: 33835269-33835269 on Build GRCh38
rs138529478 Chr.13: 33835045-33835045 on Build GRCh38
rs746545160 Chr.13: 33835296-33835296 on Build GRCh38
rs975282677 Chr.13: 33834941-33834941 on Build GRCh38
rs751166752 Chr.13: 33835162-33835162 on Build GRCh38

View Full Product Details