Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227248225-227248705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3
Entrez Gene ID
1285
Gene Name
collagen type IV alpha 3 chain
Gene Aliases
-
UniGene
Hs.570065
Gene Chromosome Location
Chr.2: 227164565-227314792 on Build GRCh38
Transcripts
NM_000091.4 XM_005246277.3 XM_005246280.3 XM_006712245.3 XM_011510555.1 XM_017003295.1 NR_102371.1

Gene Symbol
LOC654841
Entrez Gene ID
654841
Gene Name
uncharacterized LOC654841
Gene Aliases
-
UniGene
Hs.659982
Gene Chromosome Location
Chr.2: 227221052-227325164 on Build GRCh38
Transcripts
NM_000091.4 XM_005246277.3 XM_005246280.3 XM_006712245.3 XM_011510555.1 XM_017003295.1 NR_102371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6633223 c.468G>T p.K156N Chr.2: 227248442-227248442 on Build GRCh38
248990 c.520G>A p.G174R Chr.2: 227248494-227248494 on Build GRCh38
6633222 c.469-1G>T p.? Chr.2: 227248442-227248442 on Build GRCh38
6499747 c.475C>G p.P159A Chr.2: 227248449-227248449 on Build GRCh38
6499748 c.475C>G p.P159A Chr.2: 227248449-227248449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766778471 Chr.2: 227248392-227248392 on Build GRCh38
rs773539131 Chr.2: 227248353-227248353 on Build GRCh38
rs764834208 Chr.2: 227248515-227248515 on Build GRCh38
rs759869958 Chr.2: 227248501-227248501 on Build GRCh38
rs540689943 Chr.2: 227248563-227248563 on Build GRCh38
rs200363147 Chr.2: 227248410-227248410 on Build GRCh38
rs927138124 Chr.2: 227248564-227248564 on Build GRCh38
rs199514043 Chr.2: 227248487-227248487 on Build GRCh38
rs555907134 Chr.2: 227248460-227248460 on Build GRCh38
rs144036466 Chr.2: 227248468-227248468 on Build GRCh38
rs1041401425 Chr.2: 227248347-227248347 on Build GRCh38
rs369769279 Chr.2: 227248471-227248471 on Build GRCh38
rs775936375 Chr.2: 227248516-227248516 on Build GRCh38
rs756992000 Chr.2: 227248335-227248335 on Build GRCh38
rs753235176 Chr.2: 227248312-227248312 on Build GRCh38
rs764451365 Chr.2: 227248443-227248443 on Build GRCh38
rs193207353 Chr.2: 227248628-227248628 on Build GRCh38
rs766133774 Chr.2: 227248564-227248567 on Build GRCh38
rs34713188 Chr.2: 227248438-227248439 on Build GRCh38
rs904159142 Chr.2: 227248616-227248616 on Build GRCh38
rs760144611 Chr.2: 227248662-227248662 on Build GRCh38
rs954758632 Chr.2: 227248256-227248256 on Build GRCh38
rs577395398 Chr.2: 227248577-227248578 on Build GRCh38
rs935550229 Chr.2: 227248584-227248584 on Build GRCh38
rs780818056 Chr.2: 227248336-227248336 on Build GRCh38
rs369945847 Chr.2: 227248455-227248455 on Build GRCh38
rs1041409668 Chr.2: 227248604-227248604 on Build GRCh38
rs942117712 Chr.2: 227248294-227248294 on Build GRCh38
rs764516702 Chr.2: 227248361-227248362 on Build GRCh38
rs554811380 Chr.2: 227248570-227248570 on Build GRCh38
rs748974369 Chr.2: 227248478-227248478 on Build GRCh38
rs183668882 Chr.2: 227248330-227248330 on Build GRCh38
rs914863009 Chr.2: 227248597-227248597 on Build GRCh38
rs755392596 Chr.2: 227248288-227248288 on Build GRCh38
rs774154890 Chr.2: 227248484-227248484 on Build GRCh38
rs1016662285 Chr.2: 227248434-227248434 on Build GRCh38
rs751986632 Chr.2: 227248528-227248528 on Build GRCh38
rs768408068 Chr.2: 227248367-227248367 on Build GRCh38
rs116673692 Chr.2: 227248679-227248679 on Build GRCh38
rs1039468617 Chr.2: 227248295-227248295 on Build GRCh38
rs543607129 Chr.2: 227248660-227248660 on Build GRCh38
rs749865170 Chr.2: 227248557-227248557 on Build GRCh38
rs573388793 Chr.2: 227248546-227248546 on Build GRCh38
rs140415172 Chr.2: 227248564-227248565 on Build GRCh38
rs983406507 Chr.2: 227248263-227248263 on Build GRCh38
rs908002134 Chr.2: 227248272-227248272 on Build GRCh38
rs188324379 Chr.2: 227248506-227248506 on Build GRCh38
rs544930411 Chr.2: 227248596-227248596 on Build GRCh38
rs574806759 Chr.2: 227248581-227248581 on Build GRCh38
rs764701248 Chr.2: 227248525-227248525 on Build GRCh38
rs371328070 Chr.2: 227248349-227248349 on Build GRCh38
rs753336288 Chr.2: 227248476-227248477 on Build GRCh38
rs779826337 Chr.2: 227248474-227248474 on Build GRCh38
rs745701812 Chr.2: 227248338-227248338 on Build GRCh38
rs777375490 Chr.2: 227248331-227248331 on Build GRCh38
rs749276331 Chr.2: 227248359-227248359 on Build GRCh38
rs751343987 Chr.2: 227248563-227248564 on Build GRCh38
rs1014839148 Chr.2: 227248494-227248494 on Build GRCh38
rs773350204 Chr.2: 227248373-227248373 on Build GRCh38
rs759455097 Chr.2: 227248490-227248490 on Build GRCh38
rs377575924 Chr.2: 227248488-227248488 on Build GRCh38
rs761358728 Chr.2: 227248493-227248494 on Build GRCh38
rs750137445 Chr.2: 227248545-227248547 on Build GRCh38
rs768701718 Chr.2: 227248479-227248479 on Build GRCh38
rs1023021268 Chr.2: 227248433-227248433 on Build GRCh38
rs1039482006 Chr.2: 227248641-227248641 on Build GRCh38
rs752965775 Chr.2: 227248431-227248431 on Build GRCh38
rs970846003 Chr.2: 227248571-227248571 on Build GRCh38
rs112387221 Chr.2: 227248300-227248300 on Build GRCh38
rs574993401 Chr.2: 227248582-227248582 on Build GRCh38
rs762256794 Chr.2: 227248543-227248543 on Build GRCh38
rs746437816 Chr.2: 227248332-227248332 on Build GRCh38
rs776965001 Chr.2: 227248397-227248397 on Build GRCh38
rs945907024 Chr.2: 227248592-227248592 on Build GRCh38
rs755773167 Chr.2: 227248566-227248566 on Build GRCh38
rs745540374 Chr.2: 227248469-227248469 on Build GRCh38
rs758949595 Chr.2: 227248439-227248439 on Build GRCh38
rs111564047 Chr.2: 227248390-227248390 on Build GRCh38
rs6436669 Chr.2: 227248459-227248459 on Build GRCh38

View Full Product Details