Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114323470-114324061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAMP1
Entrez Gene ID
283489
Gene Name
chromosome alignment maintaining phosphoprotein 1
Gene Aliases
C13orf8, CAMP, CHAMP, MRD40, ZNF828
UniGene
Hs.7542
Gene Chromosome Location
Chr.13: 114314490-114327328 on Build GRCh38
Transcripts
NM_001164144.1 NM_001164145.1 NM_032436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5767042 c.79G>C p.E27Q Chr.13: 114323921-114323921 on Build GRCh38
6627728 c.22C>T p.R8C Chr.13: 114323864-114323864 on Build GRCh38
4671918 c.38G>A p.R13H Chr.13: 114323880-114323880 on Build GRCh38
3467478 c.167A>T p.Q56L Chr.13: 114324009-114324009 on Build GRCh38
3467479 c.192C>T p.C64C Chr.13: 114324034-114324034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146349096 Chr.13: 114323923-114323923 on Build GRCh38
rs188279098 Chr.13: 114323817-114323817 on Build GRCh38
rs143869900 Chr.13: 114323896-114323896 on Build GRCh38
rs1024243092 Chr.13: 114323569-114323569 on Build GRCh38
rs782635495 Chr.13: 114323812-114323812 on Build GRCh38
rs1028143785 Chr.13: 114323829-114323829 on Build GRCh38
rs782287224 Chr.13: 114323792-114323792 on Build GRCh38
rs782567790 Chr.13: 114323799-114323799 on Build GRCh38
rs552664873 Chr.13: 114323709-114323709 on Build GRCh38
rs751336149 Chr.13: 114323571-114323571 on Build GRCh38
rs782421881 Chr.13: 114324034-114324034 on Build GRCh38
rs782620299 Chr.13: 114323916-114323916 on Build GRCh38
rs201032426 Chr.13: 114323859-114323859 on Build GRCh38
rs903359991 Chr.13: 114323695-114323695 on Build GRCh38
rs782600313 Chr.13: 114323512-114323512 on Build GRCh38
rs782162864 Chr.13: 114324012-114324012 on Build GRCh38
rs944959675 Chr.13: 114323655-114323655 on Build GRCh38
rs201268340 Chr.13: 114323879-114323879 on Build GRCh38
rs781942990 Chr.13: 114324038-114324038 on Build GRCh38
rs782050747 Chr.13: 114323823-114323823 on Build GRCh38
rs1024277494 Chr.13: 114323750-114323750 on Build GRCh38
rs986900129 Chr.13: 114323652-114323652 on Build GRCh38
rs370877959 Chr.13: 114323914-114323914 on Build GRCh38
rs267603766 Chr.13: 114323936-114323936 on Build GRCh38
rs782356892 Chr.13: 114323835-114323835 on Build GRCh38
rs1009139477 Chr.13: 114323519-114323519 on Build GRCh38
rs114596472 Chr.13: 114323553-114323553 on Build GRCh38
rs1041923688 Chr.13: 114323528-114323528 on Build GRCh38
rs754959663 Chr.13: 114323699-114323699 on Build GRCh38
rs932399165 Chr.13: 114323640-114323640 on Build GRCh38
rs781801530 Chr.13: 114323877-114323877 on Build GRCh38
rs371511810 Chr.13: 114323864-114323864 on Build GRCh38
rs782471389 Chr.13: 114323795-114323795 on Build GRCh38
rs747725810 Chr.13: 114323919-114323919 on Build GRCh38
rs921051848 Chr.13: 114323636-114323636 on Build GRCh38
rs60243226 Chr.13: 114323836-114323836 on Build GRCh38
rs782377487 Chr.13: 114323810-114323810 on Build GRCh38
rs962825290 Chr.13: 114323602-114323602 on Build GRCh38
rs781909783 Chr.13: 114323868-114323868 on Build GRCh38
rs1033479717 Chr.13: 114323542-114323542 on Build GRCh38
rs143588592 Chr.13: 114323533-114323533 on Build GRCh38
rs782665511 Chr.13: 114323895-114323895 on Build GRCh38
rs781988354 Chr.13: 114323948-114323948 on Build GRCh38
rs184650795 Chr.13: 114323765-114323765 on Build GRCh38
rs1013372584 Chr.13: 114323563-114323563 on Build GRCh38
rs782387383 Chr.13: 114323939-114323939 on Build GRCh38
rs971318067 Chr.13: 114323570-114323570 on Build GRCh38
rs782083386 Chr.13: 114323926-114323926 on Build GRCh38
rs908384796 Chr.13: 114323582-114323582 on Build GRCh38
rs201860837 Chr.13: 114323887-114323887 on Build GRCh38
rs782156847 Chr.13: 114323872-114323872 on Build GRCh38
rs782202585 Chr.13: 114323802-114323802 on Build GRCh38
rs531745345 Chr.13: 114323727-114323727 on Build GRCh38
rs1012991307 Chr.13: 114323725-114323725 on Build GRCh38
rs781950831 Chr.13: 114323836-114323836 on Build GRCh38
rs193071805 Chr.13: 114323654-114323654 on Build GRCh38
rs374161591 Chr.13: 114323876-114323876 on Build GRCh38
rs782231322 Chr.13: 114323578-114323578 on Build GRCh38
rs530792654 Chr.13: 114323686-114323686 on Build GRCh38
rs782131661 Chr.13: 114323852-114323852 on Build GRCh38
rs532970702 Chr.13: 114323681-114323681 on Build GRCh38
rs1004092408 Chr.13: 114323773-114323773 on Build GRCh38
rs536826000 Chr.13: 114323713-114323713 on Build GRCh38

View Full Product Details