Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114178140-114178646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
UniGene
Hs.494892
Gene Chromosome Location
Chr.9: 114154271-114312516 on Build GRCh38
Transcripts
NM_032888.3 XM_006717308.2 XM_006717310.3 XM_011519138.2 XM_011519140.1 XM_011519142.2 XM_011519143.2 XM_011519144.2 XM_017015239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3847437 c.1960C>T p.R654W Chr.9: 114178342-114178342 on Build GRCh38
1104349 c.1909G>A p.G637S Chr.9: 114178291-114178291 on Build GRCh38
1201863 c.1933C>T p.P645S Chr.9: 114178315-114178315 on Build GRCh38
7297310 c.1930C>A p.L644I Chr.9: 114178312-114178312 on Build GRCh38
7331884 c.1932delA p.P645fs*103 Chr.9: 114178314-114178314 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs13295729 Chr.9: 114178209-114178209 on Build GRCh38
rs747859207 Chr.9: 114178258-114178258 on Build GRCh38
rs902816326 Chr.9: 114178182-114178182 on Build GRCh38
rs753952153 Chr.9: 114178298-114178298 on Build GRCh38
rs755822672 Chr.9: 114178379-114178379 on Build GRCh38
rs934294786 Chr.9: 114178469-114178469 on Build GRCh38
rs1013111766 Chr.9: 114178276-114178276 on Build GRCh38
rs35468375 Chr.9: 114178209-114178210 on Build GRCh38
rs1040568035 Chr.9: 114178451-114178451 on Build GRCh38
rs376853468 Chr.9: 114178334-114178334 on Build GRCh38
rs551332975 Chr.9: 114178599-114178599 on Build GRCh38
rs569678751 Chr.9: 114178295-114178295 on Build GRCh38
rs780732482 Chr.9: 114178360-114178360 on Build GRCh38
rs1002721566 Chr.9: 114178570-114178570 on Build GRCh38
rs1054673491 Chr.9: 114178527-114178527 on Build GRCh38
rs555301537 Chr.9: 114178180-114178180 on Build GRCh38
rs898458064 Chr.9: 114178623-114178623 on Build GRCh38
rs776853346 Chr.9: 114178460-114178460 on Build GRCh38
rs1055043869 Chr.9: 114178268-114178268 on Build GRCh38
rs368954439 Chr.9: 114178254-114178254 on Build GRCh38
rs369455576 Chr.9: 114178361-114178361 on Build GRCh38
rs538164777 Chr.9: 114178297-114178305 on Build GRCh38
rs369313105 Chr.9: 114178447-114178447 on Build GRCh38
rs889310560 Chr.9: 114178172-114178172 on Build GRCh38
rs1054107138 Chr.9: 114178530-114178530 on Build GRCh38
rs944878213 Chr.9: 114178436-114178436 on Build GRCh38
rs752000426 Chr.9: 114178624-114178624 on Build GRCh38
rs995857456 Chr.9: 114178584-114178584 on Build GRCh38
rs1012668078 Chr.9: 114178550-114178550 on Build GRCh38
rs900004198 Chr.9: 114178554-114178554 on Build GRCh38
rs759981220 Chr.9: 114178295-114178296 on Build GRCh38
rs767253126 Chr.9: 114178425-114178425 on Build GRCh38
rs562471664 Chr.9: 114178204-114178204 on Build GRCh38
rs903457254 Chr.9: 114178453-114178453 on Build GRCh38
rs539692896 Chr.9: 114178556-114178556 on Build GRCh38
rs565845108 Chr.9: 114178571-114178571 on Build GRCh38
rs760156443 Chr.9: 114178342-114178342 on Build GRCh38
rs536118096 Chr.9: 114178297-114178297 on Build GRCh38
rs998538002 Chr.9: 114178183-114178183 on Build GRCh38
rs1000437819 Chr.9: 114178210-114178210 on Build GRCh38
rs531637592 Chr.9: 114178223-114178223 on Build GRCh38
rs139991707 Chr.9: 114178333-114178333 on Build GRCh38
rs375701996 Chr.9: 114178417-114178417 on Build GRCh38
rs936267674 Chr.9: 114178510-114178510 on Build GRCh38
rs986653729 Chr.9: 114178409-114178409 on Build GRCh38
rs777242533 Chr.9: 114178385-114178385 on Build GRCh38
rs201890855 Chr.9: 114178541-114178541 on Build GRCh38
rs530059083 Chr.9: 114178544-114178544 on Build GRCh38
rs772657447 Chr.9: 114178294-114178294 on Build GRCh38
rs1051262082 Chr.9: 114178566-114178566 on Build GRCh38
rs774821525 Chr.9: 114178278-114178278 on Build GRCh38
rs202026497 Chr.9: 114178269-114178269 on Build GRCh38
rs780679157 Chr.9: 114178255-114178255 on Build GRCh38
rs972108987 Chr.9: 114178343-114178343 on Build GRCh38
rs949787670 Chr.9: 114178446-114178446 on Build GRCh38
rs764408402 Chr.9: 114178390-114178390 on Build GRCh38
rs981318664 Chr.9: 114178328-114178328 on Build GRCh38
rs3810930 Chr.9: 114178314-114178314 on Build GRCh38
rs759283560 Chr.9: 114178396-114178396 on Build GRCh38
rs753310687 Chr.9: 114178538-114178538 on Build GRCh38
rs79869936 Chr.9: 114178228-114178228 on Build GRCh38
rs746256798 Chr.9: 114178395-114178395 on Build GRCh38
rs569026789 Chr.9: 114178418-114178418 on Build GRCh38
rs762171990 Chr.9: 114178302-114178302 on Build GRCh38
rs750259036 Chr.9: 114178330-114178330 on Build GRCh38
rs2116404 Chr.9: 114178605-114178605 on Build GRCh38
rs917037490 Chr.9: 114178443-114178443 on Build GRCh38
rs79374574 Chr.9: 114178583-114178583 on Build GRCh38
rs147746522 Chr.9: 114178181-114178181 on Build GRCh38
rs907002751 Chr.9: 114178561-114178561 on Build GRCh38

View Full Product Details