Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146458916-146459428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCERG1
Entrez Gene ID
10915
Gene Name
transcription elongation regulator 1
Gene Aliases
CA150, TAF2S, Urn1
UniGene
Hs.443465
Gene Chromosome Location
Chr.5: 146447124-146511508 on Build GRCh38
Transcripts
NM_001040006.1 NM_006706.3 XM_005268365.2 XM_017008977.1 XM_017008978.1 XM_017008979.1 XM_017008980.1 XM_017008981.1 XM_017008982.1 XM_017008983.1 XM_017008984.1 XM_017008985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
482282 c.693T>C p.A231A Chr.5: 146459138-146459138 on Build GRCh38
5577111 c.730C>T p.Q244* Chr.5: 146459175-146459175 on Build GRCh38
6798187 c.651C>A p.A217A Chr.5: 146459096-146459096 on Build GRCh38
4159607 c.654G>A p.Q218Q Chr.5: 146459099-146459099 on Build GRCh38
6169701 c.506A>T p.Q169L Chr.5: 146458951-146458951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184183473 Chr.5: 146459347-146459347 on Build GRCh38
rs758266082 Chr.5: 146459121-146459150 on Build GRCh38
rs772707990 Chr.5: 146459179-146459180 on Build GRCh38
rs780077931 Chr.5: 146459239-146459239 on Build GRCh38
rs758231104 Chr.5: 146459067-146459090 on Build GRCh38
rs764669499 Chr.5: 146459051-146459052 on Build GRCh38
rs746588350 Chr.5: 146459126-146459126 on Build GRCh38
rs768265456 Chr.5: 146459130-146459130 on Build GRCh38
rs773095770 Chr.5: 146459031-146459048 on Build GRCh38
rs771088123 Chr.5: 146459073-146459084 on Build GRCh38
rs532984875 Chr.5: 146459081-146459081 on Build GRCh38
rs746150906 Chr.5: 146459399-146459399 on Build GRCh38
rs772057166 Chr.5: 146459016-146459051 on Build GRCh38
rs906781434 Chr.5: 146459136-146459136 on Build GRCh38
rs192744974 Chr.5: 146459287-146459287 on Build GRCh38
rs748150998 Chr.5: 146458994-146458995 on Build GRCh38
rs141891503 Chr.5: 146459203-146459203 on Build GRCh38
rs747155353 Chr.5: 146459024-146459024 on Build GRCh38
rs765798582 Chr.5: 146458993-146458993 on Build GRCh38
rs747040868 Chr.5: 146459072-146459073 on Build GRCh38
rs1041574130 Chr.5: 146459054-146459054 on Build GRCh38
rs569890952 Chr.5: 146459093-146459093 on Build GRCh38
rs772720489 Chr.5: 146459366-146459366 on Build GRCh38
rs766972196 Chr.5: 146459186-146459186 on Build GRCh38
rs777657722 Chr.5: 146459041-146459041 on Build GRCh38
rs150864535 Chr.5: 146459352-146459352 on Build GRCh38
rs781190996 Chr.5: 146459147-146459148 on Build GRCh38
rs1037012767 Chr.5: 146458942-146458942 on Build GRCh38
rs771012405 Chr.5: 146459031-146459078 on Build GRCh38
rs769497017 Chr.5: 146459278-146459278 on Build GRCh38
rs568528395 Chr.5: 146459165-146459165 on Build GRCh38
rs777792044 Chr.5: 146459068-146459072 on Build GRCh38
rs758454423 Chr.5: 146459009-146459009 on Build GRCh38
rs527340881 Chr.5: 146459156-146459173 on Build GRCh38
rs375149530 Chr.5: 146459197-146459197 on Build GRCh38
rs1047264528 Chr.5: 146459301-146459301 on Build GRCh38
rs775049784 Chr.5: 146459289-146459289 on Build GRCh38
rs372905217 Chr.5: 146459011-146459011 on Build GRCh38
rs759866842 Chr.5: 146459034-146459051 on Build GRCh38
rs765967676 Chr.5: 146459105-146459106 on Build GRCh38
rs112548582 Chr.5: 146459090-146459090 on Build GRCh38
rs151176678 Chr.5: 146459048-146459048 on Build GRCh38
rs373180463 Chr.5: 146459386-146459386 on Build GRCh38
rs754214673 Chr.5: 146458989-146458989 on Build GRCh38
rs760843114 Chr.5: 146459031-146459060 on Build GRCh38
rs775735592 Chr.5: 146459157-146459168 on Build GRCh38
rs369242802 Chr.5: 146459125-146459125 on Build GRCh38
rs756436468 Chr.5: 146459212-146459212 on Build GRCh38
rs764485976 Chr.5: 146459093-146459094 on Build GRCh38
rs777544244 Chr.5: 146459127-146459132 on Build GRCh38
rs371911957 Chr.5: 146459370-146459370 on Build GRCh38
rs566609153 Chr.5: 146459092-146459092 on Build GRCh38
rs749062766 Chr.5: 146459102-146459102 on Build GRCh38
rs765915833 Chr.5: 146459066-146459066 on Build GRCh38
rs202040285 Chr.5: 146459382-146459382 on Build GRCh38
rs754911983 Chr.5: 146459329-146459329 on Build GRCh38
rs770017912 Chr.5: 146459073-146459078 on Build GRCh38
rs776830305 Chr.5: 146459073-146459090 on Build GRCh38
rs1018383550 Chr.5: 146459374-146459374 on Build GRCh38
rs745952325 Chr.5: 146459150-146459151 on Build GRCh38
rs776965127 Chr.5: 146459031-146459066 on Build GRCh38
rs377038308 Chr.5: 146459313-146459313 on Build GRCh38
rs760398636 Chr.5: 146458946-146458946 on Build GRCh38
rs749584163 Chr.5: 146459156-146459157 on Build GRCh38
rs115619679 Chr.5: 146459281-146459281 on Build GRCh38
rs754608446 Chr.5: 146459106-146459117 on Build GRCh38
rs752828469 Chr.5: 146459299-146459299 on Build GRCh38
rs140816235 Chr.5: 146459030-146459030 on Build GRCh38
rs768611051 Chr.5: 146458988-146458989 on Build GRCh38
rs200533952 Chr.5: 146459291-146459291 on Build GRCh38
rs763396233 Chr.5: 146459094-146459117 on Build GRCh38
rs769131018 Chr.5: 146459162-146459185 on Build GRCh38
rs967357981 Chr.5: 146459185-146459185 on Build GRCh38
rs200722011 Chr.5: 146459006-146459006 on Build GRCh38
rs190572930 Chr.5: 146459042-146459042 on Build GRCh38
rs771280596 Chr.5: 146459350-146459350 on Build GRCh38
rs750482907 Chr.5: 146459003-146459003 on Build GRCh38
rs773627794 Chr.5: 146459177-146459177 on Build GRCh38
rs113361353 Chr.5: 146459117-146459117 on Build GRCh38
rs889664676 Chr.5: 146459056-146459056 on Build GRCh38
rs763393898 Chr.5: 146459048-146459051 on Build GRCh38
rs374406846 Chr.5: 146459385-146459385 on Build GRCh38
rs779776464 Chr.5: 146458983-146459000 on Build GRCh38
rs753610800 Chr.5: 146459067-146459084 on Build GRCh38
rs760099242 Chr.5: 146459099-146459100 on Build GRCh38
rs778789072 Chr.5: 146459066-146459067 on Build GRCh38
rs763306838 Chr.5: 146459181-146459181 on Build GRCh38
rs758260296 Chr.5: 146459323-146459323 on Build GRCh38
rs187468589 Chr.5: 146459084-146459084 on Build GRCh38
rs761474918 Chr.5: 146459133-146459133 on Build GRCh38
rs775767815 Chr.5: 146459088-146459117 on Build GRCh38
rs376562428 Chr.5: 146459377-146459377 on Build GRCh38
rs928458208 Chr.5: 146459211-146459211 on Build GRCh38
rs774707545 Chr.5: 146459169-146459180 on Build GRCh38
rs199852950 Chr.5: 146459326-146459326 on Build GRCh38
rs765476368 Chr.5: 146459161-146459161 on Build GRCh38
rs112714517 Chr.5: 146459051-146459051 on Build GRCh38
rs528556803 Chr.5: 146459018-146459018 on Build GRCh38
rs906901287 Chr.5: 146459364-146459364 on Build GRCh38
rs757747670 Chr.5: 146458992-146458992 on Build GRCh38
rs760849269 Chr.5: 146459380-146459380 on Build GRCh38
rs764442857 Chr.5: 146459306-146459306 on Build GRCh38
rs749851313 Chr.5: 146459052-146459069 on Build GRCh38
rs371770535 Chr.5: 146459129-146459129 on Build GRCh38
rs559455499 Chr.5: 146459012-146459012 on Build GRCh38
rs768102251 Chr.5: 146459266-146459266 on Build GRCh38
rs570448344 Chr.5: 146459063-146459063 on Build GRCh38
rs754374551 Chr.5: 146459072-146459072 on Build GRCh38
rs35276149 Chr.5: 146458986-146458986 on Build GRCh38
rs752342392 Chr.5: 146458943-146458943 on Build GRCh38
rs200699212 Chr.5: 146459089-146459089 on Build GRCh38
rs765657371 Chr.5: 146459033-146459034 on Build GRCh38
rs371433599 Chr.5: 146459324-146459324 on Build GRCh38
rs939447846 Chr.5: 146459060-146459060 on Build GRCh38
rs755697880 Chr.5: 146459053-146459060 on Build GRCh38
rs765010576 Chr.5: 146459117-146459118 on Build GRCh38
rs111965890 Chr.5: 146459096-146459096 on Build GRCh38
rs767666701 Chr.5: 146459292-146459292 on Build GRCh38
rs760752160 Chr.5: 146459305-146459305 on Build GRCh38
rs775964149 Chr.5: 146459046-146459051 on Build GRCh38
rs764402744 Chr.5: 146458988-146458988 on Build GRCh38
rs767168572 Chr.5: 146458938-146458938 on Build GRCh38
rs775097411 Chr.5: 146459062-146459062 on Build GRCh38
rs755241284 Chr.5: 146459099-146459099 on Build GRCh38
rs187719340 Chr.5: 146459230-146459230 on Build GRCh38
rs776974133 Chr.5: 146459078-146459078 on Build GRCh38
rs1056577399 Chr.5: 146459241-146459241 on Build GRCh38
rs757169617 Chr.5: 146459105-146459105 on Build GRCh38
rs779390603 Chr.5: 146459123-146459123 on Build GRCh38
rs749068903 Chr.5: 146458983-146458994 on Build GRCh38
rs200737262 Chr.5: 146458977-146458977 on Build GRCh38
rs990490844 Chr.5: 146459036-146459036 on Build GRCh38
rs541669774 Chr.5: 146459033-146459033 on Build GRCh38
rs770990778 Chr.5: 146459045-146459045 on Build GRCh38
rs557400659 Chr.5: 146459199-146459199 on Build GRCh38
rs148544834 Chr.5: 146459132-146459132 on Build GRCh38
rs757571272 Chr.5: 146459168-146459168 on Build GRCh38
rs778505919 Chr.5: 146459223-146459223 on Build GRCh38
rs778985502 Chr.5: 146458995-146459012 on Build GRCh38
rs762319496 Chr.5: 146459168-146459169 on Build GRCh38
rs111879925 Chr.5: 146459111-146459111 on Build GRCh38
rs752493267 Chr.5: 146459121-146459138 on Build GRCh38
rs879062152 Chr.5: 146459077-146459077 on Build GRCh38
rs766093356 Chr.5: 146459389-146459389 on Build GRCh38
rs769330148 Chr.5: 146459332-146459332 on Build GRCh38
rs545834199 Chr.5: 146459227-146459227 on Build GRCh38
rs754802778 Chr.5: 146459067-146459078 on Build GRCh38
rs751677150 Chr.5: 146459098-146459098 on Build GRCh38
rs757270908 Chr.5: 146459139-146459162 on Build GRCh38
rs753408349 Chr.5: 146459106-146459111 on Build GRCh38
rs774962544 Chr.5: 146459094-146459111 on Build GRCh38
rs746498610 Chr.5: 146459246-146459246 on Build GRCh38
rs928448336 Chr.5: 146459059-146459059 on Build GRCh38
rs183780634 Chr.5: 146459066-146459066 on Build GRCh38
rs914202664 Chr.5: 146459135-146459135 on Build GRCh38
rs780618123 Chr.5: 146459330-146459330 on Build GRCh38
rs747038272 Chr.5: 146459126-146459127 on Build GRCh38
rs747728427 Chr.5: 146459277-146459277 on Build GRCh38
rs555146294 Chr.5: 146459138-146459138 on Build GRCh38
rs916697712 Chr.5: 146459260-146459260 on Build GRCh38
rs537772467 Chr.5: 146459173-146459173 on Build GRCh38
rs754826945 Chr.5: 146459327-146459327 on Build GRCh38
rs759733366 Chr.5: 146459201-146459201 on Build GRCh38
rs760053839 Chr.5: 146459354-146459354 on Build GRCh38
rs546601229 Chr.5: 146459087-146459087 on Build GRCh38
rs780709817 Chr.5: 146459276-146459276 on Build GRCh38

View Full Product Details