Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40389319-40389799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOP2A
Entrez Gene ID
7153
Gene Name
topoisomerase (DNA) II alpha
Gene Aliases
TOP2, TP2A
UniGene
Hs.156346
Gene Chromosome Location
Chr.17: 40388521-40417950 on Build GRCh38
Transcripts
NM_001067.3 XM_005257632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3421525 c.4480G>T p.E1494* Chr.17: 40389635-40389635 on Build GRCh38
6146634 c.4476G>A p.K1492K Chr.17: 40389639-40389639 on Build GRCh38
5796640 c.4468-1G>A p.? Chr.17: 40389648-40389648 on Build GRCh38
6290928 c.4491C>T p.D1497D Chr.17: 40389624-40389624 on Build GRCh38
4642109 c.4541G>T p.R1514L Chr.17: 40389574-40389574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555554579 Chr.17: 40389350-40389350 on Build GRCh38
rs531771826 Chr.17: 40389731-40389731 on Build GRCh38
rs766846214 Chr.17: 40389617-40389617 on Build GRCh38
rs1033966251 Chr.17: 40389505-40389505 on Build GRCh38
rs761993788 Chr.17: 40389599-40389599 on Build GRCh38
rs754626880 Chr.17: 40389558-40389558 on Build GRCh38
rs750823979 Chr.17: 40389608-40389608 on Build GRCh38
rs573725979 Chr.17: 40389450-40389450 on Build GRCh38
rs372381834 Chr.17: 40389496-40389496 on Build GRCh38
rs756079860 Chr.17: 40389351-40389351 on Build GRCh38
rs757991657 Chr.17: 40389659-40389659 on Build GRCh38
rs561536016 Chr.17: 40389688-40389688 on Build GRCh38
rs767915206 Chr.17: 40389607-40389607 on Build GRCh38
rs549230260 Chr.17: 40389652-40389652 on Build GRCh38
rs745562008 Chr.17: 40389699-40389699 on Build GRCh38
rs370574626 Chr.17: 40389582-40389582 on Build GRCh38
rs1002466256 Chr.17: 40389491-40389491 on Build GRCh38
rs778744159 Chr.17: 40389614-40389614 on Build GRCh38
rs997760271 Chr.17: 40389544-40389544 on Build GRCh38
rs748817557 Chr.17: 40389542-40389542 on Build GRCh38
rs550692280 Chr.17: 40389762-40389762 on Build GRCh38
rs777570624 Chr.17: 40389424-40389424 on Build GRCh38
rs764316886 Chr.17: 40389475-40389475 on Build GRCh38
rs1021428865 Chr.17: 40389560-40389560 on Build GRCh38
rs935414104 Chr.17: 40389689-40389689 on Build GRCh38
rs775501165 Chr.17: 40389472-40389472 on Build GRCh38
rs375995857 Chr.17: 40389627-40389627 on Build GRCh38
rs1025161122 Chr.17: 40389442-40389446 on Build GRCh38
rs1046966936 Chr.17: 40389739-40389739 on Build GRCh38
rs1004761787 Chr.17: 40389436-40389436 on Build GRCh38
rs978635075 Chr.17: 40389640-40389640 on Build GRCh38
rs746836240 Chr.17: 40389694-40389694 on Build GRCh38
rs368592045 Chr.17: 40389414-40389414 on Build GRCh38
rs774528941 Chr.17: 40389597-40389597 on Build GRCh38
rs375623549 Chr.17: 40389575-40389575 on Build GRCh38
rs750923941 Chr.17: 40389515-40389515 on Build GRCh38
rs756643129 Chr.17: 40389531-40389531 on Build GRCh38
rs746962457 Chr.17: 40389578-40389578 on Build GRCh38
rs915739180 Chr.17: 40389356-40389356 on Build GRCh38
rs184329132 Chr.17: 40389473-40389473 on Build GRCh38
rs770779298 Chr.17: 40389580-40389580 on Build GRCh38
rs111733098 Chr.17: 40389620-40389620 on Build GRCh38
rs11540720 Chr.17: 40389572-40389572 on Build GRCh38
rs949834288 Chr.17: 40389691-40389691 on Build GRCh38
rs767816690 Chr.17: 40389513-40389513 on Build GRCh38
rs972248234 Chr.17: 40389602-40389602 on Build GRCh38
rs901867454 Chr.17: 40389527-40389527 on Build GRCh38
rs866510555 Chr.17: 40389587-40389587 on Build GRCh38
rs754498405 Chr.17: 40389629-40389629 on Build GRCh38
rs1047406616 Chr.17: 40389645-40389645 on Build GRCh38
rs947190282 Chr.17: 40389638-40389638 on Build GRCh38
rs924461240 Chr.17: 40389654-40389654 on Build GRCh38
rs544998359 Chr.17: 40389574-40389574 on Build GRCh38
rs778529043 Chr.17: 40389586-40389587 on Build GRCh38
rs959735528 Chr.17: 40389619-40389619 on Build GRCh38
rs560441128 Chr.17: 40389589-40389589 on Build GRCh38
rs779546795 Chr.17: 40389532-40389532 on Build GRCh38
rs75106179 Chr.17: 40389773-40389773 on Build GRCh38
rs753742643 Chr.17: 40389509-40389509 on Build GRCh38
rs201054615 Chr.17: 40389634-40389634 on Build GRCh38
rs764121407 Chr.17: 40389455-40389455 on Build GRCh38

View Full Product Details