Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32451164-32451699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
UniGene
Hs.585464
Gene Chromosome Location
Chr.10: 32446082-32882864 on Build GRCh38
Transcripts
NM_001026383.2 NM_001321115.1 NM_145023.5 NR_109826.1 NR_109827.1 XM_006717505.1 XM_006717506.1 XM_011519663.1 XM_011519664.1 XM_011519666.1 XM_011519667.1 XM_011519668.1 XM_011519669.1 XM_011519670.1 XM_011519671.1 XM_011519672.1 XM_011519673.1 XM_011519674.1 XM_011519675.1 XM_011519676.1 XM_011519677.1 XM_011519678.1 XM_011519679.1 XM_011519684.2 XM_011519686.2 XM_011519688.2 XM_017016638.1 XM_017016639.1 XM_017016640.1 XM_017016644.1 XM_017016647.1 XM_017016650.1 XM_017016651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622389 c.47A>G p.N16S Chr.10: 32451689-32451689 on Build GRCh38
4573587 c.41C>T p.S14L Chr.10: 32451683-32451683 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888504026 Chr.10: 32451323-32451323 on Build GRCh38
rs747900923 Chr.10: 32451421-32451421 on Build GRCh38
rs922785487 Chr.10: 32451274-32451274 on Build GRCh38
rs994853579 Chr.10: 32451429-32451429 on Build GRCh38
rs374511671 Chr.10: 32451613-32451613 on Build GRCh38
rs986544254 Chr.10: 32451192-32451192 on Build GRCh38
rs945993095 Chr.10: 32451244-32451244 on Build GRCh38
rs955595003 Chr.10: 32451450-32451450 on Build GRCh38
rs544742751 Chr.10: 32451598-32451598 on Build GRCh38
rs897193470 Chr.10: 32451349-32451351 on Build GRCh38
rs1041616825 Chr.10: 32451283-32451283 on Build GRCh38
rs986930253 Chr.10: 32451451-32451451 on Build GRCh38
rs747971791 Chr.10: 32451596-32451596 on Build GRCh38
rs138558761 Chr.10: 32451581-32451581 on Build GRCh38
rs942412309 Chr.10: 32451216-32451216 on Build GRCh38
rs907063435 Chr.10: 32451299-32451299 on Build GRCh38
rs567893423 Chr.10: 32451646-32451646 on Build GRCh38
rs769716566 Chr.10: 32451457-32451457 on Build GRCh38
rs531497310 Chr.10: 32451448-32451448 on Build GRCh38
rs945719248 Chr.10: 32451574-32451574 on Build GRCh38
rs910076669 Chr.10: 32451603-32451603 on Build GRCh38
rs747295658 Chr.10: 32451228-32451228 on Build GRCh38
rs186178595 Chr.10: 32451592-32451592 on Build GRCh38
rs77970650 Chr.10: 32451608-32451608 on Build GRCh38
rs911355333 Chr.10: 32451452-32451452 on Build GRCh38
rs11008943 Chr.10: 32451530-32451530 on Build GRCh38
rs202207315 Chr.10: 32451663-32451663 on Build GRCh38
rs1036057581 Chr.10: 32451417-32451417 on Build GRCh38
rs1025884065 Chr.10: 32451437-32451437 on Build GRCh38
rs894305292 Chr.10: 32451589-32451589 on Build GRCh38
rs779659559 Chr.10: 32451678-32451678 on Build GRCh38
rs938540635 Chr.10: 32451582-32451582 on Build GRCh38
rs1041746443 Chr.10: 32451579-32451579 on Build GRCh38
rs758154744 Chr.10: 32451675-32451675 on Build GRCh38
rs61735137 Chr.10: 32451619-32451619 on Build GRCh38
rs910985451 Chr.10: 32451199-32451199 on Build GRCh38
rs768312073 Chr.10: 32451363-32451363 on Build GRCh38
rs963794674 Chr.10: 32451499-32451499 on Build GRCh38
rs549308577 Chr.10: 32451623-32451623 on Build GRCh38
rs750068430 Chr.10: 32451668-32451668 on Build GRCh38
rs1031327687 Chr.10: 32451463-32451463 on Build GRCh38
rs773462288 Chr.10: 32451604-32451604 on Build GRCh38
rs773155816 Chr.10: 32451525-32451525 on Build GRCh38
rs1002719935 Chr.10: 32451324-32451324 on Build GRCh38
rs1012304052 Chr.10: 32451370-32451370 on Build GRCh38
rs978530918 Chr.10: 32451453-32451453 on Build GRCh38
rs761083569 Chr.10: 32451639-32451639 on Build GRCh38
rs532116292 Chr.10: 32451588-32451588 on Build GRCh38
rs1054534975 Chr.10: 32451223-32451223 on Build GRCh38
rs1036975691 Chr.10: 32451340-32451340 on Build GRCh38
rs1034161579 Chr.10: 32451337-32451337 on Build GRCh38
rs955931862 Chr.10: 32451494-32451494 on Build GRCh38
rs1049935585 Chr.10: 32451318-32451318 on Build GRCh38
rs1017384040 Chr.10: 32451420-32451420 on Build GRCh38
rs532629742 Chr.10: 32451364-32451364 on Build GRCh38

View Full Product Details