Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 78006662-78007236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAV3
Entrez Gene ID
89795
Gene Name
neuron navigator 3
Gene Aliases
POMFIL1, STEERIN3, unc53H3
UniGene
Hs.655301
Gene Chromosome Location
Chr.12: 77571862-78213012 on Build GRCh38
Transcripts
NM_001024383.1 NM_014903.5 XM_005269215.3 XM_011538940.2 XM_011538941.2 XM_011538944.2 XM_017020164.1 XM_017020165.1 XM_017020166.1 XM_017020167.1 XM_017020168.1 XM_017020169.1 XM_017020170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522837 c.1182G>A p.R394R Chr.12: 78006720-78006720 on Build GRCh38
325909 c.1167G>C p.K389N Chr.12: 78006705-78006705 on Build GRCh38
4910711 c.1177G>A p.A393T Chr.12: 78006715-78006715 on Build GRCh38
1992428 c.1698T>G p.S566S Chr.12: 78007236-78007236 on Build GRCh38
4757899 c.1192C>T p.R398C Chr.12: 78006730-78006730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748494676 Chr.12: 78007019-78007019 on Build GRCh38
rs114303302 Chr.12: 78006786-78006786 on Build GRCh38
rs774279306 Chr.12: 78007047-78007047 on Build GRCh38
rs370224002 Chr.12: 78006737-78006737 on Build GRCh38
rs182114538 Chr.12: 78006736-78006736 on Build GRCh38
rs766399842 Chr.12: 78007083-78007083 on Build GRCh38
rs534710627 Chr.12: 78007108-78007108 on Build GRCh38
rs746334714 Chr.12: 78007191-78007191 on Build GRCh38
rs762329198 Chr.12: 78006946-78006946 on Build GRCh38
rs775648848 Chr.12: 78006775-78006777 on Build GRCh38
rs758212678 Chr.12: 78007080-78007080 on Build GRCh38
rs189238808 Chr.12: 78006733-78006733 on Build GRCh38
rs780975122 Chr.12: 78006689-78006689 on Build GRCh38
rs763299667 Chr.12: 78007029-78007029 on Build GRCh38
rs1027711441 Chr.12: 78007138-78007138 on Build GRCh38
rs770375734 Chr.12: 78006718-78006718 on Build GRCh38
rs766325464 Chr.12: 78006959-78006959 on Build GRCh38
rs376504786 Chr.12: 78007165-78007165 on Build GRCh38
rs796852973 Chr.12: 78006884-78006884 on Build GRCh38
rs746544935 Chr.12: 78007005-78007005 on Build GRCh38
rs746603031 Chr.12: 78006920-78006920 on Build GRCh38
rs755907283 Chr.12: 78006763-78006763 on Build GRCh38
rs747641848 Chr.12: 78006759-78006759 on Build GRCh38
rs777476104 Chr.12: 78006870-78006870 on Build GRCh38
rs770856490 Chr.12: 78006797-78006797 on Build GRCh38
rs773789932 Chr.12: 78006812-78006812 on Build GRCh38
rs201452320 Chr.12: 78007143-78007143 on Build GRCh38
rs748769258 Chr.12: 78006716-78006716 on Build GRCh38
rs369117734 Chr.12: 78006690-78006690 on Build GRCh38
rs915656076 Chr.12: 78006917-78006917 on Build GRCh38
rs373671296 Chr.12: 78006885-78006885 on Build GRCh38
rs760909219 Chr.12: 78007065-78007065 on Build GRCh38
rs1027742467 Chr.12: 78006787-78006787 on Build GRCh38
rs750238403 Chr.12: 78007071-78007071 on Build GRCh38
rs529588724 Chr.12: 78007085-78007085 on Build GRCh38
rs200389736 Chr.12: 78006835-78006835 on Build GRCh38
rs370427296 Chr.12: 78007040-78007040 on Build GRCh38
rs779639003 Chr.12: 78006908-78006908 on Build GRCh38
rs751781706 Chr.12: 78006864-78006864 on Build GRCh38
rs777167259 Chr.12: 78006833-78006833 on Build GRCh38
rs767782554 Chr.12: 78006988-78006988 on Build GRCh38
rs527791398 Chr.12: 78006952-78006952 on Build GRCh38
rs772871173 Chr.12: 78007127-78007127 on Build GRCh38
rs549639857 Chr.12: 78007100-78007100 on Build GRCh38
rs775044959 Chr.12: 78006820-78006820 on Build GRCh38
rs755256058 Chr.12: 78006865-78006865 on Build GRCh38
rs770422527 Chr.12: 78006951-78006951 on Build GRCh38
rs766198467 Chr.12: 78007131-78007131 on Build GRCh38
rs766596287 Chr.12: 78006858-78006858 on Build GRCh38
rs557857297 Chr.12: 78007110-78007110 on Build GRCh38
rs754639465 Chr.12: 78006731-78006731 on Build GRCh38
rs376072848 Chr.12: 78006890-78006890 on Build GRCh38
rs754578095 Chr.12: 78007090-78007090 on Build GRCh38
rs777564080 Chr.12: 78006785-78006785 on Build GRCh38
rs879033793 Chr.12: 78007156-78007156 on Build GRCh38
rs202050601 Chr.12: 78006738-78006738 on Build GRCh38
rs559117211 Chr.12: 78006876-78006876 on Build GRCh38
rs773093683 Chr.12: 78006707-78006707 on Build GRCh38
rs779166824 Chr.12: 78007188-78007188 on Build GRCh38
rs1045110801 Chr.12: 78006729-78006729 on Build GRCh38
rs184775230 Chr.12: 78006719-78006719 on Build GRCh38
rs541081657 Chr.12: 78006977-78006977 on Build GRCh38
rs961283779 Chr.12: 78007198-78007198 on Build GRCh38
rs34276383 Chr.12: 78007104-78007104 on Build GRCh38
rs375778220 Chr.12: 78006730-78006730 on Build GRCh38
rs769759722 Chr.12: 78006699-78006699 on Build GRCh38
rs763070116 Chr.12: 78006957-78006957 on Build GRCh38
rs750520245 Chr.12: 78006852-78006852 on Build GRCh38
rs775577911 Chr.12: 78007113-78007113 on Build GRCh38
rs200464108 Chr.12: 78007111-78007111 on Build GRCh38
rs757081545 Chr.12: 78006996-78006996 on Build GRCh38
rs778139901 Chr.12: 78006911-78006911 on Build GRCh38
rs755811434 Chr.12: 78007170-78007170 on Build GRCh38
rs771335214 Chr.12: 78007109-78007109 on Build GRCh38
rs779364938 Chr.12: 78007003-78007003 on Build GRCh38
rs760821087 Chr.12: 78007115-78007115 on Build GRCh38
rs61754793 Chr.12: 78007067-78007067 on Build GRCh38
rs541972974 Chr.12: 78007123-78007123 on Build GRCh38
rs778799852 Chr.12: 78006999-78006999 on Build GRCh38
rs35015855 Chr.12: 78006885-78006885 on Build GRCh38
rs1035276571 Chr.12: 78007181-78007181 on Build GRCh38
rs771531149 Chr.12: 78006902-78006902 on Build GRCh38
rs757687933 Chr.12: 78007179-78007179 on Build GRCh38
rs201438479 Chr.12: 78006734-78006734 on Build GRCh38
rs758726790 Chr.12: 78006688-78006688 on Build GRCh38
rs34682552 Chr.12: 78006715-78006716 on Build GRCh38
rs781061916 Chr.12: 78007018-78007018 on Build GRCh38
rs751658778 Chr.12: 78006965-78006965 on Build GRCh38
rs768360774 Chr.12: 78006923-78006923 on Build GRCh38
rs761280532 Chr.12: 78007152-78007153 on Build GRCh38
rs765854258 Chr.12: 78006850-78006850 on Build GRCh38
rs780201858 Chr.12: 78006755-78006755 on Build GRCh38
rs928235913 Chr.12: 78006857-78006857 on Build GRCh38
rs533952691 Chr.12: 78007159-78007159 on Build GRCh38
rs772313322 Chr.12: 78006731-78006732 on Build GRCh38
rs761077508 Chr.12: 78006821-78006821 on Build GRCh38
rs372042441 Chr.12: 78007212-78007212 on Build GRCh38
rs904358931 Chr.12: 78006723-78006723 on Build GRCh38
rs778887459 Chr.12: 78006883-78006883 on Build GRCh38
rs556939248 Chr.12: 78006747-78006747 on Build GRCh38
rs753624120 Chr.12: 78006990-78006990 on Build GRCh38
rs34422147 Chr.12: 78006978-78006979 on Build GRCh38
rs201405730 Chr.12: 78007176-78007176 on Build GRCh38
rs371746674 Chr.12: 78007125-78007125 on Build GRCh38
rs754991804 Chr.12: 78007154-78007154 on Build GRCh38
rs747975674 Chr.12: 78006760-78006760 on Build GRCh38
rs777079303 Chr.12: 78006929-78006929 on Build GRCh38
rs775907789 Chr.12: 78006732-78006732 on Build GRCh38
rs772572462 Chr.12: 78007201-78007201 on Build GRCh38
rs758592180 Chr.12: 78006749-78006749 on Build GRCh38
rs540019767 Chr.12: 78007062-78007062 on Build GRCh38
rs746110023 Chr.12: 78007202-78007202 on Build GRCh38
rs200981782 Chr.12: 78007052-78007052 on Build GRCh38
rs370063853 Chr.12: 78006834-78006834 on Build GRCh38
rs200010458 Chr.12: 78006823-78006823 on Build GRCh38
rs768411330 Chr.12: 78007014-78007014 on Build GRCh38
rs781577801 Chr.12: 78007175-78007175 on Build GRCh38
rs770336478 Chr.12: 78007021-78007021 on Build GRCh38
rs370324426 Chr.12: 78007103-78007103 on Build GRCh38
rs780974001 Chr.12: 78007091-78007091 on Build GRCh38
rs79537918 Chr.12: 78007022-78007022 on Build GRCh38
rs1039410388 Chr.12: 78007072-78007072 on Build GRCh38
rs763012068 Chr.12: 78006855-78006855 on Build GRCh38
rs909661884 Chr.12: 78007049-78007049 on Build GRCh38
rs762874130 Chr.12: 78007128-78007128 on Build GRCh38
rs564529583 Chr.12: 78007002-78007002 on Build GRCh38

View Full Product Details