Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30921258-30921822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEDAG
Entrez Gene ID
84935
Gene Name
mesenteric estrogen dependent adipogenesis
Gene Aliases
AWMS3, C13orf33, MEDA-4, MEDA4, hAWMS3
UniGene
Hs.147880
Gene Chromosome Location
Chr.13: 30906175-30925577 on Build GRCh38
Transcripts
NM_032849.3 XM_017020801.1 NR_038287.1

Gene Symbol
TEX26-AS1
Entrez Gene ID
100507064
Gene Name
TEX26 antisense RNA 1
Gene Aliases
LINC00447
UniGene
Hs.654878
Gene Chromosome Location
Chr.13: 30882835-30932608 on Build GRCh38
Transcripts
NM_032849.3 XM_017020801.1 NR_038287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6711038 c.540A>C p.P180P Chr.13: 30921599-30921599 on Build GRCh38
3468276 c.741C>T p.I247I Chr.13: 30921800-30921800 on Build GRCh38
6711039 c.763C>A p.R255R Chr.13: 30921822-30921822 on Build GRCh38
279270 c.525G>T p.K175N Chr.13: 30921584-30921584 on Build GRCh38
3688672 c.718G>A p.E240K Chr.13: 30921777-30921777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781576303 Chr.13: 30921463-30921463 on Build GRCh38
rs753872842 Chr.13: 30921777-30921778 on Build GRCh38
rs780585256 Chr.13: 30921760-30921760 on Build GRCh38
rs762883125 Chr.13: 30921700-30921700 on Build GRCh38
rs536772013 Chr.13: 30921770-30921770 on Build GRCh38
rs923905314 Chr.13: 30921332-30921332 on Build GRCh38
rs373899952 Chr.13: 30921533-30921533 on Build GRCh38
rs149352249 Chr.13: 30921699-30921699 on Build GRCh38
rs530927304 Chr.13: 30921487-30921487 on Build GRCh38
rs745329303 Chr.13: 30921632-30921632 on Build GRCh38
rs767135524 Chr.13: 30921735-30921735 on Build GRCh38
rs775385301 Chr.13: 30921613-30921613 on Build GRCh38
rs771305307 Chr.13: 30921605-30921606 on Build GRCh38
rs773177464 Chr.13: 30921693-30921693 on Build GRCh38
rs138712634 Chr.13: 30921449-30921449 on Build GRCh38
rs895167670 Chr.13: 30921556-30921556 on Build GRCh38
rs748486713 Chr.13: 30921785-30921785 on Build GRCh38
rs147350400 Chr.13: 30921697-30921697 on Build GRCh38
rs963945772 Chr.13: 30921764-30921764 on Build GRCh38
rs765200612 Chr.13: 30921595-30921595 on Build GRCh38
rs763890240 Chr.13: 30921599-30921599 on Build GRCh38
rs772250763 Chr.13: 30921792-30921792 on Build GRCh38
rs142828800 Chr.13: 30921799-30921799 on Build GRCh38
rs974214495 Chr.13: 30921795-30921795 on Build GRCh38
rs576074999 Chr.13: 30921745-30921745 on Build GRCh38
rs767463116 Chr.13: 30921543-30921543 on Build GRCh38
rs749965380 Chr.13: 30921736-30921736 on Build GRCh38
rs929686324 Chr.13: 30921508-30921508 on Build GRCh38
rs561395600 Chr.13: 30921473-30921473 on Build GRCh38
rs943025022 Chr.13: 30921730-30921730 on Build GRCh38
rs150178788 Chr.13: 30921676-30921676 on Build GRCh38
rs986609971 Chr.13: 30921400-30921400 on Build GRCh38
rs968111645 Chr.13: 30921301-30921301 on Build GRCh38
rs778054883 Chr.13: 30921560-30921560 on Build GRCh38
rs544262753 Chr.13: 30921464-30921464 on Build GRCh38
rs774540988 Chr.13: 30921537-30921537 on Build GRCh38
rs773303281 Chr.13: 30921561-30921561 on Build GRCh38
rs1025802415 Chr.13: 30921662-30921662 on Build GRCh38
rs138341583 Chr.13: 30921774-30921774 on Build GRCh38
rs756580451 Chr.13: 30921636-30921636 on Build GRCh38
rs1048792380 Chr.13: 30921337-30921337 on Build GRCh38
rs749708186 Chr.13: 30921642-30921642 on Build GRCh38
rs763319508 Chr.13: 30921553-30921556 on Build GRCh38
rs374109967 Chr.13: 30921703-30921703 on Build GRCh38
rs79895157 Chr.13: 30921638-30921638 on Build GRCh38
rs911052266 Chr.13: 30921762-30921762 on Build GRCh38
rs756895959 Chr.13: 30921617-30921617 on Build GRCh38
rs774594489 Chr.13: 30921671-30921671 on Build GRCh38
rs756349736 Chr.13: 30921488-30921488 on Build GRCh38
rs997761776 Chr.13: 30921436-30921436 on Build GRCh38
rs759414279 Chr.13: 30921562-30921562 on Build GRCh38
rs758256977 Chr.13: 30921597-30921597 on Build GRCh38
rs377025280 Chr.13: 30921598-30921598 on Build GRCh38
rs1056651969 Chr.13: 30921563-30921563 on Build GRCh38
rs369962770 Chr.13: 30921405-30921405 on Build GRCh38
rs931579059 Chr.13: 30921287-30921287 on Build GRCh38
rs375543579 Chr.13: 30921761-30921761 on Build GRCh38
rs768983578 Chr.13: 30921651-30921651 on Build GRCh38
rs780594359 Chr.13: 30921618-30921618 on Build GRCh38
rs755436482 Chr.13: 30921767-30921767 on Build GRCh38
rs535179514 Chr.13: 30921602-30921602 on Build GRCh38
rs549429506 Chr.13: 30921489-30921489 on Build GRCh38
rs868844137 Chr.13: 30921776-30921777 on Build GRCh38
rs756878254 Chr.13: 30921712-30921712 on Build GRCh38
rs760523541 Chr.13: 30921750-30921752 on Build GRCh38
rs761999133 Chr.13: 30921540-30921540 on Build GRCh38
rs955299409 Chr.13: 30921756-30921756 on Build GRCh38
rs978402816 Chr.13: 30921315-30921315 on Build GRCh38
rs763880836 Chr.13: 30921763-30921767 on Build GRCh38
rs933079763 Chr.13: 30921791-30921791 on Build GRCh38
rs754308782 Chr.13: 30921763-30921763 on Build GRCh38
rs371069336 Chr.13: 30921692-30921692 on Build GRCh38
rs370922173 Chr.13: 30921521-30921521 on Build GRCh38
rs773539240 Chr.13: 30921520-30921520 on Build GRCh38
rs6563571 Chr.13: 30921596-30921596 on Build GRCh38
rs41433250 Chr.13: 30921698-30921698 on Build GRCh38
rs749583539 Chr.13: 30921523-30921523 on Build GRCh38
rs901559538 Chr.13: 30921411-30921411 on Build GRCh38
rs1038816318 Chr.13: 30921398-30921398 on Build GRCh38
rs1012172219 Chr.13: 30921559-30921559 on Build GRCh38
rs751322843 Chr.13: 30921709-30921709 on Build GRCh38
rs75477724 Chr.13: 30921344-30921344 on Build GRCh38
rs113320120 Chr.13: 30921385-30921385 on Build GRCh38

View Full Product Details