Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 24749141-24749733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFNA5
Entrez Gene ID
1687
Gene Name
DFNA5, deafness associated tumor suppressor
Gene Aliases
ICERE-1
UniGene
Hs.520708
Gene Chromosome Location
Chr.7: 24698355-24758020 on Build GRCh38
Transcripts
NM_001127453.1 NM_001127454.1 NM_004403.2 XM_017011802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5979329 c.87T>A p.D29E Chr.7: 24749688-24749688 on Build GRCh38
7364177 c.143G>A p.R48K Chr.7: 24749632-24749632 on Build GRCh38
1088701 c.55C>A p.L19M Chr.7: 24749720-24749720 on Build GRCh38
1450110 c.53A>T p.D18V Chr.7: 24749722-24749722 on Build GRCh38
6642772 c.196C>T p.Q66* Chr.7: 24749579-24749579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975807793 Chr.7: 24749226-24749226 on Build GRCh38
rs369867003 Chr.7: 24749560-24749560 on Build GRCh38
rs761407115 Chr.7: 24749587-24749587 on Build GRCh38
rs556969465 Chr.7: 24749481-24749481 on Build GRCh38
rs574436799 Chr.7: 24749458-24749458 on Build GRCh38
rs958010880 Chr.7: 24749712-24749712 on Build GRCh38
rs774533771 Chr.7: 24749515-24749515 on Build GRCh38
rs552240970 Chr.7: 24749185-24749185 on Build GRCh38
rs879137248 Chr.7: 24749497-24749497 on Build GRCh38
rs141734181 Chr.7: 24749207-24749209 on Build GRCh38
rs28490522 Chr.7: 24749249-24749249 on Build GRCh38
rs778020680 Chr.7: 24749636-24749636 on Build GRCh38
rs776319567 Chr.7: 24749539-24749539 on Build GRCh38
rs867578316 Chr.7: 24749176-24749176 on Build GRCh38
rs183252587 Chr.7: 24749491-24749491 on Build GRCh38
rs1046110601 Chr.7: 24749345-24749345 on Build GRCh38
rs143403124 Chr.7: 24749661-24749661 on Build GRCh38
rs539777831 Chr.7: 24749498-24749501 on Build GRCh38
rs906321194 Chr.7: 24749257-24749257 on Build GRCh38
rs745646987 Chr.7: 24749653-24749653 on Build GRCh38
rs989654790 Chr.7: 24749474-24749474 on Build GRCh38
rs910360160 Chr.7: 24749438-24749438 on Build GRCh38
rs763405328 Chr.7: 24749329-24749329 on Build GRCh38
rs936916914 Chr.7: 24749246-24749246 on Build GRCh38
rs776150418 Chr.7: 24749220-24749220 on Build GRCh38
rs566393085 Chr.7: 24749501-24749502 on Build GRCh38
rs570974581 Chr.7: 24749320-24749320 on Build GRCh38
rs755412241 Chr.7: 24749516-24749516 on Build GRCh38
rs982825000 Chr.7: 24749178-24749178 on Build GRCh38
rs201189985 Chr.7: 24749691-24749691 on Build GRCh38
rs779520314 Chr.7: 24749525-24749525 on Build GRCh38
rs748682263 Chr.7: 24749651-24749653 on Build GRCh38
rs149920782 Chr.7: 24749431-24749431 on Build GRCh38
rs369689648 Chr.7: 24749572-24749572 on Build GRCh38
rs1003033028 Chr.7: 24749260-24749260 on Build GRCh38
rs747192368 Chr.7: 24749637-24749637 on Build GRCh38
rs779216699 Chr.7: 24749192-24749192 on Build GRCh38
rs773824667 Chr.7: 24749697-24749697 on Build GRCh38
rs759414118 Chr.7: 24749559-24749559 on Build GRCh38
rs754333504 Chr.7: 24749513-24749513 on Build GRCh38
rs143116304 Chr.7: 24749582-24749582 on Build GRCh38
rs779556514 Chr.7: 24749600-24749600 on Build GRCh38
rs751858148 Chr.7: 24749375-24749375 on Build GRCh38
rs760212631 Chr.7: 24749182-24749182 on Build GRCh38
rs982224778 Chr.7: 24749485-24749485 on Build GRCh38
rs965378106 Chr.7: 24749218-24749218 on Build GRCh38
rs767755008 Chr.7: 24749595-24749595 on Build GRCh38
rs1047680251 Chr.7: 24749403-24749403 on Build GRCh38
rs1054900675 Chr.7: 24749545-24749545 on Build GRCh38
rs772979763 Chr.7: 24749711-24749711 on Build GRCh38
rs886249757 Chr.7: 24749424-24749424 on Build GRCh38
rs1028734668 Chr.7: 24749459-24749459 on Build GRCh38
rs1008843427 Chr.7: 24749663-24749663 on Build GRCh38
rs190587382 Chr.7: 24749227-24749227 on Build GRCh38
rs758488919 Chr.7: 24749655-24749656 on Build GRCh38
rs199834410 Chr.7: 24749528-24749528 on Build GRCh38
rs1025105164 Chr.7: 24749256-24749256 on Build GRCh38
rs146697813 Chr.7: 24749596-24749596 on Build GRCh38
rs140311101 Chr.7: 24749619-24749619 on Build GRCh38
rs552820955 Chr.7: 24749245-24749245 on Build GRCh38
rs1055195067 Chr.7: 24749372-24749372 on Build GRCh38
rs985907465 Chr.7: 24749437-24749437 on Build GRCh38
rs914040622 Chr.7: 24749323-24749323 on Build GRCh38
rs77931296 Chr.7: 24749363-24749363 on Build GRCh38
rs1050262884 Chr.7: 24749180-24749180 on Build GRCh38
rs574727291 Chr.7: 24749473-24749473 on Build GRCh38
rs773853744 Chr.7: 24749656-24749659 on Build GRCh38
rs930313983 Chr.7: 24749571-24749571 on Build GRCh38
rs148370267 Chr.7: 24749688-24749688 on Build GRCh38
rs763609133 Chr.7: 24749577-24749577 on Build GRCh38
rs994721052 Chr.7: 24749392-24749392 on Build GRCh38
rs66692319 Chr.7: 24749513-24749514 on Build GRCh38
rs753257338 Chr.7: 24749630-24749630 on Build GRCh38
rs767060533 Chr.7: 24749700-24749700 on Build GRCh38
rs370058550 Chr.7: 24749597-24749597 on Build GRCh38
rs770710188 Chr.7: 24749537-24749537 on Build GRCh38
rs920214893 Chr.7: 24749553-24749553 on Build GRCh38
rs1054887275 Chr.7: 24749547-24749547 on Build GRCh38
rs188664289 Chr.7: 24749632-24749632 on Build GRCh38
rs747247464 Chr.7: 24749535-24749535 on Build GRCh38
rs771026695 Chr.7: 24749642-24749642 on Build GRCh38
rs989725227 Chr.7: 24749267-24749267 on Build GRCh38
rs142211483 Chr.7: 24749529-24749529 on Build GRCh38
rs775475583 Chr.7: 24749564-24749564 on Build GRCh38
rs144996416 Chr.7: 24749447-24749447 on Build GRCh38
rs553611139 Chr.7: 24749501-24749501 on Build GRCh38
rs1040609324 Chr.7: 24749676-24749676 on Build GRCh38
rs927718317 Chr.7: 24749361-24749361 on Build GRCh38
rs552464750 Chr.7: 24749310-24749310 on Build GRCh38
rs1008748499 Chr.7: 24749425-24749425 on Build GRCh38
rs770893793 Chr.7: 24749673-24749674 on Build GRCh38
rs768391255 Chr.7: 24749497-24749498 on Build GRCh38
rs149188029 Chr.7: 24749643-24749643 on Build GRCh38
rs2521770 Chr.7: 24749463-24749463 on Build GRCh38
rs958927073 Chr.7: 24749268-24749268 on Build GRCh38
rs775350845 Chr.7: 24749685-24749685 on Build GRCh38
rs760895924 Chr.7: 24749656-24749656 on Build GRCh38
rs914175455 Chr.7: 24749479-24749479 on Build GRCh38

View Full Product Details