Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162008445-162008861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP4
Entrez Gene ID
1803
Gene Name
dipeptidyl peptidase 4
Gene Aliases
ADABP, ADCP2, CD26, DPPIV, TP103
UniGene
Hs.368912
Gene Chromosome Location
Chr.2: 161992241-162074542 on Build GRCh38
Transcripts
NM_001935.3 XM_005246371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707821 c.1957G>A p.V653M Chr.2: 162008592-162008592 on Build GRCh38
7221283 c.1935C>T p.G645G Chr.2: 162008614-162008614 on Build GRCh38
329327 c.1903G>A p.V635I Chr.2: 162008646-162008646 on Build GRCh38
4634624 c.1972C>T p.R658W Chr.2: 162008577-162008577 on Build GRCh38
3568488 c.1928G>A p.G643E Chr.2: 162008621-162008621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768490740 Chr.2: 162008543-162008543 on Build GRCh38
rs17848910 Chr.2: 162008614-162008614 on Build GRCh38
rs200431129 Chr.2: 162008650-162008650 on Build GRCh38
rs780928012 Chr.2: 162008601-162008601 on Build GRCh38
rs777468645 Chr.2: 162008613-162008613 on Build GRCh38
rs117796258 Chr.2: 162008631-162008631 on Build GRCh38
rs761908679 Chr.2: 162008548-162008548 on Build GRCh38
rs991661719 Chr.2: 162008659-162008659 on Build GRCh38
rs199755074 Chr.2: 162008607-162008607 on Build GRCh38
rs747045710 Chr.2: 162008706-162008706 on Build GRCh38
rs769609373 Chr.2: 162008603-162008603 on Build GRCh38
rs751172709 Chr.2: 162008592-162008592 on Build GRCh38
rs759017895 Chr.2: 162008589-162008589 on Build GRCh38
rs200164809 Chr.2: 162008666-162008666 on Build GRCh38
rs765615881 Chr.2: 162008587-162008587 on Build GRCh38
rs981791533 Chr.2: 162008596-162008596 on Build GRCh38
rs754080853 Chr.2: 162008578-162008578 on Build GRCh38
rs567095078 Chr.2: 162008518-162008518 on Build GRCh38
rs765607849 Chr.2: 162008642-162008642 on Build GRCh38
rs941280504 Chr.2: 162008819-162008819 on Build GRCh38
rs138430103 Chr.2: 162008576-162008576 on Build GRCh38
rs779763529 Chr.2: 162008529-162008529 on Build GRCh38
rs199691557 Chr.2: 162008563-162008563 on Build GRCh38
rs773220934 Chr.2: 162008557-162008557 on Build GRCh38
rs200944234 Chr.2: 162008647-162008647 on Build GRCh38
rs201024370 Chr.2: 162008602-162008602 on Build GRCh38
rs761993733 Chr.2: 162008633-162008633 on Build GRCh38
rs371412252 Chr.2: 162008639-162008639 on Build GRCh38
rs775610508 Chr.2: 162008575-162008575 on Build GRCh38
rs202156445 Chr.2: 162008528-162008528 on Build GRCh38
rs1031456284 Chr.2: 162008510-162008510 on Build GRCh38
rs747990126 Chr.2: 162008546-162008546 on Build GRCh38
rs202114910 Chr.2: 162008648-162008648 on Build GRCh38
rs770786621 Chr.2: 162008699-162008699 on Build GRCh38
rs999532414 Chr.2: 162008495-162008495 on Build GRCh38
rs535564894 Chr.2: 162008688-162008688 on Build GRCh38
rs200839327 Chr.2: 162008624-162008624 on Build GRCh38
rs779706584 Chr.2: 162008590-162008590 on Build GRCh38
rs778850699 Chr.2: 162008705-162008705 on Build GRCh38
rs561284391 Chr.2: 162008782-162008782 on Build GRCh38
rs149643982 Chr.2: 162008588-162008588 on Build GRCh38
rs201712012 Chr.2: 162008617-162008617 on Build GRCh38
rs751048103 Chr.2: 162008651-162008651 on Build GRCh38
rs755788492 Chr.2: 162008670-162008670 on Build GRCh38
rs949926358 Chr.2: 162008720-162008720 on Build GRCh38
rs552025572 Chr.2: 162008497-162008497 on Build GRCh38
rs143706793 Chr.2: 162008530-162008530 on Build GRCh38
rs754620953 Chr.2: 162008593-162008593 on Build GRCh38
rs769577567 Chr.2: 162008552-162008552 on Build GRCh38
rs2268891 Chr.2: 162008558-162008558 on Build GRCh38
rs765288567 Chr.2: 162008768-162008768 on Build GRCh38
rs886990518 Chr.2: 162008500-162008500 on Build GRCh38
rs764311023 Chr.2: 162008577-162008577 on Build GRCh38
rs754557228 Chr.2: 162008652-162008652 on Build GRCh38
rs1023848443 Chr.2: 162008823-162008823 on Build GRCh38
rs776278630 Chr.2: 162008545-162008545 on Build GRCh38
rs757673793 Chr.2: 162008579-162008579 on Build GRCh38
rs41268649 Chr.2: 162008623-162008623 on Build GRCh38
rs758009538 Chr.2: 162008527-162008527 on Build GRCh38
rs200539483 Chr.2: 162008541-162008541 on Build GRCh38
rs749091265 Chr.2: 162008692-162008692 on Build GRCh38
rs2193680 Chr.2: 162008566-162008566 on Build GRCh38
rs867660404 Chr.2: 162008571-162008571 on Build GRCh38
rs200430209 Chr.2: 162008646-162008646 on Build GRCh38
rs780614822 Chr.2: 162008658-162008658 on Build GRCh38
rs925646771 Chr.2: 162008635-162008635 on Build GRCh38

View Full Product Details