Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21691502-21691878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
UniGene
Hs.408336
Gene Chromosome Location
Chr.16: 21663857-21771468 on Build GRCh38
Transcripts
NM_001161683.1 NM_144672.3 XM_011545747.2 XM_017022950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5429891 c.636-1G>C p.? Chr.16: 21691583-21691583 on Build GRCh38
3969449 c.707C>G p.T236S Chr.16: 21691655-21691655 on Build GRCh38
3817487 c.679T>G p.S227A Chr.16: 21691627-21691627 on Build GRCh38
6464270 c.636-1G>T p.? Chr.16: 21691583-21691583 on Build GRCh38
6016080 c.739+7C>T p.? Chr.16: 21691694-21691694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767255026 Chr.16: 21691690-21691690 on Build GRCh38
rs776800144 Chr.16: 21691754-21691754 on Build GRCh38
rs775043088 Chr.16: 21691737-21691737 on Build GRCh38
rs748400139 Chr.16: 21691617-21691617 on Build GRCh38
rs751618666 Chr.16: 21691682-21691682 on Build GRCh38
rs757210776 Chr.16: 21691658-21691658 on Build GRCh38
rs1015287713 Chr.16: 21691844-21691844 on Build GRCh38
rs997472075 Chr.16: 21691850-21691850 on Build GRCh38
rs780311907 Chr.16: 21691678-21691678 on Build GRCh38
rs758589557 Chr.16: 21691569-21691569 on Build GRCh38
rs150595232 Chr.16: 21691628-21691628 on Build GRCh38
rs757057281 Chr.16: 21691846-21691846 on Build GRCh38
rs374990400 Chr.16: 21691720-21691720 on Build GRCh38
rs1053519576 Chr.16: 21691611-21691611 on Build GRCh38
rs777515522 Chr.16: 21691564-21691564 on Build GRCh38
rs945820630 Chr.16: 21691679-21691679 on Build GRCh38
rs765872605 Chr.16: 21691539-21691539 on Build GRCh38
rs763916468 Chr.16: 21691646-21691646 on Build GRCh38
rs756083859 Chr.16: 21691562-21691562 on Build GRCh38
rs771391776 Chr.16: 21691727-21691727 on Build GRCh38
rs753873826 Chr.16: 21691656-21691656 on Build GRCh38
rs1004361064 Chr.16: 21691836-21691836 on Build GRCh38
rs139292090 Chr.16: 21691608-21691608 on Build GRCh38
rs770480396 Chr.16: 21691624-21691624 on Build GRCh38
rs766851275 Chr.16: 21691632-21691632 on Build GRCh38
rs781323344 Chr.16: 21691693-21691693 on Build GRCh38
rs763421948 Chr.16: 21691621-21691621 on Build GRCh38
rs142693996 Chr.16: 21691609-21691609 on Build GRCh38
rs773855539 Chr.16: 21691728-21691728 on Build GRCh38
rs760446852 Chr.16: 21691645-21691645 on Build GRCh38
rs116395625 Chr.16: 21691549-21691549 on Build GRCh38
rs375972309 Chr.16: 21691620-21691620 on Build GRCh38
rs762505816 Chr.16: 21691537-21691537 on Build GRCh38
rs759683702 Chr.16: 21691593-21691593 on Build GRCh38
rs1056841301 Chr.16: 21691639-21691639 on Build GRCh38
rs751255122 Chr.16: 21691543-21691543 on Build GRCh38
rs926553771 Chr.16: 21691855-21691855 on Build GRCh38
rs778179657 Chr.16: 21691711-21691711 on Build GRCh38
rs900062651 Chr.16: 21691756-21691756 on Build GRCh38
rs759218668 Chr.16: 21691631-21691631 on Build GRCh38
rs752551276 Chr.16: 21691642-21691642 on Build GRCh38
rs771599502 Chr.16: 21691732-21691732 on Build GRCh38
rs1027122023 Chr.16: 21691787-21691787 on Build GRCh38
rs781620079 Chr.16: 21691612-21691612 on Build GRCh38
rs186092865 Chr.16: 21691698-21691698 on Build GRCh38
rs981717906 Chr.16: 21691853-21691853 on Build GRCh38
rs770098432 Chr.16: 21691701-21691701 on Build GRCh38
rs758989785 Chr.16: 21691731-21691731 on Build GRCh38
rs1006781580 Chr.16: 21691812-21691812 on Build GRCh38
rs199709157 Chr.16: 21691584-21691584 on Build GRCh38
rs753807547 Chr.16: 21691568-21691568 on Build GRCh38
rs770178828 Chr.16: 21691619-21691619 on Build GRCh38
rs767326746 Chr.16: 21691552-21691552 on Build GRCh38
rs997030855 Chr.16: 21691782-21691782 on Build GRCh38
rs139606123 Chr.16: 21691683-21691683 on Build GRCh38
rs1039584147 Chr.16: 21691813-21691813 on Build GRCh38
rs752395490 Chr.16: 21691557-21691557 on Build GRCh38

View Full Product Details