Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42211816-42212314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT5B
Entrez Gene ID
6777
Gene Name
signal transducer and activator of transcription 5B
Gene Aliases
STAT5
UniGene
Hs.595276
Gene Chromosome Location
Chr.17: 42199177-42276460 on Build GRCh38
Transcripts
NM_012448.3 XM_017024977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6967094 c.1615A>G p.N539D Chr.17: 42212049-42212049 on Build GRCh38
1710303 c.1655C>T p.S552F Chr.17: 42212009-42212009 on Build GRCh38
6407895 c.1648G>T p.G550C Chr.17: 42212016-42212016 on Build GRCh38
6964009 c.1474-2A>G p.? Chr.17: 42212192-42212192 on Build GRCh38
6005723 c.1529C>T p.A510V Chr.17: 42212135-42212135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951963710 Chr.17: 42212033-42212033 on Build GRCh38
rs761733940 Chr.17: 42212040-42212040 on Build GRCh38
rs756489521 Chr.17: 42211961-42211961 on Build GRCh38
rs139437059 Chr.17: 42212020-42212020 on Build GRCh38
rs1011790676 Chr.17: 42211876-42211876 on Build GRCh38
rs143172354 Chr.17: 42212095-42212095 on Build GRCh38
rs761303284 Chr.17: 42212074-42212074 on Build GRCh38
rs960461125 Chr.17: 42212230-42212230 on Build GRCh38
rs147581021 Chr.17: 42211841-42211841 on Build GRCh38
rs565566347 Chr.17: 42211873-42211873 on Build GRCh38
rs773473797 Chr.17: 42212194-42212194 on Build GRCh38
rs946937022 Chr.17: 42211928-42211928 on Build GRCh38
rs528130301 Chr.17: 42211918-42211918 on Build GRCh38
rs769636293 Chr.17: 42212149-42212149 on Build GRCh38
rs200200711 Chr.17: 42212135-42212135 on Build GRCh38
rs140437438 Chr.17: 42211927-42211927 on Build GRCh38
rs768821055 Chr.17: 42212028-42212028 on Build GRCh38
rs142918872 Chr.17: 42212073-42212073 on Build GRCh38
rs1016642236 Chr.17: 42212156-42212156 on Build GRCh38
rs936224520 Chr.17: 42212247-42212247 on Build GRCh38
rs140632704 Chr.17: 42212083-42212083 on Build GRCh38
rs373036522 Chr.17: 42212113-42212113 on Build GRCh38
rs771406626 Chr.17: 42212201-42212201 on Build GRCh38
rs764726823 Chr.17: 42212099-42212099 on Build GRCh38
rs752406003 Chr.17: 42212101-42212101 on Build GRCh38
rs953945138 Chr.17: 42212100-42212100 on Build GRCh38
rs777292743 Chr.17: 42212110-42212110 on Build GRCh38
rs916710604 Chr.17: 42212266-42212266 on Build GRCh38
rs753497612 Chr.17: 42211969-42211969 on Build GRCh38
rs749508222 Chr.17: 42212027-42212027 on Build GRCh38
rs777198405 Chr.17: 42212214-42212214 on Build GRCh38
rs766828492 Chr.17: 42212081-42212081 on Build GRCh38
rs372815322 Chr.17: 42211962-42211962 on Build GRCh38
rs754634807 Chr.17: 42211974-42211974 on Build GRCh38
rs753274920 Chr.17: 42212231-42212231 on Build GRCh38
rs748404440 Chr.17: 42212172-42212172 on Build GRCh38
rs761250147 Chr.17: 42212200-42212200 on Build GRCh38
rs972531539 Chr.17: 42212224-42212224 on Build GRCh38
rs772239606 Chr.17: 42212173-42212173 on Build GRCh38
rs146678334 Chr.17: 42212162-42212162 on Build GRCh38
rs375789837 Chr.17: 42212060-42212060 on Build GRCh38
rs990957485 Chr.17: 42212243-42212243 on Build GRCh38
rs777627496 Chr.17: 42211998-42211998 on Build GRCh38
rs778265187 Chr.17: 42212183-42212183 on Build GRCh38
rs781030237 Chr.17: 42212134-42212134 on Build GRCh38
rs939824870 Chr.17: 42211869-42211869 on Build GRCh38
rs547984168 Chr.17: 42211985-42211985 on Build GRCh38
rs559125581 Chr.17: 42211926-42211926 on Build GRCh38
rs182744930 Chr.17: 42211874-42211874 on Build GRCh38
rs569804189 Chr.17: 42212031-42212031 on Build GRCh38
rs747677045 Chr.17: 42211981-42211981 on Build GRCh38
rs758513855 Chr.17: 42212008-42212008 on Build GRCh38
rs1043713070 Chr.17: 42211939-42211939 on Build GRCh38
rs778469957 Chr.17: 42211977-42211977 on Build GRCh38
rs1019434255 Chr.17: 42211984-42211984 on Build GRCh38
rs750971483 Chr.17: 42211934-42211934 on Build GRCh38
rs776333900 Chr.17: 42212026-42212026 on Build GRCh38
rs765654328 Chr.17: 42212229-42212229 on Build GRCh38
rs547374958 Chr.17: 42212295-42212295 on Build GRCh38
rs757850017 Chr.17: 42212109-42212109 on Build GRCh38
rs1041889535 Chr.17: 42211898-42211898 on Build GRCh38
rs151102468 Chr.17: 42212265-42212265 on Build GRCh38
rs369259420 Chr.17: 42212228-42212228 on Build GRCh38

View Full Product Details