Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 137880699-137881181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOT
Entrez Gene ID
9499
Gene Name
myotilin
Gene Aliases
LGMD1, LGMD1A, MFM3, TTID, TTOD
UniGene
Hs.84665
Gene Chromosome Location
Chr.5: 137867282-137887851 on Build GRCh38
Transcripts
NM_001135940.1 NM_001300911.1 NM_006790.2 XM_017010060.1 XM_017010061.1 XM_017010062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6546503 c.653C>T p.A218V Chr.5: 137880835-137880835 on Build GRCh38
1619505 c.656G>A p.R219Q Chr.5: 137880838-137880838 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761266355 Chr.5: 137880802-137880802 on Build GRCh38
rs577720504 Chr.5: 137881105-137881105 on Build GRCh38
rs780269433 Chr.5: 137880792-137880794 on Build GRCh38
rs775405759 Chr.5: 137880842-137880842 on Build GRCh38
rs756397543 Chr.5: 137880779-137880779 on Build GRCh38
rs752632356 Chr.5: 137880923-137880924 on Build GRCh38
rs777234976 Chr.5: 137880891-137880891 on Build GRCh38
rs907150566 Chr.5: 137881118-137881118 on Build GRCh38
rs779574125 Chr.5: 137881022-137881022 on Build GRCh38
rs768353951 Chr.5: 137880859-137880859 on Build GRCh38
rs573678193 Chr.5: 137881023-137881023 on Build GRCh38
rs201494400 Chr.5: 137881090-137881090 on Build GRCh38
rs115598221 Chr.5: 137880801-137880801 on Build GRCh38
rs776296887 Chr.5: 137880778-137880778 on Build GRCh38
rs909280353 Chr.5: 137880981-137880981 on Build GRCh38
rs923536803 Chr.5: 137880911-137880911 on Build GRCh38
rs948567231 Chr.5: 137881087-137881087 on Build GRCh38
rs545347602 Chr.5: 137881157-137881157 on Build GRCh38
rs752962882 Chr.5: 137880831-137880831 on Build GRCh38
rs71578936 Chr.5: 137880727-137880727 on Build GRCh38
rs745792335 Chr.5: 137880838-137880838 on Build GRCh38
rs747808820 Chr.5: 137880799-137880803 on Build GRCh38
rs775014749 Chr.5: 137880859-137880862 on Build GRCh38
rs760217241 Chr.5: 137880868-137880868 on Build GRCh38
rs762666087 Chr.5: 137880797-137880797 on Build GRCh38
rs1039053577 Chr.5: 137880882-137880882 on Build GRCh38
rs374136468 Chr.5: 137880950-137880950 on Build GRCh38
rs754283113 Chr.5: 137880821-137880821 on Build GRCh38
rs748974656 Chr.5: 137880848-137880848 on Build GRCh38
rs758565747 Chr.5: 137880832-137880832 on Build GRCh38
rs766928007 Chr.5: 137880874-137880874 on Build GRCh38
rs112495924 Chr.5: 137880813-137880813 on Build GRCh38
rs763748887 Chr.5: 137880798-137880798 on Build GRCh38
rs761463082 Chr.5: 137880872-137880872 on Build GRCh38
rs752826238 Chr.5: 137880742-137880742 on Build GRCh38
rs537687347 Chr.5: 137881046-137881046 on Build GRCh38
rs769466850 Chr.5: 137880782-137880782 on Build GRCh38
rs759093150 Chr.5: 137880902-137880905 on Build GRCh38
rs368115567 Chr.5: 137880784-137880784 on Build GRCh38
rs749732000 Chr.5: 137880794-137880797 on Build GRCh38
rs556333322 Chr.5: 137881055-137881055 on Build GRCh38
rs773718498 Chr.5: 137881044-137881045 on Build GRCh38
rs929274580 Chr.5: 137880953-137880953 on Build GRCh38
rs957169726 Chr.5: 137880824-137880824 on Build GRCh38
rs566153061 Chr.5: 137880809-137880809 on Build GRCh38
rs934469446 Chr.5: 137880795-137880795 on Build GRCh38
rs533510304 Chr.5: 137880835-137880835 on Build GRCh38
rs781249546 Chr.5: 137880837-137880837 on Build GRCh38
rs757882238 Chr.5: 137881007-137881007 on Build GRCh38
rs779115239 Chr.5: 137880828-137880828 on Build GRCh38
rs186960367 Chr.5: 137881013-137881013 on Build GRCh38
rs776251083 Chr.5: 137880892-137880893 on Build GRCh38
rs770530872 Chr.5: 137880867-137880868 on Build GRCh38
rs781358498 Chr.5: 137880729-137880729 on Build GRCh38
rs142477496 Chr.5: 137880836-137880836 on Build GRCh38
rs983501027 Chr.5: 137880962-137880962 on Build GRCh38
rs990723283 Chr.5: 137880825-137880825 on Build GRCh38
rs976303663 Chr.5: 137880890-137880890 on Build GRCh38
rs769674891 Chr.5: 137880840-137880840 on Build GRCh38
rs886044029 Chr.5: 137880802-137880806 on Build GRCh38

View Full Product Details