Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235696790-235697204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYST
Entrez Gene ID
1130
Gene Name
lysosomal trafficking regulator
Gene Aliases
CHS, CHS1
UniGene
Hs.532411
Gene Chromosome Location
Chr.1: 235661031-235883708 on Build GRCh38
Transcripts
NM_000081.3 NM_001301365.1 XM_011544031.1 XM_011544032.1 XM_011544033.2 XM_011544034.1 XM_011544036.2 XM_017000150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3485548 c.10557G>A p.K3519K Chr.1: 235697090-235697090 on Build GRCh38
2000175 c.10467C>T p.H3489H Chr.1: 235697180-235697180 on Build GRCh38
372590 c.10480G>T p.G3494C Chr.1: 235697167-235697167 on Build GRCh38
5380153 c.10464C>T p.P3488P Chr.1: 235697183-235697183 on Build GRCh38
5380152 c.10465C>T p.H3489Y Chr.1: 235697182-235697182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755381657 Chr.1: 235697087-235697087 on Build GRCh38
rs200511787 Chr.1: 235697170-235697170 on Build GRCh38
rs773170507 Chr.1: 235697180-235697180 on Build GRCh38
rs373529106 Chr.1: 235697179-235697179 on Build GRCh38
rs1025150478 Chr.1: 235696988-235696988 on Build GRCh38
rs756551414 Chr.1: 235697097-235697097 on Build GRCh38
rs150222948 Chr.1: 235697177-235697177 on Build GRCh38
rs1021856814 Chr.1: 235696951-235696951 on Build GRCh38
rs950440861 Chr.1: 235696933-235696933 on Build GRCh38
rs745483295 Chr.1: 235697122-235697122 on Build GRCh38
rs890229523 Chr.1: 235696856-235696856 on Build GRCh38
rs943185330 Chr.1: 235697147-235697147 on Build GRCh38
rs752990332 Chr.1: 235697094-235697094 on Build GRCh38
rs997023012 Chr.1: 235696930-235696930 on Build GRCh38
rs758070828 Chr.1: 235697133-235697133 on Build GRCh38
rs746536848 Chr.1: 235697163-235697163 on Build GRCh38
rs750720378 Chr.1: 235697051-235697051 on Build GRCh38
rs1047401204 Chr.1: 235696829-235696829 on Build GRCh38
rs201576299 Chr.1: 235697169-235697169 on Build GRCh38
rs761731196 Chr.1: 235697045-235697045 on Build GRCh38
rs761761594 Chr.1: 235697154-235697154 on Build GRCh38
rs144619475 Chr.1: 235696890-235696890 on Build GRCh38
rs920490616 Chr.1: 235696827-235696827 on Build GRCh38
rs993355287 Chr.1: 235697036-235697036 on Build GRCh38
rs900033927 Chr.1: 235696929-235696929 on Build GRCh38
rs577606781 Chr.1: 235697156-235697156 on Build GRCh38
rs187342486 Chr.1: 235697066-235697066 on Build GRCh38
rs998830275 Chr.1: 235696984-235696984 on Build GRCh38
rs960157920 Chr.1: 235697019-235697019 on Build GRCh38
rs373195142 Chr.1: 235697065-235697065 on Build GRCh38
rs970317620 Chr.1: 235697041-235697041 on Build GRCh38
rs767424814 Chr.1: 235697183-235697183 on Build GRCh38
rs945464331 Chr.1: 235697100-235697100 on Build GRCh38
rs779484570 Chr.1: 235697144-235697144 on Build GRCh38
rs781724659 Chr.1: 235697092-235697092 on Build GRCh38
rs929186143 Chr.1: 235697076-235697076 on Build GRCh38
rs138936105 Chr.1: 235697121-235697121 on Build GRCh38
rs751710591 Chr.1: 235697071-235697071 on Build GRCh38
rs926848225 Chr.1: 235697029-235697029 on Build GRCh38
rs768655106 Chr.1: 235697038-235697038 on Build GRCh38
rs556548776 Chr.1: 235696906-235696906 on Build GRCh38
rs542505499 Chr.1: 235696956-235696956 on Build GRCh38
rs920523160 Chr.1: 235697070-235697070 on Build GRCh38
rs765360919 Chr.1: 235697049-235697049 on Build GRCh38
rs928983426 Chr.1: 235696828-235696828 on Build GRCh38
rs13374850 Chr.1: 235697004-235697004 on Build GRCh38
rs574905913 Chr.1: 235696918-235696918 on Build GRCh38
rs7541057 Chr.1: 235697040-235697040 on Build GRCh38
rs546420201 Chr.1: 235697044-235697044 on Build GRCh38
rs994894525 Chr.1: 235696913-235696914 on Build GRCh38
rs763216711 Chr.1: 235697181-235697181 on Build GRCh38
rs970862282 Chr.1: 235696996-235696996 on Build GRCh38

View Full Product Details