Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61515732-61516321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT7
Entrez Gene ID
9066
Gene Name
synaptotagmin 7
Gene Aliases
IPCA-7, IPCA7, PCANAP7, SYT-VII, SYTVII
UniGene
Hs.131188
Gene Chromosome Location
Chr.11: 61513716-61588404 on Build GRCh38
Transcripts
NM_001252065.1 NM_001300773.1 NM_004200.3 XM_005274383.4 XM_005274384.2 XM_005274385.4 XM_005274387.4 XM_005274390.4 XM_006718736.3 XM_011545335.2 XM_011545336.2 XM_011545337.2 XM_011545338.2 XM_011545339.2 XM_011545340.2 XM_011545341.2 XM_011545343.2

SNP Detail

SNP ID Chromosome Location
rs755883736 Chr.11: 61516195-61516195 on Build GRCh38
rs938134268 Chr.11: 61515980-61515980 on Build GRCh38
rs560348640 Chr.11: 61516212-61516212 on Build GRCh38
rs936502572 Chr.11: 61516206-61516206 on Build GRCh38
rs961039886 Chr.11: 61515831-61515831 on Build GRCh38
rs56374633 Chr.11: 61515773-61515773 on Build GRCh38
rs376084539 Chr.11: 61515816-61515816 on Build GRCh38
rs565114907 Chr.11: 61516198-61516198 on Build GRCh38
rs889538460 Chr.11: 61516219-61516219 on Build GRCh38
rs562693516 Chr.11: 61516282-61516282 on Build GRCh38
rs923193243 Chr.11: 61515779-61515779 on Build GRCh38
rs551883405 Chr.11: 61516293-61516293 on Build GRCh38
rs1007883784 Chr.11: 61515786-61515786 on Build GRCh38
rs201841247 Chr.11: 61516164-61516165 on Build GRCh38
rs952229452 Chr.11: 61515933-61515933 on Build GRCh38
rs977226242 Chr.11: 61516136-61516136 on Build GRCh38
rs1000615413 Chr.11: 61516016-61516016 on Build GRCh38
rs1042304351 Chr.11: 61516163-61516163 on Build GRCh38
rs983804697 Chr.11: 61516174-61516174 on Build GRCh38
rs577602477 Chr.11: 61516084-61516084 on Build GRCh38
rs1010467969 Chr.11: 61515931-61515931 on Build GRCh38
rs964338659 Chr.11: 61516080-61516080 on Build GRCh38
rs975761369 Chr.11: 61516119-61516119 on Build GRCh38
rs909038897 Chr.11: 61516211-61516211 on Build GRCh38
rs1047615671 Chr.11: 61515819-61515819 on Build GRCh38
rs909064807 Chr.11: 61515846-61515846 on Build GRCh38
rs536076377 Chr.11: 61515836-61515836 on Build GRCh38
rs574960974 Chr.11: 61515784-61515784 on Build GRCh38
rs1026544252 Chr.11: 61515847-61515847 on Build GRCh38
rs892064340 Chr.11: 61515913-61515913 on Build GRCh38
rs372626255 Chr.11: 61515771-61515771 on Build GRCh38
rs1019276928 Chr.11: 61515805-61515805 on Build GRCh38
rs536403468 Chr.11: 61515764-61515764 on Build GRCh38
rs1044909564 Chr.11: 61515865-61515865 on Build GRCh38
rs556350593 Chr.11: 61515780-61515780 on Build GRCh38
rs1028052381 Chr.11: 61515772-61515772 on Build GRCh38
rs750199175 Chr.11: 61516049-61516049 on Build GRCh38
rs891826255 Chr.11: 61516268-61516270 on Build GRCh38
rs145169218 Chr.11: 61516230-61516230 on Build GRCh38
rs188500806 Chr.11: 61516226-61516226 on Build GRCh38
rs758709057 Chr.11: 61516276-61516276 on Build GRCh38
rs1040787250 Chr.11: 61516227-61516227 on Build GRCh38
rs950597785 Chr.11: 61516172-61516172 on Build GRCh38
rs1002832541 Chr.11: 61515878-61515878 on Build GRCh38
rs910209801 Chr.11: 61515768-61515768 on Build GRCh38
rs868423956 Chr.11: 61515793-61515793 on Build GRCh38
rs551369214 Chr.11: 61516026-61516026 on Build GRCh38
rs993819004 Chr.11: 61516253-61516253 on Build GRCh38
rs141187033 Chr.11: 61515872-61515872 on Build GRCh38
rs184224343 Chr.11: 61515956-61515956 on Build GRCh38
rs975956276 Chr.11: 61515775-61515775 on Build GRCh38
rs972262207 Chr.11: 61515841-61515841 on Build GRCh38
rs945372451 Chr.11: 61516106-61516106 on Build GRCh38
rs985394202 Chr.11: 61515939-61515939 on Build GRCh38
rs1057041415 Chr.11: 61516259-61516259 on Build GRCh38
rs1057286564 Chr.11: 61515890-61515890 on Build GRCh38
rs540147323 Chr.11: 61516169-61516169 on Build GRCh38

View Full Product Details