Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40463610-40464204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
UniGene
Hs.22109
Gene Chromosome Location
Chr.15: 40439721-40468242 on Build GRCh38
Transcripts
NM_001301132.1 NM_014952.4 NR_125372.1 XM_011521366.2 XM_011521367.2 XM_011521368.2 XM_011521369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629525 c.1873A>G p.T625A Chr.15: 40463918-40463918 on Build GRCh38
238935 c.1894G>A p.V632I Chr.15: 40463939-40463939 on Build GRCh38
4054371 c.1968C>T p.P656P Chr.15: 40464013-40464013 on Build GRCh38
6332786 c.1849C>T p.Q617* Chr.15: 40463894-40463894 on Build GRCh38
7169886 c.1956C>G p.L652L Chr.15: 40464001-40464001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541613586 Chr.15: 40463790-40463790 on Build GRCh38
rs568365877 Chr.15: 40464126-40464126 on Build GRCh38
rs74011157 Chr.15: 40463664-40463664 on Build GRCh38
rs901928629 Chr.15: 40463680-40463680 on Build GRCh38
rs1041751043 Chr.15: 40463668-40463668 on Build GRCh38
rs199640484 Chr.15: 40463899-40463899 on Build GRCh38
rs1046588607 Chr.15: 40464124-40464124 on Build GRCh38
rs1034024149 Chr.15: 40464152-40464152 on Build GRCh38
rs547843269 Chr.15: 40464125-40464125 on Build GRCh38
rs993657980 Chr.15: 40463776-40463776 on Build GRCh38
rs993690946 Chr.15: 40464109-40464109 on Build GRCh38
rs368995400 Chr.15: 40464041-40464041 on Build GRCh38
rs796686286 Chr.15: 40463713-40463713 on Build GRCh38
rs759654273 Chr.15: 40464013-40464013 on Build GRCh38
rs1002897614 Chr.15: 40464164-40464165 on Build GRCh38
rs751957418 Chr.15: 40463848-40463848 on Build GRCh38
rs766986121 Chr.15: 40463998-40463998 on Build GRCh38
rs778939650 Chr.15: 40463886-40463886 on Build GRCh38
rs768299856 Chr.15: 40464047-40464047 on Build GRCh38
rs765166730 Chr.15: 40463930-40463930 on Build GRCh38
rs761988826 Chr.15: 40463925-40463925 on Build GRCh38
rs923384929 Chr.15: 40463997-40463997 on Build GRCh38
rs749932489 Chr.15: 40464045-40464045 on Build GRCh38
rs971119373 Chr.15: 40463804-40463804 on Build GRCh38
rs778497440 Chr.15: 40463690-40463690 on Build GRCh38
rs551902335 Chr.15: 40463896-40463896 on Build GRCh38
rs758841269 Chr.15: 40463844-40463844 on Build GRCh38
rs371867016 Chr.15: 40463856-40463856 on Build GRCh38
rs138254188 Chr.15: 40464130-40464130 on Build GRCh38
rs976481347 Chr.15: 40464112-40464112 on Build GRCh38
rs768973571 Chr.15: 40463914-40463914 on Build GRCh38
rs750858648 Chr.15: 40463839-40463839 on Build GRCh38
rs756793845 Chr.15: 40464036-40464036 on Build GRCh38
rs760947937 Chr.15: 40463814-40463814 on Build GRCh38
rs73389007 Chr.15: 40463714-40463714 on Build GRCh38
rs746914141 Chr.15: 40463910-40463910 on Build GRCh38
rs770041650 Chr.15: 40464069-40464069 on Build GRCh38
rs776829428 Chr.15: 40464052-40464052 on Build GRCh38
rs1041984028 Chr.15: 40464153-40464153 on Build GRCh38
rs62018029 Chr.15: 40463753-40463753 on Build GRCh38
rs1025437129 Chr.15: 40463777-40463777 on Build GRCh38
rs932131578 Chr.15: 40464113-40464114 on Build GRCh38
rs1050654464 Chr.15: 40463806-40463806 on Build GRCh38
rs947594301 Chr.15: 40464147-40464147 on Build GRCh38
rs990012428 Chr.15: 40463658-40463658 on Build GRCh38
rs370186242 Chr.15: 40464179-40464179 on Build GRCh38
rs765799624 Chr.15: 40463835-40463835 on Build GRCh38
rs773489978 Chr.15: 40464071-40464071 on Build GRCh38
rs368432448 Chr.15: 40463816-40463816 on Build GRCh38
rs773872277 Chr.15: 40463938-40463938 on Build GRCh38
rs1012022530 Chr.15: 40463633-40463633 on Build GRCh38
rs776671654 Chr.15: 40463920-40463920 on Build GRCh38
rs903194592 Chr.15: 40463901-40463901 on Build GRCh38
rs777256504 Chr.15: 40463868-40463868 on Build GRCh38
rs958583579 Chr.15: 40464176-40464176 on Build GRCh38
rs140705656 Chr.15: 40463667-40463667 on Build GRCh38
rs898073527 Chr.15: 40464073-40464073 on Build GRCh38
rs763934576 Chr.15: 40463813-40463814 on Build GRCh38
rs201945733 Chr.15: 40463889-40463889 on Build GRCh38
rs761900493 Chr.15: 40463728-40463749 on Build GRCh38
rs923410790 Chr.15: 40463766-40463766 on Build GRCh38
rs73389005 Chr.15: 40463645-40463645 on Build GRCh38
rs1050623447 Chr.15: 40464115-40464115 on Build GRCh38
rs766788912 Chr.15: 40463847-40463847 on Build GRCh38
rs779945165 Chr.15: 40464046-40464046 on Build GRCh38
rs866089509 Chr.15: 40464092-40464092 on Build GRCh38
rs564303133 Chr.15: 40463884-40463884 on Build GRCh38
rs761932531 Chr.15: 40464096-40464096 on Build GRCh38
rs755371846 Chr.15: 40464011-40464011 on Build GRCh38
rs761780873 Chr.15: 40463826-40463826 on Build GRCh38
rs201224593 Chr.15: 40464048-40464048 on Build GRCh38
rs375723003 Chr.15: 40464014-40464014 on Build GRCh38
rs745669222 Chr.15: 40463693-40463693 on Build GRCh38
rs576533675 Chr.15: 40463684-40463684 on Build GRCh38
rs371439925 Chr.15: 40463907-40463907 on Build GRCh38
rs780322888 Chr.15: 40464183-40464183 on Build GRCh38
rs889076303 Chr.15: 40464044-40464044 on Build GRCh38
rs1011235637 Chr.15: 40464178-40464178 on Build GRCh38
rs763609859 Chr.15: 40463939-40463939 on Build GRCh38
rs990436832 Chr.15: 40463869-40463869 on Build GRCh38
rs140236027 Chr.15: 40464010-40464010 on Build GRCh38
rs562419625 Chr.15: 40463717-40463717 on Build GRCh38
rs776813467 Chr.15: 40463825-40463825 on Build GRCh38
rs903156446 Chr.15: 40464156-40464156 on Build GRCh38
rs982888375 Chr.15: 40463724-40463725 on Build GRCh38
rs908283623 Chr.15: 40463725-40463725 on Build GRCh38
rs932162697 Chr.15: 40463785-40463785 on Build GRCh38
rs1002600569 Chr.15: 40464145-40464145 on Build GRCh38
rs372507246 Chr.15: 40464039-40464039 on Build GRCh38
rs1038392565 Chr.15: 40464058-40464058 on Build GRCh38
rs374555410 Chr.15: 40463811-40463811 on Build GRCh38
rs576830948 Chr.15: 40463757-40463757 on Build GRCh38

View Full Product Details