Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75342340-75342935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCR
Entrez Gene ID
3156
Gene Name
3-hydroxy-3-methylglutaryl-CoA reductase
Gene Aliases
LDLCQ3
UniGene
Hs.628096
Gene Chromosome Location
Chr.5: 75336334-75362101 on Build GRCh38
Transcripts
NM_000859.2 NM_001130996.1 XM_011543357.1 XM_011543358.1 XM_011543359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754330 c.19C>G p.R7G Chr.5: 75342624-75342624 on Build GRCh38
1730758 c.145G>T p.E49* Chr.5: 75342750-75342750 on Build GRCh38
5081178 c.109T>C p.F37L Chr.5: 75342714-75342714 on Build GRCh38
4955978 c.128T>G p.I43S Chr.5: 75342733-75342733 on Build GRCh38
5508587 c.14_15insT p.R7fs*34 Chr.5: 75342619-75342620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055353341 Chr.5: 75342434-75342434 on Build GRCh38
rs748446721 Chr.5: 75342843-75342843 on Build GRCh38
rs749746293 Chr.5: 75342830-75342830 on Build GRCh38
rs1047730379 Chr.5: 75342509-75342509 on Build GRCh38
rs17244778 Chr.5: 75342480-75342480 on Build GRCh38
rs1022403092 Chr.5: 75342758-75342758 on Build GRCh38
rs772612286 Chr.5: 75342848-75342848 on Build GRCh38
rs769016687 Chr.5: 75342696-75342696 on Build GRCh38
rs113949962 Chr.5: 75342608-75342608 on Build GRCh38
rs775427700 Chr.5: 75342510-75342510 on Build GRCh38
rs561136077 Chr.5: 75342382-75342382 on Build GRCh38
rs1039995871 Chr.5: 75342496-75342496 on Build GRCh38
rs372224533 Chr.5: 75342777-75342777 on Build GRCh38
rs767136822 Chr.5: 75342798-75342798 on Build GRCh38
rs773624826 Chr.5: 75342732-75342732 on Build GRCh38
rs772179314 Chr.5: 75342727-75342727 on Build GRCh38
rs751311113 Chr.5: 75342791-75342791 on Build GRCh38
rs765251670 Chr.5: 75342552-75342552 on Build GRCh38
rs367706109 Chr.5: 75342684-75342684 on Build GRCh38
rs371769368 Chr.5: 75342700-75342700 on Build GRCh38
rs764673047 Chr.5: 75342763-75342763 on Build GRCh38
rs745371345 Chr.5: 75342667-75342667 on Build GRCh38
rs566848802 Chr.5: 75342432-75342432 on Build GRCh38
rs982580473 Chr.5: 75342466-75342466 on Build GRCh38
rs545722250 Chr.5: 75342443-75342443 on Build GRCh38
rs1009595037 Chr.5: 75342709-75342709 on Build GRCh38
rs538406998 Chr.5: 75342541-75342541 on Build GRCh38
rs374951543 Chr.5: 75342557-75342557 on Build GRCh38
rs376265388 Chr.5: 75342792-75342792 on Build GRCh38
rs766602846 Chr.5: 75342740-75342740 on Build GRCh38
rs201541062 Chr.5: 75342711-75342711 on Build GRCh38
rs763611929 Chr.5: 75342646-75342646 on Build GRCh38
rs779540718 Chr.5: 75342842-75342842 on Build GRCh38
rs758178619 Chr.5: 75342583-75342583 on Build GRCh38
rs141103473 Chr.5: 75342410-75342410 on Build GRCh38
rs369987181 Chr.5: 75342753-75342753 on Build GRCh38
rs760814083 Chr.5: 75342737-75342737 on Build GRCh38
rs547050856 Chr.5: 75342423-75342423 on Build GRCh38
rs747263814 Chr.5: 75342855-75342855 on Build GRCh38
rs182458266 Chr.5: 75342522-75342522 on Build GRCh38
rs900365501 Chr.5: 75342545-75342545 on Build GRCh38
rs534202385 Chr.5: 75342885-75342885 on Build GRCh38
rs763887826 Chr.5: 75342789-75342789 on Build GRCh38
rs554858635 Chr.5: 75342818-75342818 on Build GRCh38
rs751171582 Chr.5: 75342625-75342625 on Build GRCh38
rs755445207 Chr.5: 75342831-75342831 on Build GRCh38
rs191050559 Chr.5: 75342437-75342437 on Build GRCh38
rs886518875 Chr.5: 75342516-75342516 on Build GRCh38
rs771080862 Chr.5: 75342861-75342861 on Build GRCh38
rs769051191 Chr.5: 75342555-75342556 on Build GRCh38
rs949375617 Chr.5: 75342495-75342495 on Build GRCh38
rs996427226 Chr.5: 75342602-75342602 on Build GRCh38
rs1051046202 Chr.5: 75342628-75342628 on Build GRCh38
rs941220701 Chr.5: 75342486-75342486 on Build GRCh38
rs777456432 Chr.5: 75342592-75342592 on Build GRCh38
rs773534750 Chr.5: 75342852-75342852 on Build GRCh38
rs892502039 Chr.5: 75342665-75342665 on Build GRCh38
rs779564235 Chr.5: 75342828-75342828 on Build GRCh38
rs144812438 Chr.5: 75342821-75342821 on Build GRCh38
rs780893840 Chr.5: 75342663-75342663 on Build GRCh38
rs769345462 Chr.5: 75342677-75342677 on Build GRCh38

View Full Product Details