Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154585999-154586597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGA
Entrez Gene ID
2243
Gene Name
fibrinogen alpha chain
Gene Aliases
Fib2
UniGene
Hs.351593
Gene Chromosome Location
Chr.4: 154583126-154590766 on Build GRCh38
Transcripts
NM_000508.4 NM_021871.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1284957 c.1201G>T p.G401W Chr.4: 154586228-154586228 on Build GRCh38
4785970 c.1302C>A p.V434V Chr.4: 154586127-154586127 on Build GRCh38
3696502 c.1209G>A p.A403A Chr.4: 154586220-154586220 on Build GRCh38
4924746 c.992C>A p.T331N Chr.4: 154586437-154586437 on Build GRCh38
6313291 c.1297C>A p.L433M Chr.4: 154586132-154586132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534934417 Chr.4: 154586386-154586386 on Build GRCh38
rs769422386 Chr.4: 154586330-154586330 on Build GRCh38
rs763620151 Chr.4: 154586254-154586254 on Build GRCh38
rs764255790 Chr.4: 154586429-154586429 on Build GRCh38
rs774564301 Chr.4: 154586132-154586141 on Build GRCh38
rs768140528 Chr.4: 154586223-154586223 on Build GRCh38
rs772887890 Chr.4: 154586341-154586341 on Build GRCh38
rs750763343 Chr.4: 154586049-154586049 on Build GRCh38
rs184635235 Chr.4: 154586230-154586230 on Build GRCh38
rs750564329 Chr.4: 154586510-154586510 on Build GRCh38
rs781309738 Chr.4: 154586180-154586180 on Build GRCh38
rs745804153 Chr.4: 154586310-154586310 on Build GRCh38
rs779785102 Chr.4: 154586372-154586372 on Build GRCh38
rs756998245 Chr.4: 154586539-154586539 on Build GRCh38
rs121909610 Chr.4: 154586071-154586071 on Build GRCh38
rs763510420 Chr.4: 154586088-154586088 on Build GRCh38
rs752823417 Chr.4: 154586339-154586343 on Build GRCh38
rs373276670 Chr.4: 154586241-154586241 on Build GRCh38
rs753333518 Chr.4: 154586112-154586112 on Build GRCh38
rs764131074 Chr.4: 154586359-154586359 on Build GRCh38
rs781111021 Chr.4: 154586455-154586455 on Build GRCh38
rs1033085077 Chr.4: 154586194-154586194 on Build GRCh38
rs960708731 Chr.4: 154586544-154586544 on Build GRCh38
rs760305794 Chr.4: 154586099-154586099 on Build GRCh38
rs771843535 Chr.4: 154586218-154586218 on Build GRCh38
rs759288432 Chr.4: 154586082-154586082 on Build GRCh38
rs576468194 Chr.4: 154586538-154586538 on Build GRCh38
rs141997057 Chr.4: 154586151-154586151 on Build GRCh38
rs867808266 Chr.4: 154586293-154586293 on Build GRCh38
rs774664670 Chr.4: 154586404-154586404 on Build GRCh38
rs776003123 Chr.4: 154586502-154586502 on Build GRCh38
rs202063639 Chr.4: 154586208-154586208 on Build GRCh38
rs752326694 Chr.4: 154586521-154586521 on Build GRCh38
rs957385652 Chr.4: 154586203-154586203 on Build GRCh38
rs765412352 Chr.4: 154586445-154586445 on Build GRCh38
rs982572268 Chr.4: 154586541-154586541 on Build GRCh38
rs371236168 Chr.4: 154586495-154586495 on Build GRCh38
rs138496061 Chr.4: 154586090-154586090 on Build GRCh38
rs900665797 Chr.4: 154586402-154586402 on Build GRCh38
rs759819642 Chr.4: 154586147-154586148 on Build GRCh38
rs778270823 Chr.4: 154586540-154586540 on Build GRCh38
rs757775109 Chr.4: 154586036-154586036 on Build GRCh38
rs760149256 Chr.4: 154586411-154586411 on Build GRCh38
rs751799830 Chr.4: 154586163-154586163 on Build GRCh38
rs774954357 Chr.4: 154586348-154586348 on Build GRCh38
rs779880607 Chr.4: 154586323-154586323 on Build GRCh38
rs754360283 Chr.4: 154586025-154586025 on Build GRCh38
rs776817952 Chr.4: 154586507-154586507 on Build GRCh38
rs6051 Chr.4: 154586358-154586358 on Build GRCh38
rs760868274 Chr.4: 154586421-154586421 on Build GRCh38
rs762964238 Chr.4: 154586518-154586518 on Build GRCh38
rs754059450 Chr.4: 154586430-154586430 on Build GRCh38
rs751918931 Chr.4: 154586274-154586274 on Build GRCh38
rs780354544 Chr.4: 154586056-154586056 on Build GRCh38
rs772322188 Chr.4: 154586415-154586415 on Build GRCh38
rs777590326 Chr.4: 154586381-154586381 on Build GRCh38
rs755392951 Chr.4: 154586279-154586279 on Build GRCh38
rs746647055 Chr.4: 154586221-154586221 on Build GRCh38
rs765197711 Chr.4: 154586509-154586509 on Build GRCh38
rs947709167 Chr.4: 154586572-154586572 on Build GRCh38
rs201382224 Chr.4: 154586345-154586345 on Build GRCh38
rs558935860 Chr.4: 154586085-154586085 on Build GRCh38
rs1009160284 Chr.4: 154586129-154586129 on Build GRCh38
rs369849217 Chr.4: 154586079-154586079 on Build GRCh38
rs779394316 Chr.4: 154586037-154586037 on Build GRCh38
rs374710954 Chr.4: 154586183-154586183 on Build GRCh38
rs746736351 Chr.4: 154586326-154586326 on Build GRCh38
rs200203992 Chr.4: 154586525-154586525 on Build GRCh38
rs767596060 Chr.4: 154586281-154586281 on Build GRCh38
rs530693284 Chr.4: 154586321-154586321 on Build GRCh38
rs907226575 Chr.4: 154586490-154586490 on Build GRCh38
rs747567499 Chr.4: 154586562-154586562 on Build GRCh38
rs373577582 Chr.4: 154586130-154586130 on Build GRCh38
rs747858344 Chr.4: 154586328-154586328 on Build GRCh38
rs751444190 Chr.4: 154586446-154586446 on Build GRCh38
rs780493488 Chr.4: 154586558-154586558 on Build GRCh38
rs752970242 Chr.4: 154586307-154586307 on Build GRCh38
rs757384888 Chr.4: 154586371-154586371 on Build GRCh38
rs121909615 Chr.4: 154586390-154586390 on Build GRCh38
rs766746856 Chr.4: 154586347-154586347 on Build GRCh38
rs35633886 Chr.4: 154586279-154586280 on Build GRCh38
rs766734861 Chr.4: 154586259-154586259 on Build GRCh38
rs139005577 Chr.4: 154586327-154586327 on Build GRCh38
rs369650423 Chr.4: 154586352-154586352 on Build GRCh38
rs770479433 Chr.4: 154586389-154586389 on Build GRCh38
rs868313512 Chr.4: 154586332-154586332 on Build GRCh38
rs754981956 Chr.4: 154586448-154586448 on Build GRCh38
rs117761234 Chr.4: 154586485-154586485 on Build GRCh38
rs1011614133 Chr.4: 154586482-154586482 on Build GRCh38
rs755193875 Chr.4: 154586171-154586171 on Build GRCh38
rs747770355 Chr.4: 154586376-154586376 on Build GRCh38
rs760992799 Chr.4: 154586506-154586506 on Build GRCh38
rs192689898 Chr.4: 154586184-154586184 on Build GRCh38
rs755916126 Chr.4: 154586527-154586527 on Build GRCh38
rs757904815 Chr.4: 154586556-154586556 on Build GRCh38
rs745321779 Chr.4: 154586551-154586551 on Build GRCh38
rs767571876 Chr.4: 154586210-154586211 on Build GRCh38
rs6052 Chr.4: 154586093-154586093 on Build GRCh38
rs758775703 Chr.4: 154586152-154586152 on Build GRCh38
rs1021878997 Chr.4: 154586499-154586499 on Build GRCh38
rs200607781 Chr.4: 154586140-154586140 on Build GRCh38
rs763659799 Chr.4: 154586105-154586105 on Build GRCh38
rs150696254 Chr.4: 154586245-154586245 on Build GRCh38
rs1020674911 Chr.4: 154586487-154586487 on Build GRCh38
rs778756908 Chr.4: 154586212-154586212 on Build GRCh38
rs774294357 Chr.4: 154586069-154586069 on Build GRCh38
rs145956993 Chr.4: 154586419-154586419 on Build GRCh38
rs575433349 Chr.4: 154586428-154586428 on Build GRCh38
rs752490927 Chr.4: 154586469-154586469 on Build GRCh38
rs753399163 Chr.4: 154586533-154586533 on Build GRCh38
rs748962767 Chr.4: 154586492-154586492 on Build GRCh38
rs2070031 Chr.4: 154586063-154586063 on Build GRCh38
rs977651675 Chr.4: 154586143-154586143 on Build GRCh38
rs200459889 Chr.4: 154586367-154586367 on Build GRCh38
rs772688113 Chr.4: 154586501-154586501 on Build GRCh38
rs777770166 Chr.4: 154586529-154586529 on Build GRCh38
rs757449859 Chr.4: 154586260-154586260 on Build GRCh38
rs189096541 Chr.4: 154586559-154586559 on Build GRCh38
rs756131362 Chr.4: 154586308-154586308 on Build GRCh38
rs201105899 Chr.4: 154586357-154586357 on Build GRCh38
rs780620686 Chr.4: 154586162-154586162 on Build GRCh38
rs958559004 Chr.4: 154586316-154586316 on Build GRCh38
rs781106032 Chr.4: 154586375-154586375 on Build GRCh38
rs6050 Chr.4: 154586438-154586438 on Build GRCh38
rs10921 Chr.4: 154586253-154586253 on Build GRCh38
rs751773097 Chr.4: 154586232-154586232 on Build GRCh38
rs769893639 Chr.4: 154586202-154586202 on Build GRCh38
rs758861856 Chr.4: 154586053-154586053 on Build GRCh38
rs376629642 Chr.4: 154586062-154586062 on Build GRCh38
rs775218633 Chr.4: 154586220-154586220 on Build GRCh38
rs376065633 Chr.4: 154586216-154586216 on Build GRCh38
rs770335622 Chr.4: 154586494-154586494 on Build GRCh38
rs749217129 Chr.4: 154586060-154586060 on Build GRCh38
rs373154873 Chr.4: 154586374-154586374 on Build GRCh38
rs927095063 Chr.4: 154586294-154586294 on Build GRCh38
rs916352754 Chr.4: 154586565-154586565 on Build GRCh38
rs756004130 Chr.4: 154586484-154586484 on Build GRCh38
rs760776311 Chr.4: 154586346-154586347 on Build GRCh38
rs761337850 Chr.4: 154586231-154586231 on Build GRCh38
rs781195832 Chr.4: 154586059-154586059 on Build GRCh38
rs143170281 Chr.4: 154586058-154586058 on Build GRCh38

View Full Product Details